Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100678008-100678511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB2
Entrez Gene ID
2783
Gene Name
G protein subunit beta 2
Gene Aliases
-
UniGene
Hs.185172
Gene Chromosome Location
Chr.7: 100673740-100679169 on Build GRCh38
Transcripts
NM_005273.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208409 c.682G>A p.D228N Chr.7: 100678282-100678282 on Build GRCh38
1292385 c.571T>G p.S191A Chr.7: 100678171-100678171 on Build GRCh38
4153677 c.791A>T p.Y264F Chr.7: 100678489-100678489 on Build GRCh38
4428584 c.580C>G p.P194A Chr.7: 100678180-100678180 on Build GRCh38
4654155 c.777G>A p.Q259Q Chr.7: 100678475-100678475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763156002 Chr.7: 100678321-100678321 on Build GRCh38
rs112855313 Chr.7: 100678300-100678300 on Build GRCh38
rs61758990 Chr.7: 100678452-100678452 on Build GRCh38
rs756555248 Chr.7: 100678343-100678343 on Build GRCh38
rs753114827 Chr.7: 100678333-100678333 on Build GRCh38
rs369459358 Chr.7: 100678392-100678392 on Build GRCh38
rs774089228 Chr.7: 100678354-100678355 on Build GRCh38
rs774398152 Chr.7: 100678387-100678387 on Build GRCh38
rs756394669 Chr.7: 100678237-100678237 on Build GRCh38
rs755355032 Chr.7: 100678161-100678161 on Build GRCh38
rs374877478 Chr.7: 100678275-100678275 on Build GRCh38
rs766519665 Chr.7: 100678323-100678323 on Build GRCh38
rs1001964230 Chr.7: 100678389-100678389 on Build GRCh38
rs200947399 Chr.7: 100678403-100678403 on Build GRCh38
rs74851555 Chr.7: 100678360-100678360 on Build GRCh38
rs774483254 Chr.7: 100678189-100678189 on Build GRCh38
rs374626655 Chr.7: 100678063-100678063 on Build GRCh38
rs752126417 Chr.7: 100678222-100678222 on Build GRCh38
rs1016502162 Chr.7: 100678030-100678030 on Build GRCh38
rs759487978 Chr.7: 100678330-100678330 on Build GRCh38
rs779424974 Chr.7: 100678457-100678457 on Build GRCh38
rs79631023 Chr.7: 100678183-100678183 on Build GRCh38
rs150959922 Chr.7: 100678131-100678131 on Build GRCh38
rs1038338672 Chr.7: 100678341-100678341 on Build GRCh38
rs138214277 Chr.7: 100678120-100678120 on Build GRCh38
rs775247316 Chr.7: 100678310-100678310 on Build GRCh38
rs993199714 Chr.7: 100678042-100678042 on Build GRCh38
rs139214321 Chr.7: 100678190-100678190 on Build GRCh38
rs772067269 Chr.7: 100678307-100678307 on Build GRCh38
rs781496683 Chr.7: 100678234-100678234 on Build GRCh38
rs1016962982 Chr.7: 100678101-100678101 on Build GRCh38
rs373871017 Chr.7: 100678182-100678182 on Build GRCh38
rs771248426 Chr.7: 100678048-100678049 on Build GRCh38
rs542867104 Chr.7: 100678226-100678226 on Build GRCh38
rs141859943 Chr.7: 100678179-100678179 on Build GRCh38
rs761264175 Chr.7: 100678395-100678395 on Build GRCh38
rs748396013 Chr.7: 100678176-100678176 on Build GRCh38
rs746905977 Chr.7: 100678367-100678367 on Build GRCh38
rs889272812 Chr.7: 100678272-100678272 on Build GRCh38
rs201948712 Chr.7: 100678427-100678427 on Build GRCh38
rs114089976 Chr.7: 100678346-100678346 on Build GRCh38
rs983636308 Chr.7: 100678090-100678090 on Build GRCh38
rs2272573 Chr.7: 100678415-100678415 on Build GRCh38
rs112334033 Chr.7: 100678302-100678302 on Build GRCh38
rs144309481 Chr.7: 100678469-100678469 on Build GRCh38
rs754921203 Chr.7: 100678466-100678466 on Build GRCh38
rs942240263 Chr.7: 100678335-100678335 on Build GRCh38
rs377281989 Chr.7: 100678372-100678372 on Build GRCh38
rs200210649 Chr.7: 100678206-100678206 on Build GRCh38
rs377520963 Chr.7: 100678485-100678485 on Build GRCh38
rs771831778 Chr.7: 100678193-100678193 on Build GRCh38
rs768416926 Chr.7: 100678314-100678314 on Build GRCh38
rs769501179 Chr.7: 100678177-100678177 on Build GRCh38
rs757652261 Chr.7: 100678429-100678429 on Build GRCh38
rs751674938 Chr.7: 100678327-100678327 on Build GRCh38
rs570986478 Chr.7: 100678409-100678409 on Build GRCh38
rs565561031 Chr.7: 100678045-100678045 on Build GRCh38
rs757811881 Chr.7: 100678351-100678351 on Build GRCh38
rs957854880 Chr.7: 100678430-100678430 on Build GRCh38
rs780085774 Chr.7: 100678364-100678364 on Build GRCh38
rs369552295 Chr.7: 100678375-100678375 on Build GRCh38
rs769252464 Chr.7: 100678061-100678061 on Build GRCh38
rs746067913 Chr.7: 100678486-100678486 on Build GRCh38
rs770474986 Chr.7: 100678064-100678068 on Build GRCh38
rs2406154 Chr.7: 100678146-100678146 on Build GRCh38
rs368070921 Chr.7: 100678194-100678194 on Build GRCh38
rs561037473 Chr.7: 100678355-100678360 on Build GRCh38
rs140356994 Chr.7: 100678460-100678460 on Build GRCh38
rs766442414 Chr.7: 100678070-100678070 on Build GRCh38
rs770626958 Chr.7: 100678281-100678281 on Build GRCh38
rs780578984 Chr.7: 100678134-100678134 on Build GRCh38
rs569427998 Chr.7: 100678370-100678375 on Build GRCh38
rs112857834 Chr.7: 100678397-100678397 on Build GRCh38
rs150086689 Chr.7: 100678436-100678436 on Build GRCh38
rs776413782 Chr.7: 100678373-100678373 on Build GRCh38
rs754068566 Chr.7: 100678100-100678100 on Build GRCh38
rs747822004 Chr.7: 100678060-100678060 on Build GRCh38
rs747491823 Chr.7: 100678149-100678149 on Build GRCh38
rs140950077 Chr.7: 100678031-100678031 on Build GRCh38
rs375187958 Chr.7: 100678391-100678391 on Build GRCh38
rs5001693 Chr.7: 100678209-100678209 on Build GRCh38
rs757499352 Chr.7: 100678119-100678119 on Build GRCh38
rs145101383 Chr.7: 100678089-100678089 on Build GRCh38
rs923325507 Chr.7: 100678224-100678224 on Build GRCh38
rs778564494 Chr.7: 100678293-100678293 on Build GRCh38
rs371447650 Chr.7: 100678269-100678269 on Build GRCh38
rs760554032 Chr.7: 100678313-100678313 on Build GRCh38
rs768101002 Chr.7: 100678332-100678332 on Build GRCh38
rs769547114 Chr.7: 100678470-100678470 on Build GRCh38
rs372498022 Chr.7: 100678384-100678384 on Build GRCh38
rs776126189 Chr.7: 100678393-100678393 on Build GRCh38
rs149502189 Chr.7: 100678242-100678242 on Build GRCh38
rs200586443 Chr.7: 100678094-100678094 on Build GRCh38
rs574237260 Chr.7: 100678239-100678239 on Build GRCh38
rs375676173 Chr.7: 100678419-100678419 on Build GRCh38
rs781614369 Chr.7: 100678171-100678171 on Build GRCh38
rs762496734 Chr.7: 100678069-100678069 on Build GRCh38
rs560853556 Chr.7: 100678320-100678320 on Build GRCh38
rs773258449 Chr.7: 100678059-100678062 on Build GRCh38
rs769033152 Chr.7: 100678056-100678056 on Build GRCh38
rs201331669 Chr.7: 100678055-100678055 on Build GRCh38
rs753129673 Chr.7: 100678236-100678236 on Build GRCh38
rs1050520443 Chr.7: 100678267-100678267 on Build GRCh38
rs2272574 Chr.7: 100678345-100678345 on Build GRCh38
rs769947840 Chr.7: 100678067-100678068 on Build GRCh38
rs766432973 Chr.7: 100678220-100678220 on Build GRCh38
rs373371911 Chr.7: 100678439-100678439 on Build GRCh38
rs201168795 Chr.7: 100678445-100678445 on Build GRCh38
rs771165202 Chr.7: 100678386-100678386 on Build GRCh38
rs758428726 Chr.7: 100678132-100678132 on Build GRCh38
rs376741553 Chr.7: 100678083-100678083 on Build GRCh38

View Full Product Details