Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157749319-157749840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL2
Entrez Gene ID
79368
Gene Name
Fc receptor like 2
Gene Aliases
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
UniGene
Hs.437393
Gene Chromosome Location
Chr.1: 157745733-157777472 on Build GRCh38
Transcripts
NM_001159488.1 NM_030764.3 NR_125358.1 XM_011509974.2 XM_011509975.2 XM_011509976.2 XM_017002316.1 XM_017002317.1 XM_017002318.1 XM_017002319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368020 c.1303C>T p.P435S Chr.1: 157749654-157749654 on Build GRCh38
7250054 c.1300G>T p.E434* Chr.1: 157749657-157749657 on Build GRCh38
4023554 c.1282G>T p.E428* Chr.1: 157749675-157749675 on Build GRCh38
1252324 c.1285A>C p.S429R Chr.1: 157749672-157749672 on Build GRCh38
5531167 c.1280G>A p.G427E Chr.1: 157749677-157749677 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs188058971 Chr.1: 157749690-157749690 on Build GRCh38
rs1027130079 Chr.1: 157749517-157749517 on Build GRCh38
rs891961435 Chr.1: 157749801-157749801 on Build GRCh38
rs773919103 Chr.1: 157749730-157749730 on Build GRCh38
rs146287852 Chr.1: 157749566-157749566 on Build GRCh38
rs751078395 Chr.1: 157749641-157749641 on Build GRCh38
rs551070642 Chr.1: 157749587-157749587 on Build GRCh38
rs16839038 Chr.1: 157749634-157749634 on Build GRCh38
rs77127214 Chr.1: 157749429-157749429 on Build GRCh38
rs764543898 Chr.1: 157749599-157749599 on Build GRCh38
rs567715061 Chr.1: 157749511-157749511 on Build GRCh38
rs770334300 Chr.1: 157749657-157749664 on Build GRCh38
rs754991734 Chr.1: 157749647-157749647 on Build GRCh38
rs935752752 Chr.1: 157749772-157749772 on Build GRCh38
rs530997459 Chr.1: 157749388-157749388 on Build GRCh38
rs906704791 Chr.1: 157749786-157749786 on Build GRCh38
rs1009143889 Chr.1: 157749507-157749507 on Build GRCh38
rs748115778 Chr.1: 157749623-157749623 on Build GRCh38
rs1027667550 Chr.1: 157749571-157749571 on Build GRCh38
rs961345668 Chr.1: 157749484-157749484 on Build GRCh38
rs745386434 Chr.1: 157749479-157749479 on Build GRCh38
rs906777371 Chr.1: 157749426-157749426 on Build GRCh38
rs546237441 Chr.1: 157749506-157749506 on Build GRCh38
rs530561419 Chr.1: 157749787-157749787 on Build GRCh38
rs995209218 Chr.1: 157749495-157749495 on Build GRCh38
rs746115849 Chr.1: 157749704-157749704 on Build GRCh38
rs139649563 Chr.1: 157749738-157749738 on Build GRCh38
rs375675141 Chr.1: 157749653-157749653 on Build GRCh38
rs562187995 Chr.1: 157749650-157749650 on Build GRCh38
rs1000749667 Chr.1: 157749451-157749451 on Build GRCh38
rs772118228 Chr.1: 157749725-157749725 on Build GRCh38
rs1005113480 Chr.1: 157749798-157749798 on Build GRCh38
rs765484730 Chr.1: 157749632-157749632 on Build GRCh38
rs948153739 Chr.1: 157749754-157749754 on Build GRCh38
rs769202863 Chr.1: 157749478-157749478 on Build GRCh38
rs1057232414 Chr.1: 157749366-157749366 on Build GRCh38
rs1054294326 Chr.1: 157749380-157749380 on Build GRCh38
rs1046569259 Chr.1: 157749779-157749779 on Build GRCh38
rs550850102 Chr.1: 157749509-157749509 on Build GRCh38
rs750471317 Chr.1: 157749568-157749568 on Build GRCh38
rs892072009 Chr.1: 157749500-157749500 on Build GRCh38
rs777724460 Chr.1: 157749441-157749441 on Build GRCh38
rs1012403917 Chr.1: 157749403-157749403 on Build GRCh38
rs757687668 Chr.1: 157749613-157749613 on Build GRCh38
rs895480671 Chr.1: 157749376-157749376 on Build GRCh38
rs1018942701 Chr.1: 157749539-157749539 on Build GRCh38
rs79445400 Chr.1: 157749655-157749655 on Build GRCh38
rs1053730133 Chr.1: 157749800-157749800 on Build GRCh38
rs553285523 Chr.1: 157749413-157749413 on Build GRCh38
rs533416602 Chr.1: 157749744-157749744 on Build GRCh38
rs754222558 Chr.1: 157749603-157749603 on Build GRCh38
rs541954716 Chr.1: 157749633-157749633 on Build GRCh38
rs536333378 Chr.1: 157749753-157749753 on Build GRCh38

View Full Product Details