Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157831225-157831737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD5L
Entrez Gene ID
922
Gene Name
CD5 molecule like
Gene Aliases
AIM, API6, CT-2, PRO229, SP-ALPHA, Spalpha, hAIM
UniGene
Hs.134035
Gene Chromosome Location
Chr.1: 157827071-157841844 on Build GRCh38
Transcripts
NM_005894.2 XM_005245602.1 XM_017002806.1

SNP Detail

SNP ID Chromosome Location
rs775373844 Chr.1: 157831480-157831480 on Build GRCh38
rs900900466 Chr.1: 157831552-157831552 on Build GRCh38
rs773508490 Chr.1: 157831670-157831670 on Build GRCh38
rs564289849 Chr.1: 157831434-157831434 on Build GRCh38
rs187859976 Chr.1: 157831527-157831527 on Build GRCh38
rs1045122405 Chr.1: 157831309-157831309 on Build GRCh38
rs148584519 Chr.1: 157831598-157831598 on Build GRCh38
rs917955625 Chr.1: 157831557-157831557 on Build GRCh38
rs374847728 Chr.1: 157831283-157831283 on Build GRCh38
rs112301931 Chr.1: 157831440-157831440 on Build GRCh38
rs1006530766 Chr.1: 157831526-157831526 on Build GRCh38
rs61818260 Chr.1: 157831439-157831439 on Build GRCh38
rs1054726383 Chr.1: 157831712-157831712 on Build GRCh38
rs969319591 Chr.1: 157831330-157831330 on Build GRCh38
rs930837140 Chr.1: 157831455-157831455 on Build GRCh38
rs16839250 Chr.1: 157831503-157831503 on Build GRCh38
rs16839246 Chr.1: 157831334-157831334 on Build GRCh38
rs769251618 Chr.1: 157831549-157831549 on Build GRCh38
rs909362936 Chr.1: 157831486-157831486 on Build GRCh38
rs920797606 Chr.1: 157831385-157831385 on Build GRCh38
rs1050629797 Chr.1: 157831476-157831476 on Build GRCh38
rs75445239 Chr.1: 157831452-157831453 on Build GRCh38
rs1023480333 Chr.1: 157831700-157831700 on Build GRCh38
rs749743644 Chr.1: 157831387-157831387 on Build GRCh38
rs536723216 Chr.1: 157831281-157831281 on Build GRCh38
rs1045008809 Chr.1: 157831600-157831600 on Build GRCh38
rs16839249 Chr.1: 157831351-157831351 on Build GRCh38
rs759519936 Chr.1: 157831272-157831272 on Build GRCh38
rs189680766 Chr.1: 157831323-157831323 on Build GRCh38
rs866745022 Chr.1: 157831531-157831531 on Build GRCh38
rs921688740 Chr.1: 157831440-157831441 on Build GRCh38
rs751647203 Chr.1: 157831641-157831641 on Build GRCh38
rs772300875 Chr.1: 157831594-157831594 on Build GRCh38
rs145866521 Chr.1: 157831491-157831491 on Build GRCh38
rs111770134 Chr.1: 157831663-157831663 on Build GRCh38
rs780460630 Chr.1: 157831324-157831324 on Build GRCh38
rs764286952 Chr.1: 157831561-157831561 on Build GRCh38
rs905297781 Chr.1: 157831628-157831628 on Build GRCh38
rs775833616 Chr.1: 157831441-157831441 on Build GRCh38
rs369849878 Chr.1: 157831373-157831373 on Build GRCh38
rs547627312 Chr.1: 157831710-157831710 on Build GRCh38
rs115662283 Chr.1: 157831287-157831287 on Build GRCh38
rs940795413 Chr.1: 157831352-157831352 on Build GRCh38
rs761911519 Chr.1: 157831566-157831566 on Build GRCh38
rs182904391 Chr.1: 157831337-157831337 on Build GRCh38
rs1036529633 Chr.1: 157831361-157831361 on Build GRCh38
rs957542287 Chr.1: 157831346-157831346 on Build GRCh38

View Full Product Details