Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158254996-158255502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1A
Entrez Gene ID
909
Gene Name
CD1a molecule
Gene Aliases
CD1, FCB6, HTA1, R4, T6
UniGene
Hs.1309
Gene Chromosome Location
Chr.1: 158248329-158258269 on Build GRCh38
Transcripts
NM_001320652.1 NM_001763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1688835 c.151G>A p.D51N Chr.1: 158255176-158255176 on Build GRCh38
6121589 c.269G>C p.R90P Chr.1: 158255294-158255294 on Build GRCh38
5378018 c.212C>T p.S71F Chr.1: 158255237-158255237 on Build GRCh38
3475942 c.81C>T p.F27F Chr.1: 158255106-158255106 on Build GRCh38
6863466 c.199C>A p.L67M Chr.1: 158255224-158255224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778866732 Chr.1: 158255383-158255383 on Build GRCh38
rs559573053 Chr.1: 158255202-158255202 on Build GRCh38
rs933100350 Chr.1: 158255230-158255230 on Build GRCh38
rs202190888 Chr.1: 158255235-158255235 on Build GRCh38
rs572935311 Chr.1: 158255437-158255437 on Build GRCh38
rs549161271 Chr.1: 158255220-158255220 on Build GRCh38
rs768704597 Chr.1: 158255318-158255318 on Build GRCh38
rs761707042 Chr.1: 158255089-158255089 on Build GRCh38
rs1035926182 Chr.1: 158255463-158255463 on Build GRCh38
rs192062712 Chr.1: 158255080-158255080 on Build GRCh38
rs11800924 Chr.1: 158255340-158255340 on Build GRCh38
rs977116148 Chr.1: 158255429-158255429 on Build GRCh38
rs767795358 Chr.1: 158255176-158255176 on Build GRCh38
rs764917107 Chr.1: 158255090-158255090 on Build GRCh38
rs147750753 Chr.1: 158255328-158255328 on Build GRCh38
rs776765700 Chr.1: 158255395-158255395 on Build GRCh38
rs748118650 Chr.1: 158255223-158255223 on Build GRCh38
rs150169121 Chr.1: 158255329-158255329 on Build GRCh38
rs201755458 Chr.1: 158255347-158255347 on Build GRCh38
rs2269716 Chr.1: 158255453-158255453 on Build GRCh38
rs774096922 Chr.1: 158255056-158255056 on Build GRCh38
rs1038767555 Chr.1: 158255095-158255095 on Build GRCh38
rs531484228 Chr.1: 158255072-158255072 on Build GRCh38
rs569038953 Chr.1: 158255206-158255206 on Build GRCh38
rs187686385 Chr.1: 158255352-158255352 on Build GRCh38
rs146048507 Chr.1: 158255303-158255303 on Build GRCh38
rs774994865 Chr.1: 158255126-158255126 on Build GRCh38
rs746482126 Chr.1: 158255127-158255127 on Build GRCh38
rs769695209 Chr.1: 158255149-158255149 on Build GRCh38
rs750898544 Chr.1: 158255263-158255263 on Build GRCh38
rs373705897 Chr.1: 158255100-158255100 on Build GRCh38
rs116290813 Chr.1: 158255293-158255293 on Build GRCh38
rs775908388 Chr.1: 158255236-158255236 on Build GRCh38
rs2269715 Chr.1: 158255229-158255229 on Build GRCh38
rs375586275 Chr.1: 158255054-158255054 on Build GRCh38
rs199579196 Chr.1: 158255251-158255251 on Build GRCh38
rs150885713 Chr.1: 158255315-158255315 on Build GRCh38
rs771647135 Chr.1: 158255124-158255124 on Build GRCh38
rs773077647 Chr.1: 158255231-158255231 on Build GRCh38
rs750728957 Chr.1: 158255216-158255216 on Build GRCh38
rs764303588 Chr.1: 158255246-158255246 on Build GRCh38
rs761641729 Chr.1: 158255143-158255143 on Build GRCh38
rs411089 Chr.1: 158255035-158255035 on Build GRCh38
rs780561704 Chr.1: 158255222-158255222 on Build GRCh38
rs187184165 Chr.1: 158255079-158255079 on Build GRCh38
rs765336915 Chr.1: 158255325-158255325 on Build GRCh38
rs149019370 Chr.1: 158255118-158255118 on Build GRCh38
rs767578429 Chr.1: 158255280-158255282 on Build GRCh38
rs148007865 Chr.1: 158255260-158255260 on Build GRCh38
rs765680591 Chr.1: 158255036-158255036 on Build GRCh38
rs759543827 Chr.1: 158255240-158255240 on Build GRCh38
rs990456775 Chr.1: 158255174-158255174 on Build GRCh38
rs371301715 Chr.1: 158255178-158255178 on Build GRCh38
rs762395400 Chr.1: 158255254-158255254 on Build GRCh38
rs778292155 Chr.1: 158255045-158255045 on Build GRCh38
rs768955173 Chr.1: 158255392-158255392 on Build GRCh38
rs954435984 Chr.1: 158255445-158255445 on Build GRCh38
rs745601974 Chr.1: 158255391-158255391 on Build GRCh38
rs772813755 Chr.1: 158255165-158255165 on Build GRCh38
rs138585396 Chr.1: 158255287-158255287 on Build GRCh38
rs776678576 Chr.1: 158255136-158255136 on Build GRCh38
rs776795053 Chr.1: 158255320-158255320 on Build GRCh38
rs775233831 Chr.1: 158255294-158255294 on Build GRCh38
rs780382684 Chr.1: 158255283-158255283 on Build GRCh38
rs763962809 Chr.1: 158255082-158255082 on Build GRCh38
rs755245058 Chr.1: 158255094-158255094 on Build GRCh38
rs3087218 Chr.1: 158255452-158255452 on Build GRCh38
rs768122492 Chr.1: 158255128-158255128 on Build GRCh38
rs760969425 Chr.1: 158255243-158255243 on Build GRCh38
rs759847473 Chr.1: 158255170-158255170 on Build GRCh38
rs967100894 Chr.1: 158255194-158255194 on Build GRCh38
rs754517946 Chr.1: 158255288-158255288 on Build GRCh38
rs777412961 Chr.1: 158255356-158255356 on Build GRCh38
rs140466685 Chr.1: 158255121-158255121 on Build GRCh38
rs764062414 Chr.1: 158255198-158255198 on Build GRCh38
rs754429586 Chr.1: 158255200-158255200 on Build GRCh38
rs377400670 Chr.1: 158255217-158255217 on Build GRCh38
rs1053231401 Chr.1: 158255142-158255142 on Build GRCh38
rs770910864 Chr.1: 158255304-158255304 on Build GRCh38
rs375993315 Chr.1: 158255189-158255189 on Build GRCh38
rs749731331 Chr.1: 158255116-158255116 on Build GRCh38
rs758811925 Chr.1: 158255282-158255282 on Build GRCh38
rs140904380 Chr.1: 158255122-158255122 on Build GRCh38
rs3087217 Chr.1: 158255091-158255091 on Build GRCh38
rs776321553 Chr.1: 158255085-158255085 on Build GRCh38
rs914278858 Chr.1: 158255025-158255025 on Build GRCh38
rs990232671 Chr.1: 158255021-158255021 on Build GRCh38
rs193227561 Chr.1: 158255446-158255446 on Build GRCh38
rs958942210 Chr.1: 158255133-158255133 on Build GRCh38
rs749619335 Chr.1: 158255046-158255046 on Build GRCh38
rs918892628 Chr.1: 158255059-158255059 on Build GRCh38
rs372873025 Chr.1: 158255076-158255076 on Build GRCh38
rs865995777 Chr.1: 158255207-158255207 on Build GRCh38
rs771303365 Chr.1: 158255110-158255110 on Build GRCh38
rs537550225 Chr.1: 158255218-158255218 on Build GRCh38
rs77627581 Chr.1: 158255418-158255418 on Build GRCh38
rs757196117 Chr.1: 158255368-158255368 on Build GRCh38
rs1049444427 Chr.1: 158255069-158255069 on Build GRCh38
rs538916791 Chr.1: 158255306-158255306 on Build GRCh38
rs142769781 Chr.1: 158255302-158255302 on Build GRCh38
rs2269714 Chr.1: 158255114-158255114 on Build GRCh38
rs538168250 Chr.1: 158255431-158255431 on Build GRCh38
rs752359352 Chr.1: 158255351-158255351 on Build GRCh38
rs139349958 Chr.1: 158255321-158255321 on Build GRCh38
rs745488261 Chr.1: 158255060-158255060 on Build GRCh38
rs745776518 Chr.1: 158255204-158255204 on Build GRCh38
rs373702670 Chr.1: 158255177-158255177 on Build GRCh38
rs749308248 Chr.1: 158255360-158255360 on Build GRCh38
rs866676954 Chr.1: 158255145-158255145 on Build GRCh38
rs950431908 Chr.1: 158255262-158255262 on Build GRCh38
rs1022289158 Chr.1: 158255193-158255193 on Build GRCh38

View Full Product Details