Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158353618-158354117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1E
Entrez Gene ID
913
Gene Name
CD1e molecule
Gene Aliases
CD1A, R2
UniGene
Hs.249217
Gene Chromosome Location
Chr.1: 158353157-158357554 on Build GRCh38
Transcripts
NM_001042583.2 NM_001042584.2 NM_001042585.2 NM_001042586.2 NM_001042587.2 NM_001185107.1 NM_001185108.1 NM_001185110.1 NM_001185112.1 NM_001185113.1 NM_001185114.1 NM_001185115.1 NM_030893.3 XM_011510133.2 XM_011510134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718360 c.46G>A p.E16K Chr.1: 158354034-158354034 on Build GRCh38
1335156 c.51T>C p.N17N Chr.1: 158354039-158354039 on Build GRCh38
6271380 c.27G>T p.E9D Chr.1: 158354015-158354015 on Build GRCh38
414414 c.24C>G p.F8L Chr.1: 158354012-158354012 on Build GRCh38
1247677 c.16C>A p.L6I Chr.1: 158354004-158354004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899793008 Chr.1: 158353815-158353815 on Build GRCh38
rs773356484 Chr.1: 158353967-158353967 on Build GRCh38
rs371742057 Chr.1: 158353958-158353958 on Build GRCh38
rs933963391 Chr.1: 158354095-158354095 on Build GRCh38
rs771896654 Chr.1: 158353954-158353954 on Build GRCh38
rs11809933 Chr.1: 158353820-158353820 on Build GRCh38
rs557256689 Chr.1: 158353777-158353777 on Build GRCh38
rs768143512 Chr.1: 158354089-158354089 on Build GRCh38
rs937263276 Chr.1: 158353693-158353693 on Build GRCh38
rs962947918 Chr.1: 158353665-158353665 on Build GRCh38
rs746811165 Chr.1: 158354066-158354066 on Build GRCh38
rs759632386 Chr.1: 158354087-158354087 on Build GRCh38
rs774514834 Chr.1: 158354084-158354084 on Build GRCh38
rs753516066 Chr.1: 158354013-158354013 on Build GRCh38
rs373877618 Chr.1: 158354000-158354000 on Build GRCh38
rs747930055 Chr.1: 158353801-158353801 on Build GRCh38
rs1029903730 Chr.1: 158353869-158353869 on Build GRCh38
rs770954870 Chr.1: 158354081-158354081 on Build GRCh38
rs573383371 Chr.1: 158353825-158353825 on Build GRCh38
rs989955199 Chr.1: 158353798-158353798 on Build GRCh38
rs571315266 Chr.1: 158353737-158353737 on Build GRCh38
rs982648987 Chr.1: 158354046-158354046 on Build GRCh38
rs1009757983 Chr.1: 158353897-158353897 on Build GRCh38
rs554894177 Chr.1: 158353821-158353821 on Build GRCh38
rs868718790 Chr.1: 158354034-158354034 on Build GRCh38
rs768092068 Chr.1: 158353981-158353981 on Build GRCh38
rs929858202 Chr.1: 158354009-158354009 on Build GRCh38
rs183557427 Chr.1: 158353959-158353959 on Build GRCh38
rs376989417 Chr.1: 158354012-158354012 on Build GRCh38
rs917104781 Chr.1: 158353733-158353733 on Build GRCh38
rs201300311 Chr.1: 158353989-158353989 on Build GRCh38
rs779908083 Chr.1: 158354055-158354055 on Build GRCh38
rs368524094 Chr.1: 158353719-158353719 on Build GRCh38
rs968417202 Chr.1: 158353644-158353644 on Build GRCh38
rs552211899 Chr.1: 158354024-158354024 on Build GRCh38
rs978412077 Chr.1: 158353659-158353659 on Build GRCh38
rs543003746 Chr.1: 158354023-158354023 on Build GRCh38
rs71632610 Chr.1: 158354073-158354073 on Build GRCh38
rs899866375 Chr.1: 158354093-158354093 on Build GRCh38
rs753128759 Chr.1: 158353983-158353983 on Build GRCh38
rs773359926 Chr.1: 158354075-158354075 on Build GRCh38
rs1051031519 Chr.1: 158353911-158353911 on Build GRCh38
rs926976054 Chr.1: 158353766-158353766 on Build GRCh38
rs544192630 Chr.1: 158354016-158354016 on Build GRCh38
rs11809955 Chr.1: 158353970-158353970 on Build GRCh38
rs760055201 Chr.1: 158353980-158353980 on Build GRCh38
rs994306541 Chr.1: 158354033-158354033 on Build GRCh38
rs752052168 Chr.1: 158353971-158353971 on Build GRCh38
rs971627077 Chr.1: 158354076-158354076 on Build GRCh38
rs888556374 Chr.1: 158353757-158353757 on Build GRCh38
rs937067880 Chr.1: 158353790-158353790 on Build GRCh38
rs763210901 Chr.1: 158354077-158354077 on Build GRCh38
rs763005827 Chr.1: 158353969-158353969 on Build GRCh38
rs368752831 Chr.1: 158353957-158353957 on Build GRCh38
rs1022858955 Chr.1: 158353898-158353898 on Build GRCh38
rs993182806 Chr.1: 158353978-158353978 on Build GRCh38
rs753251534 Chr.1: 158354090-158354090 on Build GRCh38
rs11805538 Chr.1: 158353943-158353943 on Build GRCh38
rs11806627 Chr.1: 158353743-158353743 on Build GRCh38
rs745332808 Chr.1: 158354019-158354019 on Build GRCh38
rs575816125 Chr.1: 158353800-158353800 on Build GRCh38
rs957118468 Chr.1: 158353892-158353892 on Build GRCh38
rs747031931 Chr.1: 158353941-158353941 on Build GRCh38
rs3180089 Chr.1: 158354032-158354032 on Build GRCh38
rs756767963 Chr.1: 158354015-158354015 on Build GRCh38
rs1056974831 Chr.1: 158353709-158353709 on Build GRCh38
rs926928682 Chr.1: 158353669-158353669 on Build GRCh38
rs548896452 Chr.1: 158353918-158353918 on Build GRCh38
rs562487369 Chr.1: 158354054-158354054 on Build GRCh38

View Full Product Details