Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158357023-158357573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1E
Entrez Gene ID
913
Gene Name
CD1e molecule
Gene Aliases
CD1A, R2
UniGene
Hs.249217
Gene Chromosome Location
Chr.1: 158353157-158357554 on Build GRCh38
Transcripts
NM_001042583.2 NM_001042584.2 NM_001042585.2 NM_001042586.2 NM_001042587.2 NM_001185107.1 NM_001185108.1 NM_001185110.1 NM_001185112.1 NM_001185113.1 NM_001185114.1 NM_001185115.1 NM_030893.3 XM_011510133.2 XM_011510134.2

SNP Detail

SNP ID Chromosome Location
rs1003638372 Chr.1: 158357186-158357186 on Build GRCh38
rs149274495 Chr.1: 158357132-158357132 on Build GRCh38
rs961961718 Chr.1: 158357202-158357202 on Build GRCh38
rs934881100 Chr.1: 158357106-158357106 on Build GRCh38
rs909235655 Chr.1: 158357464-158357464 on Build GRCh38
rs777025250 Chr.1: 158357162-158357162 on Build GRCh38
rs920390360 Chr.1: 158357048-158357048 on Build GRCh38
rs369261045 Chr.1: 158357072-158357072 on Build GRCh38
rs934954774 Chr.1: 158357365-158357365 on Build GRCh38
rs548025801 Chr.1: 158357144-158357144 on Build GRCh38
rs765629 Chr.1: 158357096-158357096 on Build GRCh38
rs541321251 Chr.1: 158357204-158357204 on Build GRCh38
rs770362576 Chr.1: 158357425-158357425 on Build GRCh38
rs892204557 Chr.1: 158357338-158357338 on Build GRCh38
rs543425228 Chr.1: 158357130-158357130 on Build GRCh38
rs796107361 Chr.1: 158357235-158357236 on Build GRCh38
rs965623729 Chr.1: 158357366-158357366 on Build GRCh38
rs987908350 Chr.1: 158357110-158357110 on Build GRCh38
rs79136060 Chr.1: 158357432-158357432 on Build GRCh38
rs1032010187 Chr.1: 158357104-158357104 on Build GRCh38
rs771712604 Chr.1: 158357129-158357129 on Build GRCh38
rs191290538 Chr.1: 158357211-158357211 on Build GRCh38
rs142523394 Chr.1: 158357449-158357449 on Build GRCh38
rs951847472 Chr.1: 158357270-158357270 on Build GRCh38
rs183051737 Chr.1: 158357342-158357342 on Build GRCh38
rs896684481 Chr.1: 158357279-158357279 on Build GRCh38
rs563207612 Chr.1: 158357310-158357310 on Build GRCh38
rs763106342 Chr.1: 158357509-158357509 on Build GRCh38
rs904953629 Chr.1: 158357486-158357486 on Build GRCh38
rs35362687 Chr.1: 158357474-158357475 on Build GRCh38
rs571479434 Chr.1: 158357453-158357453 on Build GRCh38
rs567247240 Chr.1: 158357416-158357416 on Build GRCh38
rs146040069 Chr.1: 158357476-158357476 on Build GRCh38
rs139325584 Chr.1: 158357388-158357388 on Build GRCh38
rs577910868 Chr.1: 158357070-158357070 on Build GRCh38
rs1023296356 Chr.1: 158357082-158357082 on Build GRCh38
rs965308586 Chr.1: 158357117-158357117 on Build GRCh38
rs910584007 Chr.1: 158357076-158357076 on Build GRCh38
rs1019436717 Chr.1: 158357113-158357113 on Build GRCh38
rs918015163 Chr.1: 158357490-158357490 on Build GRCh38
rs931942459 Chr.1: 158357141-158357141 on Build GRCh38
rs1000913010 Chr.1: 158357316-158357316 on Build GRCh38
rs571401953 Chr.1: 158357112-158357112 on Build GRCh38
rs879502951 Chr.1: 158357236-158357236 on Build GRCh38
rs1019405380 Chr.1: 158357360-158357360 on Build GRCh38
rs993373511 Chr.1: 158357241-158357241 on Build GRCh38
rs984776430 Chr.1: 158357450-158357450 on Build GRCh38
rs145203260 Chr.1: 158357499-158357499 on Build GRCh38
rs976496743 Chr.1: 158357309-158357309 on Build GRCh38
rs949652300 Chr.1: 158357282-158357282 on Build GRCh38
rs866146436 Chr.1: 158357278-158357278 on Build GRCh38
rs910648662 Chr.1: 158357288-158357288 on Build GRCh38
rs1027527438 Chr.1: 158357260-158357260 on Build GRCh38
rs369758730 Chr.1: 158357156-158357156 on Build GRCh38
rs1045688615 Chr.1: 158357284-158357284 on Build GRCh38
rs1028620146 Chr.1: 158357441-158357442 on Build GRCh38
rs764136491 Chr.1: 158357517-158357517 on Build GRCh38
rs948882311 Chr.1: 158357471-158357471 on Build GRCh38
rs909266867 Chr.1: 158357158-158357158 on Build GRCh38
rs1034661065 Chr.1: 158357187-158357187 on Build GRCh38
rs924576406 Chr.1: 158357328-158357328 on Build GRCh38
rs61818738 Chr.1: 158357108-158357108 on Build GRCh38
rs944688092 Chr.1: 158357094-158357094 on Build GRCh38
rs557933251 Chr.1: 158357098-158357098 on Build GRCh38
rs962117459 Chr.1: 158357470-158357470 on Build GRCh38
rs146198103 Chr.1: 158357280-158357280 on Build GRCh38
rs940799536 Chr.1: 158357066-158357066 on Build GRCh38
rs904880262 Chr.1: 158357172-158357172 on Build GRCh38
rs551458470 Chr.1: 158357304-158357304 on Build GRCh38

View Full Product Details