Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160151742-160152254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A4
Entrez Gene ID
480
Gene Name
ATPase Na+/K+ transporting subunit alpha 4
Gene Aliases
ATP1A1, ATP1AL2
UniGene
Hs.662219
Gene Chromosome Location
Chr.1: 160151562-160186977 on Build GRCh38
Transcripts
NM_144699.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
424305 c.78C>G p.I26M Chr.1: 160152118-160152118 on Build GRCh38
6607416 c.55C>T p.R19* Chr.1: 160152095-160152095 on Build GRCh38
4939118 c.33T>G p.A11A Chr.1: 160152073-160152073 on Build GRCh38
5993345 c.96G>T p.K32N Chr.1: 160152136-160152136 on Build GRCh38
5992720 c.80A>C p.K27T Chr.1: 160152120-160152120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147557247 Chr.1: 160152152-160152152 on Build GRCh38
rs145797000 Chr.1: 160151897-160151897 on Build GRCh38
rs987174117 Chr.1: 160151954-160151954 on Build GRCh38
rs930476618 Chr.1: 160152217-160152217 on Build GRCh38
rs746362 Chr.1: 160151956-160151956 on Build GRCh38
rs115248829 Chr.1: 160151941-160151941 on Build GRCh38
rs751566558 Chr.1: 160151829-160151829 on Build GRCh38
rs1054048664 Chr.1: 160151971-160151971 on Build GRCh38
rs934590150 Chr.1: 160151834-160151834 on Build GRCh38
rs758607510 Chr.1: 160151783-160151783 on Build GRCh38
rs749011760 Chr.1: 160151985-160151985 on Build GRCh38
rs756778817 Chr.1: 160152056-160152056 on Build GRCh38
rs912693734 Chr.1: 160151799-160151799 on Build GRCh38
rs368224464 Chr.1: 160152136-160152136 on Build GRCh38
rs776997077 Chr.1: 160152121-160152122 on Build GRCh38
rs565588321 Chr.1: 160151816-160151816 on Build GRCh38
rs778343721 Chr.1: 160152214-160152214 on Build GRCh38
rs748584646 Chr.1: 160152140-160152141 on Build GRCh38
rs17376841 Chr.1: 160152088-160152088 on Build GRCh38
rs900596570 Chr.1: 160151902-160151902 on Build GRCh38
rs752364981 Chr.1: 160152108-160152108 on Build GRCh38
rs924239118 Chr.1: 160151813-160151813 on Build GRCh38
rs749834741 Chr.1: 160152197-160152197 on Build GRCh38
rs529208980 Chr.1: 160151804-160151804 on Build GRCh38
rs749882078 Chr.1: 160152151-160152151 on Build GRCh38
rs944251789 Chr.1: 160151809-160151809 on Build GRCh38
rs143691169 Chr.1: 160152083-160152083 on Build GRCh38
rs1053056621 Chr.1: 160151840-160151840 on Build GRCh38
rs77332724 Chr.1: 160152109-160152109 on Build GRCh38
rs953054701 Chr.1: 160151953-160151953 on Build GRCh38
rs776546098 Chr.1: 160152187-160152187 on Build GRCh38
rs183382735 Chr.1: 160152079-160152079 on Build GRCh38
rs1023789615 Chr.1: 160151954-160151954 on Build GRCh38
rs778352783 Chr.1: 160152132-160152132 on Build GRCh38
rs745547071 Chr.1: 160152072-160152072 on Build GRCh38
rs375675846 Chr.1: 160152116-160152116 on Build GRCh38
rs988488837 Chr.1: 160152161-160152161 on Build GRCh38
rs751779437 Chr.1: 160152014-160152014 on Build GRCh38
rs539242741 Chr.1: 160151823-160151823 on Build GRCh38
rs755588909 Chr.1: 160152153-160152153 on Build GRCh38
rs372349795 Chr.1: 160152209-160152209 on Build GRCh38
rs78489282 Chr.1: 160152181-160152181 on Build GRCh38
rs963295427 Chr.1: 160151942-160151942 on Build GRCh38
rs778602426 Chr.1: 160152069-160152069 on Build GRCh38
rs74123262 Chr.1: 160151768-160151768 on Build GRCh38
rs1028575137 Chr.1: 160151948-160151948 on Build GRCh38
rs973120568 Chr.1: 160151766-160151766 on Build GRCh38
rs547649929 Chr.1: 160151812-160151812 on Build GRCh38
rs371901036 Chr.1: 160152039-160152039 on Build GRCh38
rs759737165 Chr.1: 160152096-160152096 on Build GRCh38
rs764462395 Chr.1: 160151776-160151776 on Build GRCh38
rs569475320 Chr.1: 160152101-160152101 on Build GRCh38
rs965909930 Chr.1: 160151958-160151958 on Build GRCh38
rs921799862 Chr.1: 160151993-160151993 on Build GRCh38
rs748476091 Chr.1: 160152146-160152146 on Build GRCh38
rs778394399 Chr.1: 160152150-160152150 on Build GRCh38
rs564642725 Chr.1: 160151966-160151966 on Build GRCh38
rs753818842 Chr.1: 160152118-160152118 on Build GRCh38
rs967132042 Chr.1: 160152122-160152122 on Build GRCh38
rs777007153 Chr.1: 160152194-160152194 on Build GRCh38
rs757949739 Chr.1: 160152207-160152207 on Build GRCh38
rs138370635 Chr.1: 160152005-160152005 on Build GRCh38
rs953108648 Chr.1: 160151777-160151777 on Build GRCh38
rs199933555 Chr.1: 160152016-160152016 on Build GRCh38
rs745780620 Chr.1: 160151946-160151946 on Build GRCh38
rs755990380 Chr.1: 160152022-160152022 on Build GRCh38
rs951922125 Chr.1: 160151883-160151883 on Build GRCh38
rs955667376 Chr.1: 160152143-160152143 on Build GRCh38
rs1012304023 Chr.1: 160151858-160151858 on Build GRCh38
rs757928498 Chr.1: 160152139-160152139 on Build GRCh38
rs987184092 Chr.1: 160151789-160151789 on Build GRCh38
rs146860131 Chr.1: 160152095-160152095 on Build GRCh38
rs757272308 Chr.1: 160151925-160151925 on Build GRCh38
rs760437076 Chr.1: 160152186-160152186 on Build GRCh38
rs76482004 Chr.1: 160151964-160151964 on Build GRCh38
rs748535263 Chr.1: 160152082-160152082 on Build GRCh38
rs113578898 Chr.1: 160151933-160151933 on Build GRCh38
rs557492307 Chr.1: 160151828-160151828 on Build GRCh38
rs940783725 Chr.1: 160152034-160152034 on Build GRCh38
rs549038987 Chr.1: 160152127-160152127 on Build GRCh38
rs78088664 Chr.1: 160152211-160152211 on Build GRCh38
rs769059748 Chr.1: 160152077-160152077 on Build GRCh38
rs1019096599 Chr.1: 160152120-160152120 on Build GRCh38
rs747524571 Chr.1: 160152109-160152110 on Build GRCh38
rs761796232 Chr.1: 160152190-160152190 on Build GRCh38
rs576476432 Chr.1: 160151968-160151968 on Build GRCh38
rs911693292 Chr.1: 160151841-160151842 on Build GRCh38
rs763967047 Chr.1: 160152112-160152112 on Build GRCh38
rs752669899 Chr.1: 160152019-160152019 on Build GRCh38
rs370498913 Chr.1: 160152010-160152010 on Build GRCh38
rs769261788 Chr.1: 160152122-160152122 on Build GRCh38
rs780037178 Chr.1: 160152074-160152074 on Build GRCh38
rs543885172 Chr.1: 160151986-160151986 on Build GRCh38
rs779788781 Chr.1: 160152144-160152144 on Build GRCh38
rs1053764152 Chr.1: 160151817-160151817 on Build GRCh38
rs927971848 Chr.1: 160152208-160152208 on Build GRCh38
rs730313 Chr.1: 160152046-160152046 on Build GRCh38
rs371620775 Chr.1: 160152178-160152178 on Build GRCh38
rs867969835 Chr.1: 160151889-160151889 on Build GRCh38
rs376918422 Chr.1: 160152009-160152009 on Build GRCh38
rs150079838 Chr.1: 160151961-160151961 on Build GRCh38
rs974692764 Chr.1: 160152210-160152210 on Build GRCh38
rs572718039 Chr.1: 160151963-160151963 on Build GRCh38
rs368122370 Chr.1: 160152017-160152017 on Build GRCh38
rs140784515 Chr.1: 160151959-160151959 on Build GRCh38
rs749688081 Chr.1: 160152229-160152229 on Build GRCh38
rs763103557 Chr.1: 160152084-160152084 on Build GRCh38
rs867995865 Chr.1: 160152119-160152119 on Build GRCh38

View Full Product Details