Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160190806-160191310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASQ1
Entrez Gene ID
844
Gene Name
calsequestrin 1
Gene Aliases
CASQ, PDIB1, VMCQA
UniGene
Hs.632476
Gene Chromosome Location
Chr.1: 160190495-160201886 on Build GRCh38
Transcripts
NM_001231.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4536654 c.232G>A p.D78N Chr.1: 160190983-160190983 on Build GRCh38
7185688 c.126G>A p.E42E Chr.1: 160190877-160190877 on Build GRCh38
4669401 c.129C>T p.Y43Y Chr.1: 160190880-160190880 on Build GRCh38
4669402 c.240C>A p.A80A Chr.1: 160190991-160190991 on Build GRCh38
7106235 c.193G>A p.E65K Chr.1: 160190944-160190944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781172147 Chr.1: 160190974-160190974 on Build GRCh38
rs770652925 Chr.1: 160190981-160190981 on Build GRCh38
rs999894015 Chr.1: 160190913-160190913 on Build GRCh38
rs765193262 Chr.1: 160191005-160191005 on Build GRCh38
rs762974773 Chr.1: 160190937-160190937 on Build GRCh38
rs537084023 Chr.1: 160191255-160191255 on Build GRCh38
rs41265773 Chr.1: 160191075-160191075 on Build GRCh38
rs375793287 Chr.1: 160191078-160191078 on Build GRCh38
rs531621501 Chr.1: 160190980-160190980 on Build GRCh38
rs766111752 Chr.1: 160190971-160190971 on Build GRCh38
rs757491241 Chr.1: 160190840-160190840 on Build GRCh38
rs775604272 Chr.1: 160190897-160190897 on Build GRCh38
rs75077936 Chr.1: 160191158-160191158 on Build GRCh38
rs1018332082 Chr.1: 160191165-160191165 on Build GRCh38
rs753139131 Chr.1: 160190890-160190890 on Build GRCh38
rs764759200 Chr.1: 160190904-160190904 on Build GRCh38
rs756103982 Chr.1: 160191056-160191056 on Build GRCh38
rs386636030 Chr.1: 160191158-160191160 on Build GRCh38
rs775988203 Chr.1: 160190985-160190985 on Build GRCh38
rs779923292 Chr.1: 160190869-160190869 on Build GRCh38
rs951961582 Chr.1: 160191277-160191277 on Build GRCh38
rs140253806 Chr.1: 160190881-160190881 on Build GRCh38
rs749431792 Chr.1: 160190839-160190839 on Build GRCh38
rs1019026338 Chr.1: 160191232-160191232 on Build GRCh38
rs746109416 Chr.1: 160190860-160190860 on Build GRCh38
rs947673768 Chr.1: 160191004-160191004 on Build GRCh38
rs754699536 Chr.1: 160191036-160191036 on Build GRCh38
rs1001010022 Chr.1: 160191117-160191117 on Build GRCh38
rs758679972 Chr.1: 160190956-160190956 on Build GRCh38
rs923421149 Chr.1: 160190971-160190971 on Build GRCh38
rs545910107 Chr.1: 160190972-160190972 on Build GRCh38
rs779066324 Chr.1: 160190930-160190930 on Build GRCh38
rs747983130 Chr.1: 160191049-160191049 on Build GRCh38
rs374611267 Chr.1: 160190863-160190863 on Build GRCh38
rs757219885 Chr.1: 160190921-160190921 on Build GRCh38
rs150330307 Chr.1: 160191011-160191011 on Build GRCh38
rs770706586 Chr.1: 160191063-160191063 on Build GRCh38
rs779074224 Chr.1: 160190853-160190853 on Build GRCh38
rs899530407 Chr.1: 160191196-160191196 on Build GRCh38
rs974646851 Chr.1: 160191270-160191270 on Build GRCh38
rs184949450 Chr.1: 160190902-160190902 on Build GRCh38
rs74123281 Chr.1: 160190966-160190966 on Build GRCh38
rs74123282 Chr.1: 160190988-160190988 on Build GRCh38
rs997875769 Chr.1: 160191237-160191237 on Build GRCh38
rs752633420 Chr.1: 160191095-160191095 on Build GRCh38
rs907937424 Chr.1: 160191052-160191052 on Build GRCh38
rs753966210 Chr.1: 160190916-160190916 on Build GRCh38
rs745531630 Chr.1: 160191070-160191070 on Build GRCh38
rs79469730 Chr.1: 160190977-160190977 on Build GRCh38
rs768619823 Chr.1: 160190877-160190877 on Build GRCh38
rs930493885 Chr.1: 160191037-160191038 on Build GRCh38
rs35923530 Chr.1: 160190873-160190874 on Build GRCh38
rs927336336 Chr.1: 160191058-160191058 on Build GRCh38
rs138101305 Chr.1: 160190885-160190885 on Build GRCh38
rs762407330 Chr.1: 160190995-160190995 on Build GRCh38
rs112871275 Chr.1: 160191046-160191046 on Build GRCh38
rs746909396 Chr.1: 160190875-160190875 on Build GRCh38
rs748891339 Chr.1: 160191060-160191060 on Build GRCh38
rs780311434 Chr.1: 160191019-160191019 on Build GRCh38
rs1046894505 Chr.1: 160191120-160191120 on Build GRCh38
rs1005317138 Chr.1: 160190845-160190845 on Build GRCh38
rs754341004 Chr.1: 160190990-160190990 on Build GRCh38
rs149468328 Chr.1: 160191105-160191105 on Build GRCh38
rs777111674 Chr.1: 160191057-160191057 on Build GRCh38
rs779793705 Chr.1: 160190965-160190965 on Build GRCh38
rs963734855 Chr.1: 160190923-160190923 on Build GRCh38
rs750057121 Chr.1: 160191084-160191084 on Build GRCh38
rs115990089 Chr.1: 160191160-160191160 on Build GRCh38
rs368579149 Chr.1: 160190894-160190894 on Build GRCh38
rs565746759 Chr.1: 160191079-160191079 on Build GRCh38
rs543422219 Chr.1: 160190880-160190880 on Build GRCh38
rs751350723 Chr.1: 160191028-160191028 on Build GRCh38
rs374653743 Chr.1: 160190976-160190976 on Build GRCh38
rs750563617 Chr.1: 160190951-160190951 on Build GRCh38
rs1028939501 Chr.1: 160191271-160191271 on Build GRCh38
rs774082512 Chr.1: 160191064-160191064 on Build GRCh38
rs371473487 Chr.1: 160190911-160190911 on Build GRCh38

View Full Product Details