Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100806256-100806669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB4
Entrez Gene ID
2050
Gene Name
EPH receptor B4
Gene Aliases
HTK, MYK1, TYRO11
UniGene
Hs.437008
Gene Chromosome Location
Chr.7: 100802565-100827521 on Build GRCh38
Transcripts
NM_004444.4 XM_017011816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238802 c.2383C>T p.R795W Chr.7: 100806521-100806521 on Build GRCh38
1643186 c.2484C>T p.D828D Chr.7: 100806420-100806420 on Build GRCh38
4825246 c.2479C>G p.Q827E Chr.7: 100806425-100806425 on Build GRCh38
3941906 c.2343G>C p.K781N Chr.7: 100806561-100806561 on Build GRCh38
5032810 c.2353C>T p.R785* Chr.7: 100806551-100806551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375461456 Chr.7: 100806521-100806521 on Build GRCh38
rs1050266760 Chr.7: 100806362-100806362 on Build GRCh38
rs188430894 Chr.7: 100806412-100806412 on Build GRCh38
rs753128639 Chr.7: 100806429-100806429 on Build GRCh38
rs372982740 Chr.7: 100806379-100806379 on Build GRCh38
rs1050602516 Chr.7: 100806281-100806281 on Build GRCh38
rs750504664 Chr.7: 100806600-100806600 on Build GRCh38
rs368960535 Chr.7: 100806611-100806611 on Build GRCh38
rs778904315 Chr.7: 100806589-100806589 on Build GRCh38
rs564917113 Chr.7: 100806641-100806642 on Build GRCh38
rs753075600 Chr.7: 100806538-100806538 on Build GRCh38
rs957116019 Chr.7: 100806554-100806554 on Build GRCh38
rs890653624 Chr.7: 100806288-100806288 on Build GRCh38
rs138574211 Chr.7: 100806444-100806444 on Build GRCh38
rs780716438 Chr.7: 100806535-100806535 on Build GRCh38
rs938113151 Chr.7: 100806644-100806644 on Build GRCh38
rs922581035 Chr.7: 100806314-100806314 on Build GRCh38
rs377702127 Chr.7: 100806486-100806486 on Build GRCh38
rs550797013 Chr.7: 100806306-100806306 on Build GRCh38
rs779755057 Chr.7: 100806464-100806464 on Build GRCh38
rs200890953 Chr.7: 100806387-100806387 on Build GRCh38
rs906578426 Chr.7: 100806548-100806548 on Build GRCh38
rs1024547571 Chr.7: 100806456-100806456 on Build GRCh38
rs760547158 Chr.7: 100806380-100806380 on Build GRCh38
rs369079556 Chr.7: 100806420-100806420 on Build GRCh38
rs761253426 Chr.7: 100806399-100806399 on Build GRCh38
rs772293299 Chr.7: 100806457-100806457 on Build GRCh38
rs746811538 Chr.7: 100806480-100806480 on Build GRCh38
rs771847518 Chr.7: 100806377-100806377 on Build GRCh38
rs201140601 Chr.7: 100806507-100806507 on Build GRCh38
rs111764210 Chr.7: 100806376-100806376 on Build GRCh38
rs552986260 Chr.7: 100806640-100806640 on Build GRCh38
rs547435407 Chr.7: 100806296-100806296 on Build GRCh38
rs776410552 Chr.7: 100806499-100806499 on Build GRCh38
rs369337751 Chr.7: 100806378-100806378 on Build GRCh38
rs750843065 Chr.7: 100806305-100806305 on Build GRCh38
rs772917108 Chr.7: 100806520-100806520 on Build GRCh38
rs1030786732 Chr.7: 100806312-100806312 on Build GRCh38
rs1050694612 Chr.7: 100806588-100806588 on Build GRCh38
rs1001170707 Chr.7: 100806534-100806534 on Build GRCh38
rs781123870 Chr.7: 100806593-100806594 on Build GRCh38
rs764852078 Chr.7: 100806404-100806404 on Build GRCh38
rs1057519264 Chr.7: 100806559-100806559 on Build GRCh38
rs761850149 Chr.7: 100806506-100806506 on Build GRCh38
rs267601190 Chr.7: 100806555-100806555 on Build GRCh38
rs572297761 Chr.7: 100806646-100806646 on Build GRCh38
rs754413533 Chr.7: 100806441-100806441 on Build GRCh38
rs778178721 Chr.7: 100806434-100806434 on Build GRCh38
rs760922001 Chr.7: 100806540-100806540 on Build GRCh38
rs377653260 Chr.7: 100806537-100806537 on Build GRCh38
rs746073038 Chr.7: 100806453-100806453 on Build GRCh38
rs140482690 Chr.7: 100806360-100806360 on Build GRCh38
rs201432441 Chr.7: 100806409-100806409 on Build GRCh38
rs541006018 Chr.7: 100806616-100806616 on Build GRCh38
rs888497992 Chr.7: 100806403-100806403 on Build GRCh38
rs201929093 Chr.7: 100806447-100806447 on Build GRCh38
rs927772349 Chr.7: 100806419-100806419 on Build GRCh38
rs147110749 Chr.7: 100806430-100806430 on Build GRCh38
rs757837205 Chr.7: 100806586-100806586 on Build GRCh38
rs988578770 Chr.7: 100806641-100806641 on Build GRCh38
rs754293421 Chr.7: 100806572-100806572 on Build GRCh38
rs199910843 Chr.7: 100806474-100806474 on Build GRCh38

View Full Product Details