Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101161125-101161658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1S1
Entrez Gene ID
1174
Gene Name
adaptor related protein complex 1 sigma 1 subunit
Gene Aliases
AP19, CLAPS1, EKV3, MEDNIK, SIGMA1A
UniGene
Hs.489365
Gene Chromosome Location
Chr.7: 101154405-101161276 on Build GRCh38
Transcripts
NM_001283.3

SNP Detail

SNP ID Chromosome Location
rs968395979 Chr.7: 101161599-101161599 on Build GRCh38
rs3177697 Chr.7: 101161261-101161261 on Build GRCh38
rs1057299987 Chr.7: 101161551-101161551 on Build GRCh38
rs958959531 Chr.7: 101161613-101161613 on Build GRCh38
rs887296180 Chr.7: 101161186-101161186 on Build GRCh38
rs1000376645 Chr.7: 101161587-101161587 on Build GRCh38
rs930517083 Chr.7: 101161407-101161407 on Build GRCh38
rs1048375 Chr.7: 101161180-101161180 on Build GRCh38
rs1036881555 Chr.7: 101161373-101161373 on Build GRCh38
rs1031973434 Chr.7: 101161619-101161619 on Build GRCh38
rs1009300821 Chr.7: 101161533-101161533 on Build GRCh38
rs13221344 Chr.7: 101161217-101161217 on Build GRCh38
rs149674622 Chr.7: 101161210-101161210 on Build GRCh38
rs1022601949 Chr.7: 101161250-101161250 on Build GRCh38
rs998733489 Chr.7: 101161218-101161218 on Build GRCh38
rs895543543 Chr.7: 101161566-101161566 on Build GRCh38
rs185916008 Chr.7: 101161287-101161287 on Build GRCh38
rs1039729260 Chr.7: 101161155-101161155 on Build GRCh38
rs950174986 Chr.7: 101161370-101161370 on Build GRCh38
rs944122836 Chr.7: 101161422-101161422 on Build GRCh38
rs778265405 Chr.7: 101161480-101161480 on Build GRCh38
rs1035880385 Chr.7: 101161204-101161204 on Build GRCh38
rs1048389 Chr.7: 101161195-101161195 on Build GRCh38
rs559224956 Chr.7: 101161570-101161570 on Build GRCh38
rs912557618 Chr.7: 101161153-101161153 on Build GRCh38
rs917295165 Chr.7: 101161359-101161359 on Build GRCh38
rs956102123 Chr.7: 101161291-101161291 on Build GRCh38
rs752646332 Chr.7: 101161345-101161345 on Build GRCh38
rs1022570715 Chr.7: 101161577-101161577 on Build GRCh38
rs991457501 Chr.7: 101161614-101161614 on Build GRCh38
rs1039697633 Chr.7: 101161423-101161423 on Build GRCh38
rs571055443 Chr.7: 101161612-101161612 on Build GRCh38
rs1049160212 Chr.7: 101161467-101161467 on Build GRCh38
rs959787740 Chr.7: 101161215-101161215 on Build GRCh38
rs1048365 Chr.7: 101161149-101161149 on Build GRCh38
rs187626778 Chr.7: 101161519-101161519 on Build GRCh38
rs991488089 Chr.7: 101161326-101161326 on Build GRCh38
rs1003961466 Chr.7: 101161191-101161191 on Build GRCh38
rs979007485 Chr.7: 101161281-101161281 on Build GRCh38
rs191612208 Chr.7: 101161292-101161292 on Build GRCh38
rs889039313 Chr.7: 101161465-101161465 on Build GRCh38
rs919077882 Chr.7: 101161375-101161375 on Build GRCh38
rs1004753405 Chr.7: 101161518-101161518 on Build GRCh38
rs955986380 Chr.7: 101161620-101161620 on Build GRCh38
rs2158611 Chr.7: 101161450-101161450 on Build GRCh38
rs77060326 Chr.7: 101161177-101161177 on Build GRCh38
rs755438433 Chr.7: 101161229-101161229 on Build GRCh38
rs1021164209 Chr.7: 101161547-101161547 on Build GRCh38
rs979112447 Chr.7: 101161265-101161265 on Build GRCh38
rs987686959 Chr.7: 101161634-101161634 on Build GRCh38
rs182592421 Chr.7: 101161293-101161293 on Build GRCh38
rs995771239 Chr.7: 101161176-101161176 on Build GRCh38
rs3188428 Chr.7: 101161213-101161213 on Build GRCh38
rs564715150 Chr.7: 101161154-101161154 on Build GRCh38
rs748486739 Chr.7: 101161365-101161365 on Build GRCh38
rs900062430 Chr.7: 101161156-101161156 on Build GRCh38
rs921293539 Chr.7: 101161432-101161432 on Build GRCh38
rs912612341 Chr.7: 101161320-101161320 on Build GRCh38

View Full Product Details