Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101237657-101238147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN15
Entrez Gene ID
24146
Gene Name
claudin 15
Gene Aliases
-
UniGene
Hs.38738
Gene Chromosome Location
Chr.7: 101232092-101238820 on Build GRCh38
Transcripts
NM_001185080.1 NM_014343.2

SNP Detail

SNP ID Chromosome Location
rs111333670 Chr.7: 101237769-101237769 on Build GRCh38
rs529104553 Chr.7: 101237896-101237896 on Build GRCh38
rs952893364 Chr.7: 101237728-101237728 on Build GRCh38
rs1053831393 Chr.7: 101237714-101237714 on Build GRCh38
rs963521671 Chr.7: 101237820-101237820 on Build GRCh38
rs773263425 Chr.7: 101237914-101237914 on Build GRCh38
rs1029537734 Chr.7: 101237944-101237944 on Build GRCh38
rs1016118375 Chr.7: 101238111-101238111 on Build GRCh38
rs58557289 Chr.7: 101238048-101238048 on Build GRCh38
rs1004717334 Chr.7: 101238096-101238096 on Build GRCh38
rs917535467 Chr.7: 101237929-101237929 on Build GRCh38
rs1008507403 Chr.7: 101237856-101237856 on Build GRCh38
rs748498620 Chr.7: 101237898-101237898 on Build GRCh38
rs543800671 Chr.7: 101237915-101237915 on Build GRCh38
rs767034599 Chr.7: 101237953-101237953 on Build GRCh38
rs772583730 Chr.7: 101237907-101237907 on Build GRCh38
rs908618737 Chr.7: 101237921-101237921 on Build GRCh38
rs935318807 Chr.7: 101237978-101237978 on Build GRCh38
rs1016588517 Chr.7: 101238113-101238113 on Build GRCh38
rs1044572888 Chr.7: 101237815-101237815 on Build GRCh38
rs796284892 Chr.7: 101237921-101237922 on Build GRCh38
rs975210625 Chr.7: 101237927-101237927 on Build GRCh38
rs576461807 Chr.7: 101237833-101237833 on Build GRCh38
rs935771001 Chr.7: 101237985-101237985 on Build GRCh38
rs999528128 Chr.7: 101237694-101237694 on Build GRCh38
rs922138403 Chr.7: 101237843-101237843 on Build GRCh38
rs532083628 Chr.7: 101237814-101237815 on Build GRCh38
rs373647714 Chr.7: 101237819-101237819 on Build GRCh38
rs865793658 Chr.7: 101238038-101238038 on Build GRCh38
rs779624611 Chr.7: 101237727-101237727 on Build GRCh38
rs1003479289 Chr.7: 101237801-101237801 on Build GRCh38
rs557308067 Chr.7: 101237832-101237832 on Build GRCh38
rs141809529 Chr.7: 101238006-101238006 on Build GRCh38
rs189595322 Chr.7: 101237960-101237960 on Build GRCh38
rs148059973 Chr.7: 101237738-101237738 on Build GRCh38
rs948175373 Chr.7: 101237921-101237921 on Build GRCh38
rs574712521 Chr.7: 101237803-101237803 on Build GRCh38
rs778962988 Chr.7: 101237773-101237774 on Build GRCh38
rs1012363607 Chr.7: 101237920-101237920 on Build GRCh38
rs900439892 Chr.7: 101238044-101238044 on Build GRCh38
rs201531161 Chr.7: 101237920-101237921 on Build GRCh38
rs565178153 Chr.7: 101237953-101237953 on Build GRCh38
rs769275366 Chr.7: 101237807-101237807 on Build GRCh38
rs961409739 Chr.7: 101237922-101237922 on Build GRCh38
rs557880219 Chr.7: 101237719-101237719 on Build GRCh38
rs948923814 Chr.7: 101237950-101237950 on Build GRCh38
rs1045162038 Chr.7: 101238028-101238028 on Build GRCh38
rs534466198 Chr.7: 101237786-101237786 on Build GRCh38
rs933546021 Chr.7: 101237846-101237846 on Build GRCh38
rs961443121 Chr.7: 101238117-101238117 on Build GRCh38
rs547566377 Chr.7: 101237923-101237923 on Build GRCh38
rs17135348 Chr.7: 101237799-101237799 on Build GRCh38
rs570024517 Chr.7: 101238066-101238066 on Build GRCh38
rs940294447 Chr.7: 101237756-101237756 on Build GRCh38
rs1027871193 Chr.7: 101237899-101237899 on Build GRCh38
rs948891240 Chr.7: 101237811-101237811 on Build GRCh38
rs529338779 Chr.7: 101238029-101238029 on Build GRCh38
rs1005246295 Chr.7: 101237923-101237924 on Build GRCh38
rs1018855699 Chr.7: 101237878-101237878 on Build GRCh38
rs895360256 Chr.7: 101237875-101237875 on Build GRCh38
rs73408493 Chr.7: 101238091-101238091 on Build GRCh38
rs779632413 Chr.7: 101237818-101237818 on Build GRCh38
rs1023536937 Chr.7: 101237771-101237771 on Build GRCh38
rs565874232 Chr.7: 101238012-101238012 on Build GRCh38
rs530381048 Chr.7: 101238086-101238086 on Build GRCh38
rs964545766 Chr.7: 101237691-101237691 on Build GRCh38
rs908500433 Chr.7: 101237748-101237748 on Build GRCh38
rs989408741 Chr.7: 101237980-101237980 on Build GRCh38
rs59928515 Chr.7: 101238007-101238007 on Build GRCh38
rs113185246 Chr.7: 101237894-101237894 on Build GRCh38
rs1005488008 Chr.7: 101238094-101238094 on Build GRCh38
rs1044605650 Chr.7: 101237952-101237952 on Build GRCh38
rs3757458 Chr.7: 101238122-101238122 on Build GRCh38
rs987745249 Chr.7: 101237851-101237851 on Build GRCh38

View Full Product Details