Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102097262-102097758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUX1
Entrez Gene ID
1523
Gene Name
cut like homeobox 1
Gene Aliases
CASP, CDP, CDP/Cut, CDP1, COY1, CUTL1, CUX, Clox, Cux/CDP, GOLIM6, Nbla10317, p100, p110, p200, p75
UniGene
Hs.191482
Gene Chromosome Location
Chr.7: 101815904-102283958 on Build GRCh38
Transcripts
NM_001202543.1 NM_001202544.2 NM_001202545.2 NM_001202546.2 NM_001913.4 NM_181500.3 NM_181552.3 XM_005250150.2 XM_005250151.2 XM_005250154.3 XM_006715854.2 XM_006715855.1 XM_011515823.2 XM_011515825.1 XM_017011759.1 XM_017011760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5452602 c.318G>A p.L106L Chr.7: 102097380-102097380 on Build GRCh38
5452600 c.317T>A p.L106* Chr.7: 102097379-102097379 on Build GRCh38
1488083 c.370G>T p.E124* Chr.7: 102097465-102097465 on Build GRCh38
3083207 c.391G>A p.E131K Chr.7: 102097486-102097486 on Build GRCh38
6281398 c.361G>A p.D121N Chr.7: 102097423-102097423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750368974 Chr.7: 102097502-102097502 on Build GRCh38
rs782020138 Chr.7: 102097486-102097486 on Build GRCh38
rs77767003 Chr.7: 102097547-102097547 on Build GRCh38
rs113120121 Chr.7: 102097711-102097711 on Build GRCh38
rs572081545 Chr.7: 102097715-102097715 on Build GRCh38
rs782302433 Chr.7: 102097508-102097508 on Build GRCh38
rs779056805 Chr.7: 102097402-102097402 on Build GRCh38
rs202141115 Chr.7: 102097343-102097343 on Build GRCh38
rs1637257 Chr.7: 102097326-102097326 on Build GRCh38
rs782605307 Chr.7: 102097509-102097510 on Build GRCh38
rs11314718 Chr.7: 102097710-102097710 on Build GRCh38
rs974344463 Chr.7: 102097642-102097642 on Build GRCh38
rs571246660 Chr.7: 102097307-102097307 on Build GRCh38
rs782462214 Chr.7: 102097422-102097422 on Build GRCh38
rs371765100 Chr.7: 102097595-102097595 on Build GRCh38
rs781951661 Chr.7: 102097352-102097352 on Build GRCh38
rs782089593 Chr.7: 102097396-102097396 on Build GRCh38
rs77240477 Chr.7: 102097436-102097436 on Build GRCh38
rs951579016 Chr.7: 102097712-102097712 on Build GRCh38
rs781863665 Chr.7: 102097404-102097404 on Build GRCh38
rs781944554 Chr.7: 102097518-102097518 on Build GRCh38
rs371126154 Chr.7: 102097489-102097489 on Build GRCh38
rs1007681758 Chr.7: 102097533-102097533 on Build GRCh38
rs782042708 Chr.7: 102097370-102097370 on Build GRCh38
rs782636888 Chr.7: 102097497-102097497 on Build GRCh38
rs781902544 Chr.7: 102097539-102097539 on Build GRCh38
rs201081607 Chr.7: 102097535-102097535 on Build GRCh38
rs578257480 Chr.7: 102097677-102097677 on Build GRCh38
rs375459438 Chr.7: 102097548-102097548 on Build GRCh38
rs556893973 Chr.7: 102097651-102097651 on Build GRCh38
rs782711546 Chr.7: 102097549-102097549 on Build GRCh38
rs368204658 Chr.7: 102097517-102097517 on Build GRCh38
rs782565144 Chr.7: 102097319-102097319 on Build GRCh38
rs10638510 Chr.7: 102097547-102097548 on Build GRCh38
rs782789147 Chr.7: 102097539-102097540 on Build GRCh38
rs1042943750 Chr.7: 102097453-102097453 on Build GRCh38
rs781786123 Chr.7: 102097387-102097387 on Build GRCh38
rs782673997 Chr.7: 102097540-102097540 on Build GRCh38
rs1734879 Chr.7: 102097330-102097330 on Build GRCh38
rs781797963 Chr.7: 102097709-102097710 on Build GRCh38
rs181804840 Chr.7: 102097287-102097287 on Build GRCh38
rs782466159 Chr.7: 102097325-102097325 on Build GRCh38
rs781883497 Chr.7: 102097415-102097415 on Build GRCh38
rs782362424 Chr.7: 102097338-102097338 on Build GRCh38
rs782382132 Chr.7: 102097369-102097369 on Build GRCh38
rs560538069 Chr.7: 102097714-102097714 on Build GRCh38
rs903050299 Chr.7: 102097467-102097467 on Build GRCh38
rs782180373 Chr.7: 102097332-102097332 on Build GRCh38
rs370206245 Chr.7: 102097313-102097313 on Build GRCh38
rs782151315 Chr.7: 102097383-102097383 on Build GRCh38
rs782225705 Chr.7: 102097320-102097320 on Build GRCh38
rs372478457 Chr.7: 102097300-102097300 on Build GRCh38
rs920120233 Chr.7: 102097295-102097295 on Build GRCh38
rs201537465 Chr.7: 102097433-102097433 on Build GRCh38
rs782121474 Chr.7: 102097534-102097534 on Build GRCh38
rs782135600 Chr.7: 102097355-102097355 on Build GRCh38
rs782411361 Chr.7: 102097515-102097515 on Build GRCh38
rs782612569 Chr.7: 102097423-102097423 on Build GRCh38
rs35412282 Chr.7: 102097548-102097549 on Build GRCh38
rs782014907 Chr.7: 102097331-102097331 on Build GRCh38
rs782721883 Chr.7: 102097398-102097398 on Build GRCh38
rs920192392 Chr.7: 102097647-102097647 on Build GRCh38
rs1047237813 Chr.7: 102097304-102097304 on Build GRCh38
rs538283487 Chr.7: 102097612-102097612 on Build GRCh38
rs782812674 Chr.7: 102097386-102097386 on Build GRCh38
rs782680467 Chr.7: 102097414-102097414 on Build GRCh38
rs34247954 Chr.7: 102097713-102097713 on Build GRCh38
rs782504553 Chr.7: 102097408-102097408 on Build GRCh38

View Full Product Details