Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102751282-102751809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM185A
Entrez Gene ID
222234
Gene Name
family with sequence similarity 185 member A
Gene Aliases
-
UniGene
Hs.202543
Gene Chromosome Location
Chr.7: 102748944-102913394 on Build GRCh38
Transcripts
NM_001145268.1 NM_001145269.1 NR_026879.1 XM_006715896.3 XM_011515922.2 XM_011515923.2 XM_011515924.2 XM_011515925.2 XM_011515926.2 XM_017011846.1 XM_017011847.1 XM_017011848.1 XM_017011849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5952592 c.560A>C p.K187T Chr.7: 102751800-102751800 on Build GRCh38
7213135 c.487A>G p.K163E Chr.7: 102751727-102751727 on Build GRCh38
1137593 c.526A>G p.T176A Chr.7: 102751766-102751766 on Build GRCh38
4745966 c.482_489delGTGTAAAA p.V162fs*4 Chr.7: 102751722-102751729 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754586765 Chr.7: 102751722-102751729 on Build GRCh38
rs560079207 Chr.7: 102751682-102751682 on Build GRCh38
rs1009592835 Chr.7: 102751554-102751554 on Build GRCh38
rs1000960599 Chr.7: 102751509-102751509 on Build GRCh38
rs764891653 Chr.7: 102751609-102751610 on Build GRCh38
rs955163439 Chr.7: 102751609-102751609 on Build GRCh38
rs556072667 Chr.7: 102751491-102751491 on Build GRCh38
rs533642875 Chr.7: 102751395-102751395 on Build GRCh38
rs1030317329 Chr.7: 102751579-102751579 on Build GRCh38
rs1046696185 Chr.7: 102751402-102751402 on Build GRCh38
rs769051661 Chr.7: 102751655-102751655 on Build GRCh38
rs929620837 Chr.7: 102751396-102751396 on Build GRCh38
rs776927695 Chr.7: 102751666-102751666 on Build GRCh38
rs770747328 Chr.7: 102751537-102751537 on Build GRCh38
rs780968191 Chr.7: 102751540-102751540 on Build GRCh38
rs773379551 Chr.7: 102751761-102751761 on Build GRCh38
rs760476934 Chr.7: 102751540-102751541 on Build GRCh38
rs973753713 Chr.7: 102751381-102751381 on Build GRCh38
rs528593607 Chr.7: 102751694-102751694 on Build GRCh38
rs200475628 Chr.7: 102751775-102751775 on Build GRCh38
rs12113137 Chr.7: 102751508-102751508 on Build GRCh38
rs986731905 Chr.7: 102751344-102751344 on Build GRCh38
rs544115336 Chr.7: 102751464-102751464 on Build GRCh38
rs555352682 Chr.7: 102751436-102751436 on Build GRCh38
rs910711800 Chr.7: 102751355-102751355 on Build GRCh38
rs12113806 Chr.7: 102751377-102751377 on Build GRCh38
rs573255452 Chr.7: 102751443-102751443 on Build GRCh38
rs917857005 Chr.7: 102751467-102751467 on Build GRCh38
rs540319110 Chr.7: 102751766-102751766 on Build GRCh38
rs1045343518 Chr.7: 102751490-102751490 on Build GRCh38
rs3748116 Chr.7: 102751527-102751527 on Build GRCh38
rs768640700 Chr.7: 102751469-102751469 on Build GRCh38
rs963548897 Chr.7: 102751774-102751774 on Build GRCh38

View Full Product Details