Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102884051-102884461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM185A
Entrez Gene ID
222234
Gene Name
family with sequence similarity 185 member A
Gene Aliases
-
UniGene
Hs.202543
Gene Chromosome Location
Chr.7: 102748944-102913394 on Build GRCh38
Transcripts
XM_011515923.2 XM_011515924.2 XM_017011846.1 XM_017011847.1 XM_017011848.1 XM_017011849.1 NM_001111038.1 NM_001287150.1 NM_145032.3 NR_105043.1 XM_005250205.3 XM_005250207.3 XM_005250208.3 XM_005250209.2 XM_006715898.2 XM_011515928.2 XM_011515929.2 XM_011515930.2 XM_011515932.2 XM_017011850.1 XM_017011851.1 XM_017011852.1 XM_017011853.1

Gene Symbol
FBXL13
Entrez Gene ID
222235
Gene Name
F-box and leucine rich repeat protein 13
Gene Aliases
DRC6, Fbl13
UniGene
Hs.660029
Gene Chromosome Location
Chr.7: 102699226-103074841 on Build GRCh38
Transcripts
XM_011515923.2 XM_011515924.2 XM_017011846.1 XM_017011847.1 XM_017011848.1 XM_017011849.1 NM_001111038.1 NM_001287150.1 NM_145032.3 NR_105043.1 XM_005250205.3 XM_005250207.3 XM_005250208.3 XM_005250209.2 XM_006715898.2 XM_011515928.2 XM_011515929.2 XM_011515930.2 XM_011515932.2 XM_017011850.1 XM_017011851.1 XM_017011852.1 XM_017011853.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6837100 c.1098C>G p.N366K Chr.7: 102884223-102884223 on Build GRCh38
7099683 c.1012T>C p.S338P Chr.7: 102884309-102884309 on Build GRCh38
4904496 c.1089G>A p.L363L Chr.7: 102884232-102884232 on Build GRCh38
4422197 c.1091C>G p.T364R Chr.7: 102884230-102884230 on Build GRCh38
5472322 c.1052G>A p.G351E Chr.7: 102884269-102884269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138676670 Chr.7: 102884238-102884238 on Build GRCh38
rs986251028 Chr.7: 102884153-102884153 on Build GRCh38
rs562927680 Chr.7: 102884300-102884300 on Build GRCh38
rs376851494 Chr.7: 102884272-102884272 on Build GRCh38
rs369576055 Chr.7: 102884318-102884318 on Build GRCh38
rs769711866 Chr.7: 102884273-102884273 on Build GRCh38
rs760119487 Chr.7: 102884349-102884349 on Build GRCh38
rs773005670 Chr.7: 102884275-102884275 on Build GRCh38
rs753424939 Chr.7: 102884296-102884296 on Build GRCh38
rs747212629 Chr.7: 102884257-102884257 on Build GRCh38
rs988053039 Chr.7: 102884087-102884087 on Build GRCh38
rs866202473 Chr.7: 102884381-102884381 on Build GRCh38
rs923124539 Chr.7: 102884097-102884097 on Build GRCh38
rs76187115 Chr.7: 102884388-102884388 on Build GRCh38
rs1013036408 Chr.7: 102884317-102884317 on Build GRCh38
rs765668960 Chr.7: 102884236-102884236 on Build GRCh38
rs574251841 Chr.7: 102884156-102884156 on Build GRCh38
rs10267104 Chr.7: 102884346-102884346 on Build GRCh38
rs372238022 Chr.7: 102884207-102884207 on Build GRCh38
rs910640810 Chr.7: 102884162-102884162 on Build GRCh38
rs772609097 Chr.7: 102884227-102884227 on Build GRCh38
rs866738051 Chr.7: 102884228-102884229 on Build GRCh38
rs369267502 Chr.7: 102884177-102884177 on Build GRCh38
rs986617918 Chr.7: 102884402-102884402 on Build GRCh38
rs138485936 Chr.7: 102884405-102884405 on Build GRCh38
rs34528283 Chr.7: 102884369-102884370 on Build GRCh38
rs749311013 Chr.7: 102884288-102884288 on Build GRCh38
rs773588752 Chr.7: 102884260-102884260 on Build GRCh38
rs1000593823 Chr.7: 102884414-102884414 on Build GRCh38
rs975752297 Chr.7: 102884431-102884431 on Build GRCh38
rs766179436 Chr.7: 102884292-102884293 on Build GRCh38
rs527300760 Chr.7: 102884437-102884437 on Build GRCh38
rs770605328 Chr.7: 102884293-102884293 on Build GRCh38
rs933129820 Chr.7: 102884108-102884108 on Build GRCh38
rs760453386 Chr.7: 102884229-102884230 on Build GRCh38
rs755271767 Chr.7: 102884259-102884259 on Build GRCh38
rs780507048 Chr.7: 102884246-102884246 on Build GRCh38
rs533077981 Chr.7: 102884301-102884301 on Build GRCh38
rs766250685 Chr.7: 102884193-102884193 on Build GRCh38
rs759230889 Chr.7: 102884224-102884224 on Build GRCh38
rs752228847 Chr.7: 102884230-102884230 on Build GRCh38
rs963881465 Chr.7: 102884408-102884408 on Build GRCh38
rs757745253 Chr.7: 102884234-102884234 on Build GRCh38
rs774915378 Chr.7: 102884325-102884325 on Build GRCh38
rs1021855460 Chr.7: 102884281-102884281 on Build GRCh38
rs896022041 Chr.7: 102884298-102884298 on Build GRCh38
rs1030744613 Chr.7: 102884382-102884382 on Build GRCh38
rs1000456080 Chr.7: 102884371-102884371 on Build GRCh38
rs758676811 Chr.7: 102884240-102884240 on Build GRCh38
rs887156498 Chr.7: 102884219-102884219 on Build GRCh38
rs556102142 Chr.7: 102884099-102884099 on Build GRCh38
rs931305680 Chr.7: 102884094-102884094 on Build GRCh38
rs766995818 Chr.7: 102884229-102884229 on Build GRCh38
rs751205666 Chr.7: 102884205-102884205 on Build GRCh38

View Full Product Details