Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 105565131-105565638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB10
Entrez Gene ID
100130771
Gene Name
EF-hand calcium binding domain 10
Gene Aliases
-
UniGene
Hs.719432
Gene Chromosome Location
Chr.7: 105565133-105581529 on Build GRCh38
Transcripts
NR_027068.1 NM_021930.4 XM_005250524.3 XM_011516458.2 XM_017012496.1 XM_017012497.1 XM_017012498.1

Gene Symbol
RINT1
Entrez Gene ID
60561
Gene Name
RAD50 interactor 1
Gene Aliases
RINT-1
UniGene
Hs.531388
Gene Chromosome Location
Chr.7: 105532085-105567688 on Build GRCh38
Transcripts
NR_027068.1 NM_021930.4 XM_005250524.3 XM_011516458.2 XM_017012496.1 XM_017012497.1 XM_017012498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3254147 c.1906C>G p.Q636E Chr.7: 105565296-105565296 on Build GRCh38
3781238 c.1927T>A p.S643T Chr.7: 105565317-105565317 on Build GRCh38
5976170 c.1906C>T p.Q636* Chr.7: 105565296-105565296 on Build GRCh38
7202056 c.2077G>T p.A693S Chr.7: 105565539-105565539 on Build GRCh38
4155357 c.2042A>T p.K681M Chr.7: 105565432-105565432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544366002 Chr.7: 105565243-105565243 on Build GRCh38
rs757812202 Chr.7: 105565316-105565316 on Build GRCh38
rs985197903 Chr.7: 105565184-105565184 on Build GRCh38
rs113164663 Chr.7: 105565364-105565364 on Build GRCh38
rs755565434 Chr.7: 105565233-105565233 on Build GRCh38
rs776280054 Chr.7: 105565292-105565292 on Build GRCh38
rs763343254 Chr.7: 105565607-105565607 on Build GRCh38
rs755904615 Chr.7: 105565550-105565550 on Build GRCh38
rs746195338 Chr.7: 105565260-105565263 on Build GRCh38
rs758518451 Chr.7: 105565419-105565419 on Build GRCh38
rs749460107 Chr.7: 105565259-105565259 on Build GRCh38
rs771142191 Chr.7: 105565262-105565262 on Build GRCh38
rs761520355 Chr.7: 105565300-105565300 on Build GRCh38
rs778220756 Chr.7: 105565339-105565339 on Build GRCh38
rs746864752 Chr.7: 105565357-105565357 on Build GRCh38
rs774027650 Chr.7: 105565575-105565575 on Build GRCh38
rs530777123 Chr.7: 105565471-105565471 on Build GRCh38
rs774643074 Chr.7: 105565265-105565265 on Build GRCh38
rs780062646 Chr.7: 105565356-105565356 on Build GRCh38
rs748582789 Chr.7: 105565240-105565240 on Build GRCh38
rs745405558 Chr.7: 105565578-105565578 on Build GRCh38
rs747097541 Chr.7: 105565435-105565435 on Build GRCh38
rs35971380 Chr.7: 105565393-105565393 on Build GRCh38
rs35531972 Chr.7: 105565368-105565368 on Build GRCh38
rs777493133 Chr.7: 105565552-105565552 on Build GRCh38
rs974799105 Chr.7: 105565162-105565162 on Build GRCh38
rs141684814 Chr.7: 105565361-105565361 on Build GRCh38
rs750563744 Chr.7: 105565418-105565418 on Build GRCh38
rs761799563 Chr.7: 105565456-105565459 on Build GRCh38
rs113457803 Chr.7: 105565183-105565183 on Build GRCh38
rs757668486 Chr.7: 105565411-105565411 on Build GRCh38
rs765744917 Chr.7: 105565380-105565380 on Build GRCh38
rs145856362 Chr.7: 105565572-105565572 on Build GRCh38
rs779310406 Chr.7: 105565416-105565416 on Build GRCh38
rs368702915 Chr.7: 105565362-105565362 on Build GRCh38
rs566796840 Chr.7: 105565605-105565605 on Build GRCh38
rs865981937 Chr.7: 105565304-105565305 on Build GRCh38
rs771659321 Chr.7: 105565585-105565585 on Build GRCh38
rs922021785 Chr.7: 105565167-105565167 on Build GRCh38
rs751630489 Chr.7: 105565519-105565519 on Build GRCh38
rs756469608 Chr.7: 105565159-105565159 on Build GRCh38
rs864687 Chr.7: 105565470-105565470 on Build GRCh38
rs986540926 Chr.7: 105565351-105565351 on Build GRCh38
rs753248481 Chr.7: 105565388-105565388 on Build GRCh38
rs770175502 Chr.7: 105565266-105565267 on Build GRCh38
rs867300610 Chr.7: 105565319-105565319 on Build GRCh38
rs904888408 Chr.7: 105565214-105565214 on Build GRCh38
rs755009395 Chr.7: 105565529-105565529 on Build GRCh38
rs768627748 Chr.7: 105565440-105565440 on Build GRCh38
rs949105262 Chr.7: 105565401-105565401 on Build GRCh38
rs553231338 Chr.7: 105565165-105565165 on Build GRCh38
rs818624 Chr.7: 105565209-105565209 on Build GRCh38
rs747965842 Chr.7: 105565448-105565448 on Build GRCh38
rs200446656 Chr.7: 105565530-105565530 on Build GRCh38
rs866767336 Chr.7: 105565295-105565296 on Build GRCh38
rs187666745 Chr.7: 105565330-105565330 on Build GRCh38
rs200970042 Chr.7: 105565363-105565363 on Build GRCh38
rs147409665 Chr.7: 105565340-105565340 on Build GRCh38
rs766277399 Chr.7: 105565484-105565485 on Build GRCh38
rs745925045 Chr.7: 105565271-105565271 on Build GRCh38
rs997658204 Chr.7: 105565547-105565547 on Build GRCh38
rs746000896 Chr.7: 105565346-105565346 on Build GRCh38
rs775261161 Chr.7: 105565280-105565280 on Build GRCh38
rs752202705 Chr.7: 105565190-105565190 on Build GRCh38
rs772098271 Chr.7: 105565274-105565274 on Build GRCh38
rs754359023 Chr.7: 105565504-105565504 on Build GRCh38
rs780694637 Chr.7: 105565541-105565542 on Build GRCh38
rs1030845401 Chr.7: 105565331-105565331 on Build GRCh38
rs762292576 Chr.7: 105565505-105565505 on Build GRCh38
rs772697416 Chr.7: 105565374-105565374 on Build GRCh38
rs860182 Chr.7: 105565494-105565494 on Build GRCh38
rs527456391 Chr.7: 105565234-105565234 on Build GRCh38
rs112914598 Chr.7: 105565463-105565463 on Build GRCh38
rs185684106 Chr.7: 105565422-105565422 on Build GRCh38
rs776469444 Chr.7: 105565518-105565519 on Build GRCh38
rs750663952 Chr.7: 105565507-105565507 on Build GRCh38
rs749487545 Chr.7: 105565363-105565364 on Build GRCh38
rs764854638 Chr.7: 105565315-105565315 on Build GRCh38
rs755107934 Chr.7: 105565563-105565563 on Build GRCh38
rs765770922 Chr.7: 105565598-105565598 on Build GRCh38
rs147460279 Chr.7: 105565246-105565246 on Build GRCh38
rs861761 Chr.7: 105565508-105565508 on Build GRCh38
rs1036206288 Chr.7: 105565178-105565178 on Build GRCh38
rs967432691 Chr.7: 105565298-105565298 on Build GRCh38
rs753092357 Chr.7: 105565336-105565336 on Build GRCh38
rs770541803 Chr.7: 105565574-105565574 on Build GRCh38
rs867880437 Chr.7: 105565310-105565311 on Build GRCh38
rs777291719 Chr.7: 105565590-105565590 on Build GRCh38
rs139766063 Chr.7: 105565596-105565596 on Build GRCh38
rs139459919 Chr.7: 105565375-105565375 on Build GRCh38
rs754306128 Chr.7: 105565406-105565406 on Build GRCh38
rs768922101 Chr.7: 105565404-105565416 on Build GRCh38
rs118063235 Chr.7: 105565187-105565187 on Build GRCh38
rs772926485 Chr.7: 105565464-105565464 on Build GRCh38
rs778159459 Chr.7: 105565254-105565254 on Build GRCh38
rs775872148 Chr.7: 105565295-105565298 on Build GRCh38
rs779258990 Chr.7: 105565344-105565344 on Build GRCh38
rs139532717 Chr.7: 105565352-105565352 on Build GRCh38
rs952224777 Chr.7: 105565475-105565475 on Build GRCh38
rs762443021 Chr.7: 105565468-105565468 on Build GRCh38
rs764689459 Chr.7: 105565400-105565400 on Build GRCh38
rs759097634 Chr.7: 105565480-105565480 on Build GRCh38
rs756510641 Chr.7: 105565338-105565338 on Build GRCh38
rs765462365 Chr.7: 105565506-105565506 on Build GRCh38
rs370890616 Chr.7: 105565513-105565513 on Build GRCh38
rs774482766 Chr.7: 105565453-105565455 on Build GRCh38
rs763828038 Chr.7: 105565281-105565281 on Build GRCh38

View Full Product Details