Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 106658169-106658687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC71L
Entrez Gene ID
168455
Gene Name
coiled-coil domain containing 71-like
Gene Aliases
C7orf74
UniGene
Hs.29692
Gene Chromosome Location
Chr.7: 106656765-106661188 on Build GRCh38
Transcripts
NM_175884.4

SNP Detail

SNP ID Chromosome Location
rs1018281394 Chr.7: 106658371-106658371 on Build GRCh38
rs555382832 Chr.7: 106658484-106658484 on Build GRCh38
rs981271720 Chr.7: 106658594-106658594 on Build GRCh38
rs10266131 Chr.7: 106658375-106658375 on Build GRCh38
rs902955455 Chr.7: 106658227-106658227 on Build GRCh38
rs776968279 Chr.7: 106658514-106658514 on Build GRCh38
rs928351568 Chr.7: 106658360-106658360 on Build GRCh38
rs929361417 Chr.7: 106658428-106658428 on Build GRCh38
rs558255489 Chr.7: 106658646-106658646 on Build GRCh38
rs1055423261 Chr.7: 106658374-106658374 on Build GRCh38
rs10236648 Chr.7: 106658381-106658381 on Build GRCh38
rs1014736004 Chr.7: 106658567-106658568 on Build GRCh38
rs762554788 Chr.7: 106658289-106658289 on Build GRCh38
rs919312455 Chr.7: 106658384-106658384 on Build GRCh38
rs941223387 Chr.7: 106658555-106658555 on Build GRCh38
rs1013880330 Chr.7: 106658218-106658218 on Build GRCh38
rs903332269 Chr.7: 106658415-106658415 on Build GRCh38
rs530661978 Chr.7: 106658303-106658303 on Build GRCh38
rs997257379 Chr.7: 106658466-106658466 on Build GRCh38
rs961810368 Chr.7: 106658324-106658324 on Build GRCh38
rs961842753 Chr.7: 106658593-106658593 on Build GRCh38
rs539868617 Chr.7: 106658478-106658478 on Build GRCh38
rs899891545 Chr.7: 106658429-106658429 on Build GRCh38
rs981139676 Chr.7: 106658349-106658350 on Build GRCh38
rs568155766 Chr.7: 106658293-106658293 on Build GRCh38
rs745337544 Chr.7: 106658531-106658531 on Build GRCh38
rs1041946768 Chr.7: 106658400-106658400 on Build GRCh38
rs771724705 Chr.7: 106658606-106658606 on Build GRCh38
rs1035847871 Chr.7: 106658596-106658596 on Build GRCh38
rs1023987589 Chr.7: 106658220-106658220 on Build GRCh38
rs1033164258 Chr.7: 106658301-106658301 on Build GRCh38
rs1045362123 Chr.7: 106658614-106658614 on Build GRCh38
rs986392544 Chr.7: 106658339-106658339 on Build GRCh38
rs1055679347 Chr.7: 106658574-106658574 on Build GRCh38
rs949695902 Chr.7: 106658609-106658609 on Build GRCh38
rs183644189 Chr.7: 106658487-106658487 on Build GRCh38
rs755483783 Chr.7: 106658622-106658622 on Build GRCh38
rs951957146 Chr.7: 106658203-106658203 on Build GRCh38
rs570907270 Chr.7: 106658365-106658365 on Build GRCh38
rs937018651 Chr.7: 106658368-106658368 on Build GRCh38
rs761105126 Chr.7: 106658425-106658425 on Build GRCh38
rs772755816 Chr.7: 106658506-106658507 on Build GRCh38
rs149964845 Chr.7: 106658666-106658666 on Build GRCh38
rs374277889 Chr.7: 106658239-106658239 on Build GRCh38
rs998613333 Chr.7: 106658626-106658626 on Build GRCh38
rs550658086 Chr.7: 106658357-106658357 on Build GRCh38
rs764744963 Chr.7: 106658271-106658272 on Build GRCh38
rs894416987 Chr.7: 106658601-106658601 on Build GRCh38
rs566854387 Chr.7: 106658438-106658438 on Build GRCh38
rs537906338 Chr.7: 106658640-106658640 on Build GRCh38

View Full Product Details