Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 106658575-106659068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC71L
Entrez Gene ID
168455
Gene Name
coiled-coil domain containing 71-like
Gene Aliases
C7orf74
UniGene
Hs.29692
Gene Chromosome Location
Chr.7: 106656765-106661188 on Build GRCh38
Transcripts
NM_175884.4

SNP Detail

SNP ID Chromosome Location
rs546012211 Chr.7: 106658679-106658679 on Build GRCh38
rs2190093 Chr.7: 106658958-106658958 on Build GRCh38
rs1017846686 Chr.7: 106658713-106658713 on Build GRCh38
rs977061167 Chr.7: 106658988-106658988 on Build GRCh38
rs193119725 Chr.7: 106658993-106658993 on Build GRCh38
rs932745628 Chr.7: 106658871-106658873 on Build GRCh38
rs765791438 Chr.7: 106658886-106658886 on Build GRCh38
rs1033210631 Chr.7: 106658687-106658687 on Build GRCh38
rs1021329353 Chr.7: 106658968-106658968 on Build GRCh38
rs934328646 Chr.7: 106658932-106658932 on Build GRCh38
rs558255489 Chr.7: 106658646-106658646 on Build GRCh38
rs968385801 Chr.7: 106658980-106658980 on Build GRCh38
rs909293511 Chr.7: 106658834-106658834 on Build GRCh38
rs75971732 Chr.7: 106658913-106658913 on Build GRCh38
rs114981053 Chr.7: 106658802-106658802 on Build GRCh38
rs561624407 Chr.7: 106658788-106658788 on Build GRCh38
rs1008144131 Chr.7: 106658712-106658712 on Build GRCh38
rs866386203 Chr.7: 106658935-106658935 on Build GRCh38
rs144885421 Chr.7: 106658724-106658724 on Build GRCh38
rs991747017 Chr.7: 106658880-106658880 on Build GRCh38
rs554115153 Chr.7: 106658908-106658909 on Build GRCh38
rs977656006 Chr.7: 106658785-106658785 on Build GRCh38
rs866962462 Chr.7: 106659005-106659005 on Build GRCh38
rs550371614 Chr.7: 106658849-106658849 on Build GRCh38
rs78890690 Chr.7: 106658778-106658778 on Build GRCh38
rs984881971 Chr.7: 106658805-106658805 on Build GRCh38
rs888430401 Chr.7: 106658881-106658881 on Build GRCh38
rs187689280 Chr.7: 106658910-106658910 on Build GRCh38
rs1054172577 Chr.7: 106658957-106658957 on Build GRCh38
rs564242813 Chr.7: 106658876-106658876 on Build GRCh38
rs902035205 Chr.7: 106658975-106658975 on Build GRCh38
rs921441361 Chr.7: 106659045-106659045 on Build GRCh38
rs568884589 Chr.7: 106659012-106659012 on Build GRCh38
rs573218391 Chr.7: 106658771-106658771 on Build GRCh38
rs915775238 Chr.7: 106658887-106658887 on Build GRCh38
rs1039205689 Chr.7: 106658973-106658973 on Build GRCh38
rs893270472 Chr.7: 106658705-106658705 on Build GRCh38
rs149043909 Chr.7: 106658922-106658922 on Build GRCh38
rs957241093 Chr.7: 106658710-106658710 on Build GRCh38
rs771724705 Chr.7: 106658606-106658606 on Build GRCh38
rs1035847871 Chr.7: 106658596-106658596 on Build GRCh38
rs1045362123 Chr.7: 106658614-106658614 on Build GRCh38
rs966295876 Chr.7: 106658738-106658738 on Build GRCh38
rs949695902 Chr.7: 106658609-106658609 on Build GRCh38
rs1048915283 Chr.7: 106658874-106658874 on Build GRCh38
rs950710456 Chr.7: 106659037-106659037 on Build GRCh38
rs755483783 Chr.7: 106658622-106658622 on Build GRCh38
rs899923989 Chr.7: 106658839-106658839 on Build GRCh38
rs149964845 Chr.7: 106658666-106658666 on Build GRCh38
rs998613333 Chr.7: 106658626-106658626 on Build GRCh38
rs1007696902 Chr.7: 106658969-106658969 on Build GRCh38
rs1006125543 Chr.7: 106658891-106658892 on Build GRCh38
rs950732500 Chr.7: 106658796-106658796 on Build GRCh38
rs1041179187 Chr.7: 106658824-106658824 on Build GRCh38
rs894416987 Chr.7: 106658601-106658601 on Build GRCh38
rs914239624 Chr.7: 106658762-106658762 on Build GRCh38
rs537906338 Chr.7: 106658640-106658640 on Build GRCh38

View Full Product Details