Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107613318-107613818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAP29
Entrez Gene ID
55973
Gene Name
B-cell receptor-associated protein 29
Gene Aliases
BAP29
UniGene
Hs.303787
Gene Chromosome Location
Chr.7: 107579977-107623317 on Build GRCh38
Transcripts
NM_001008405.2 NM_018844.3 NR_027830.1 XM_006716051.2 XM_011516406.2 XM_011516407.1 XM_011516408.2 XM_011516409.2 XM_017012436.1 XM_017012437.1 XM_017012438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4759889 c.336G>C p.M112I Chr.7: 107613360-107613360 on Build GRCh38
421646 c.641G>C p.R214T Chr.7: 107613383-107613383 on Build GRCh38
6849147 c.699T>A p.S233R Chr.7: 107613662-107613662 on Build GRCh38
1582702 c.742A>T p.I248F Chr.7: 107613705-107613705 on Build GRCh38
6610296 c.757G>T p.G253C Chr.7: 107613720-107613720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77410087 Chr.7: 107613680-107613680 on Build GRCh38
rs773979440 Chr.7: 107613409-107613409 on Build GRCh38
rs937659690 Chr.7: 107613399-107613399 on Build GRCh38
rs201006372 Chr.7: 107613440-107613440 on Build GRCh38
rs773819636 Chr.7: 107613642-107613642 on Build GRCh38
rs766693735 Chr.7: 107613764-107613765 on Build GRCh38
rs949202488 Chr.7: 107613358-107613358 on Build GRCh38
rs765934477 Chr.7: 107613402-107613402 on Build GRCh38
rs148654100 Chr.7: 107613619-107613619 on Build GRCh38
rs770584854 Chr.7: 107613742-107613742 on Build GRCh38
rs759071680 Chr.7: 107613644-107613644 on Build GRCh38
rs773539756 Chr.7: 107613768-107613768 on Build GRCh38
rs749638027 Chr.7: 107613579-107613579 on Build GRCh38
rs750736050 Chr.7: 107613448-107613448 on Build GRCh38
rs773116598 Chr.7: 107613482-107613482 on Build GRCh38
rs776650045 Chr.7: 107613364-107613364 on Build GRCh38
rs943456976 Chr.7: 107613617-107613617 on Build GRCh38
rs1008383376 Chr.7: 107613501-107613501 on Build GRCh38
rs1026985951 Chr.7: 107613763-107613763 on Build GRCh38
rs755057215 Chr.7: 107613456-107613456 on Build GRCh38
rs983233310 Chr.7: 107613649-107613650 on Build GRCh38
rs758629622 Chr.7: 107613679-107613679 on Build GRCh38
rs750801066 Chr.7: 107613677-107613677 on Build GRCh38
rs769670477 Chr.7: 107613389-107613389 on Build GRCh38
rs200184386 Chr.7: 107613685-107613685 on Build GRCh38
rs778570707 Chr.7: 107613743-107613743 on Build GRCh38
rs762360860 Chr.7: 107613676-107613676 on Build GRCh38
rs774953959 Chr.7: 107613651-107613651 on Build GRCh38
rs764599359 Chr.7: 107613419-107613419 on Build GRCh38
rs529523337 Chr.7: 107613699-107613699 on Build GRCh38
rs547002394 Chr.7: 107613555-107613555 on Build GRCh38
rs74583821 Chr.7: 107613453-107613453 on Build GRCh38
rs902025412 Chr.7: 107613716-107613716 on Build GRCh38
rs754296424 Chr.7: 107613431-107613431 on Build GRCh38
rs780858398 Chr.7: 107613455-107613455 on Build GRCh38
rs745562894 Chr.7: 107613747-107613747 on Build GRCh38
rs576177767 Chr.7: 107613650-107613699 on Build GRCh38
rs190861851 Chr.7: 107613468-107613468 on Build GRCh38
rs771694294 Chr.7: 107613748-107613748 on Build GRCh38
rs984433390 Chr.7: 107613348-107613348 on Build GRCh38
rs770622262 Chr.7: 107613631-107613631 on Build GRCh38
rs749123508 Chr.7: 107613735-107613735 on Build GRCh38
rs905145928 Chr.7: 107613391-107613391 on Build GRCh38
rs758709025 Chr.7: 107613449-107613449 on Build GRCh38
rs775043760 Chr.7: 107613750-107613750 on Build GRCh38
rs549554128 Chr.7: 107613703-107613703 on Build GRCh38
rs763404420 Chr.7: 107613772-107613772 on Build GRCh38
rs769596043 Chr.7: 107613477-107613477 on Build GRCh38
rs377663210 Chr.7: 107613418-107613418 on Build GRCh38
rs200602346 Chr.7: 107613721-107613721 on Build GRCh38
rs772850807 Chr.7: 107613390-107613390 on Build GRCh38
rs112966301 Chr.7: 107613789-107613789 on Build GRCh38
rs760107808 Chr.7: 107613762-107613762 on Build GRCh38
rs374377179 Chr.7: 107613393-107613393 on Build GRCh38
rs1054493305 Chr.7: 107613546-107613546 on Build GRCh38
rs768717062 Chr.7: 107613351-107613351 on Build GRCh38
rs142168570 Chr.7: 107613650-107613650 on Build GRCh38
rs370691308 Chr.7: 107613441-107613441 on Build GRCh38
rs748134815 Chr.7: 107613476-107613476 on Build GRCh38
rs747052665 Chr.7: 107613344-107613344 on Build GRCh38
rs868552926 Chr.7: 107613697-107613697 on Build GRCh38
rs766977316 Chr.7: 107613648-107613648 on Build GRCh38
rs538415663 Chr.7: 107613653-107613654 on Build GRCh38
rs747977259 Chr.7: 107613369-107613369 on Build GRCh38
rs770344229 Chr.7: 107613767-107613767 on Build GRCh38
rs1039112857 Chr.7: 107613664-107613664 on Build GRCh38
rs766691813 Chr.7: 107613784-107613784 on Build GRCh38

View Full Product Details