Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107903469-107903996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLD
Entrez Gene ID
1738
Gene Name
dihydrolipoamide dehydrogenase
Gene Aliases
DLDD, DLDH, E3, GCSL, LAD, PHE3
UniGene
Hs.131711
Gene Chromosome Location
Chr.7: 107891107-107921198 on Build GRCh38
Transcripts
NM_000108.4 NM_001289750.1 NM_001289751.1 NM_001289752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1083922 c.313G>T p.D105Y Chr.7: 107903523-107903523 on Build GRCh38
3255004 c.320C>T p.A107V Chr.7: 107903530-107903530 on Build GRCh38
5560223 c.298A>G p.M100V Chr.7: 107903508-107903508 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs920442260 Chr.7: 107903666-107903666 on Build GRCh38
rs552726220 Chr.7: 107903628-107903628 on Build GRCh38
rs987510839 Chr.7: 107903612-107903612 on Build GRCh38
rs754528609 Chr.7: 107903574-107903574 on Build GRCh38
rs983151451 Chr.7: 107903709-107903709 on Build GRCh38
rs573537912 Chr.7: 107903809-107903809 on Build GRCh38
rs749777272 Chr.7: 107903591-107903591 on Build GRCh38
rs565303362 Chr.7: 107903645-107903645 on Build GRCh38
rs994594761 Chr.7: 107903710-107903710 on Build GRCh38
rs765105905 Chr.7: 107903529-107903529 on Build GRCh38
rs926855738 Chr.7: 107903775-107903775 on Build GRCh38
rs796051950 Chr.7: 107903516-107903516 on Build GRCh38
rs766075550 Chr.7: 107903550-107903550 on Build GRCh38
rs149266807 Chr.7: 107903701-107903701 on Build GRCh38
rs949712492 Chr.7: 107903733-107903733 on Build GRCh38
rs935937945 Chr.7: 107903942-107903942 on Build GRCh38
rs973224187 Chr.7: 107903662-107903663 on Build GRCh38
rs117304495 Chr.7: 107903875-107903875 on Build GRCh38
rs35232221 Chr.7: 107903800-107903801 on Build GRCh38
rs1042494103 Chr.7: 107903966-107903967 on Build GRCh38
rs758078257 Chr.7: 107903532-107903532 on Build GRCh38
rs138398782 Chr.7: 107903531-107903531 on Build GRCh38
rs2158834 Chr.7: 107903872-107903872 on Build GRCh38
rs1053927543 Chr.7: 107903811-107903811 on Build GRCh38
rs376524199 Chr.7: 107903603-107903620 on Build GRCh38
rs187972465 Chr.7: 107903748-107903748 on Build GRCh38
rs751168608 Chr.7: 107903566-107903566 on Build GRCh38
rs371072449 Chr.7: 107903554-107903554 on Build GRCh38
rs781074171 Chr.7: 107903878-107903878 on Build GRCh38
rs2701043 Chr.7: 107903652-107903652 on Build GRCh38
rs893865374 Chr.7: 107903515-107903515 on Build GRCh38
rs938278452 Chr.7: 107903776-107903776 on Build GRCh38
rs958431141 Chr.7: 107903844-107903844 on Build GRCh38
rs761564212 Chr.7: 107903495-107903495 on Build GRCh38
rs992565154 Chr.7: 107903857-107903857 on Build GRCh38
rs1041605197 Chr.7: 107903509-107903509 on Build GRCh38
rs2108222 Chr.7: 107903763-107903763 on Build GRCh38
rs192797949 Chr.7: 107903867-107903867 on Build GRCh38
rs1130477 Chr.7: 107903521-107903521 on Build GRCh38
rs374458520 Chr.7: 107903579-107903579 on Build GRCh38
rs745660291 Chr.7: 107903932-107903932 on Build GRCh38
rs925858025 Chr.7: 107903916-107903916 on Build GRCh38
rs1005669434 Chr.7: 107903789-107903789 on Build GRCh38
rs752671238 Chr.7: 107903557-107903557 on Build GRCh38
rs35528839 Chr.7: 107903653-107903654 on Build GRCh38
rs11538953 Chr.7: 107903505-107903505 on Build GRCh38
rs548760589 Chr.7: 107903778-107903778 on Build GRCh38
rs889865883 Chr.7: 107903608-107903608 on Build GRCh38
rs945939975 Chr.7: 107903507-107903507 on Build GRCh38

View Full Product Details