Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 108884315-108884815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf66
Entrez Gene ID
154907
Gene Name
chromosome 7 open reading frame 66
Gene Aliases
-
UniGene
Hs.276325
Gene Chromosome Location
Chr.7: 108883975-108884587 on Build GRCh38
Transcripts
NM_001024607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616661 c.37C>T p.Q13* Chr.7: 108884496-108884496 on Build GRCh38
4475585 c.19C>T p.P7S Chr.7: 108884514-108884514 on Build GRCh38
5799427 c.8C>A p.A3D Chr.7: 108884525-108884525 on Build GRCh38
452150 c.12G>C p.V4V Chr.7: 108884521-108884521 on Build GRCh38
4556382 c.68G>A p.R23K Chr.7: 108884465-108884465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759467504 Chr.7: 108884458-108884458 on Build GRCh38
rs778961883 Chr.7: 108884408-108884408 on Build GRCh38
rs568999702 Chr.7: 108884646-108884646 on Build GRCh38
rs909661178 Chr.7: 108884689-108884689 on Build GRCh38
rs75599531 Chr.7: 108884471-108884471 on Build GRCh38
rs374820012 Chr.7: 108884405-108884405 on Build GRCh38
rs951017705 Chr.7: 108884647-108884647 on Build GRCh38
rs998079972 Chr.7: 108884580-108884580 on Build GRCh38
rs867907077 Chr.7: 108884587-108884587 on Build GRCh38
rs751839363 Chr.7: 108884579-108884579 on Build GRCh38
rs755106698 Chr.7: 108884400-108884400 on Build GRCh38
rs748494645 Chr.7: 108884388-108884388 on Build GRCh38
rs922423144 Chr.7: 108884371-108884371 on Build GRCh38
rs1018400714 Chr.7: 108884352-108884352 on Build GRCh38
rs760420832 Chr.7: 108884536-108884536 on Build GRCh38
rs928188151 Chr.7: 108884778-108884778 on Build GRCh38
rs1056893898 Chr.7: 108884478-108884478 on Build GRCh38
rs778178126 Chr.7: 108884479-108884479 on Build GRCh38
rs997926171 Chr.7: 108884632-108884632 on Build GRCh38
rs982594735 Chr.7: 108884643-108884643 on Build GRCh38
rs764059683 Chr.7: 108884393-108884394 on Build GRCh38
rs752788360 Chr.7: 108884569-108884569 on Build GRCh38
rs765367894 Chr.7: 108884464-108884464 on Build GRCh38
rs761452089 Chr.7: 108884383-108884383 on Build GRCh38
rs758831323 Chr.7: 108884413-108884413 on Build GRCh38
rs11404723 Chr.7: 108884387-108884388 on Build GRCh38
rs962551745 Chr.7: 108884720-108884720 on Build GRCh38
rs899608595 Chr.7: 108884590-108884590 on Build GRCh38
rs768589012 Chr.7: 108884545-108884545 on Build GRCh38
rs776375398 Chr.7: 108884546-108884546 on Build GRCh38
rs755802139 Chr.7: 108884399-108884400 on Build GRCh38
rs748710638 Chr.7: 108884419-108884419 on Build GRCh38
rs183230116 Chr.7: 108884567-108884567 on Build GRCh38
rs768221459 Chr.7: 108884421-108884421 on Build GRCh38
rs796670233 Chr.7: 108884388-108884389 on Build GRCh38
rs374850391 Chr.7: 108884387-108884387 on Build GRCh38
rs1041249204 Chr.7: 108884379-108884379 on Build GRCh38
rs1007658153 Chr.7: 108884516-108884516 on Build GRCh38
rs904033201 Chr.7: 108884515-108884515 on Build GRCh38
rs770262147 Chr.7: 108884504-108884504 on Build GRCh38
rs778288051 Chr.7: 108884416-108884430 on Build GRCh38
rs532788304 Chr.7: 108884441-108884441 on Build GRCh38
rs1029925424 Chr.7: 108884588-108884588 on Build GRCh38
rs991193815 Chr.7: 108884709-108884709 on Build GRCh38
rs962447204 Chr.7: 108884694-108884694 on Build GRCh38
rs376515459 Chr.7: 108884385-108884385 on Build GRCh38
rs781770947 Chr.7: 108884417-108884417 on Build GRCh38
rs939890089 Chr.7: 108884375-108884375 on Build GRCh38
rs760074695 Chr.7: 108884769-108884769 on Build GRCh38
rs935528326 Chr.7: 108884783-108884783 on Build GRCh38
rs761514610 Chr.7: 108884425-108884425 on Build GRCh38
rs763608129 Chr.7: 108884380-108884380 on Build GRCh38
rs752964189 Chr.7: 108884409-108884409 on Build GRCh38
rs747697932 Chr.7: 108884511-108884511 on Build GRCh38
rs771780211 Chr.7: 108884426-108884426 on Build GRCh38
rs202099108 Chr.7: 108884451-108884451 on Build GRCh38
rs566769968 Chr.7: 108884505-108884505 on Build GRCh38
rs145510967 Chr.7: 108884456-108884456 on Build GRCh38
rs762751537 Chr.7: 108884391-108884392 on Build GRCh38
rs141228190 Chr.7: 108884437-108884437 on Build GRCh38
rs765754917 Chr.7: 108884400-108884401 on Build GRCh38
rs773817613 Chr.7: 108884423-108884423 on Build GRCh38
rs779457018 Chr.7: 108884406-108884406 on Build GRCh38
rs759500066 Chr.7: 108884548-108884548 on Build GRCh38
rs1029316868 Chr.7: 108884395-108884396 on Build GRCh38
rs909569394 Chr.7: 108884404-108884404 on Build GRCh38
rs757731554 Chr.7: 108884481-108884481 on Build GRCh38
rs368225324 Chr.7: 108884575-108884575 on Build GRCh38
rs530619258 Chr.7: 108884392-108884392 on Build GRCh38
rs982502921 Chr.7: 108884398-108884398 on Build GRCh38
rs963765517 Chr.7: 108884365-108884365 on Build GRCh38
rs746404116 Chr.7: 108884503-108884503 on Build GRCh38
rs771208852 Chr.7: 108884540-108884540 on Build GRCh38
rs371483483 Chr.7: 108884337-108884337 on Build GRCh38
rs199829152 Chr.7: 108884395-108884395 on Build GRCh38
rs764374703 Chr.7: 108884576-108884576 on Build GRCh38
rs916852614 Chr.7: 108884429-108884429 on Build GRCh38
rs774087048 Chr.7: 108884382-108884382 on Build GRCh38
rs893876270 Chr.7: 108884532-108884532 on Build GRCh38
rs748725646 Chr.7: 108884412-108884413 on Build GRCh38
rs868236439 Chr.7: 108884737-108884737 on Build GRCh38
rs941137898 Chr.7: 108884406-108884406 on Build GRCh38
rs915543431 Chr.7: 108884725-108884725 on Build GRCh38
rs753422146 Chr.7: 108884401-108884402 on Build GRCh38
rs1029780001 Chr.7: 108884636-108884636 on Build GRCh38
rs747172103 Chr.7: 108884503-108884504 on Build GRCh38
rs767363538 Chr.7: 108884398-108884399 on Build GRCh38
rs767314349 Chr.7: 108884384-108884384 on Build GRCh38
rs762575179 Chr.7: 108884378-108884378 on Build GRCh38
rs906672949 Chr.7: 108884474-108884474 on Build GRCh38
rs368132144 Chr.7: 108884405-108884405 on Build GRCh38
rs753838364 Chr.7: 108884399-108884399 on Build GRCh38
rs879072878 Chr.7: 108884388-108884389 on Build GRCh38
rs751926747 Chr.7: 108884480-108884480 on Build GRCh38
rs1016839577 Chr.7: 108884711-108884711 on Build GRCh38
rs778265991 Chr.7: 108884415-108884415 on Build GRCh38
rs752773814 Chr.7: 108884466-108884466 on Build GRCh38
rs761581900 Chr.7: 108884397-108884398 on Build GRCh38
rs774319101 Chr.7: 108884394-108884395 on Build GRCh38
rs142526547 Chr.7: 108884513-108884513 on Build GRCh38
rs36004991 Chr.7: 108884404-108884405 on Build GRCh38
rs760510505 Chr.7: 108884439-108884439 on Build GRCh38
rs10278613 Chr.7: 108884736-108884736 on Build GRCh38
rs772872504 Chr.7: 108884514-108884514 on Build GRCh38
rs200301270 Chr.7: 108884413-108884413 on Build GRCh38
rs1052541985 Chr.7: 108884787-108884787 on Build GRCh38
rs999910532 Chr.7: 108884342-108884342 on Build GRCh38
rs781543768 Chr.7: 108884622-108884622 on Build GRCh38
rs367868708 Chr.7: 108884501-108884501 on Build GRCh38
rs555536391 Chr.7: 108884621-108884621 on Build GRCh38
rs747441925 Chr.7: 108884416-108884416 on Build GRCh38

View Full Product Details