Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 112484734-112485240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSMEM1
Entrez Gene ID
286006
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
C7orf53
UniGene
Hs.396189
Gene Chromosome Location
Chr.7: 112480219-112491062 on Build GRCh38
Transcripts
NM_001134468.1 NM_182597.2 XM_011516074.1 XM_011516075.2 XM_011516076.2 XM_017012028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
452159 c.35G>A p.G12D Chr.7: 112484851-112484851 on Build GRCh38
6704720 c.122T>C p.L41P Chr.7: 112484938-112484938 on Build GRCh38
1084123 c.99C>T p.L33L Chr.7: 112484915-112484915 on Build GRCh38
6704721 c.122T>C p.L41P Chr.7: 112484938-112484938 on Build GRCh38
6447424 c.109G>A p.G37R Chr.7: 112484925-112484925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761669301 Chr.7: 112484898-112484898 on Build GRCh38
rs866482937 Chr.7: 112484810-112484810 on Build GRCh38
rs377646980 Chr.7: 112485012-112485012 on Build GRCh38
rs562601406 Chr.7: 112485200-112485200 on Build GRCh38
rs765479355 Chr.7: 112484930-112484930 on Build GRCh38
rs1045010430 Chr.7: 112485101-112485101 on Build GRCh38
rs748276941 Chr.7: 112484925-112484925 on Build GRCh38
rs1053638178 Chr.7: 112485192-112485192 on Build GRCh38
rs779372931 Chr.7: 112484954-112484954 on Build GRCh38
rs544369703 Chr.7: 112485198-112485198 on Build GRCh38
rs201708903 Chr.7: 112484798-112484798 on Build GRCh38
rs751836961 Chr.7: 112484900-112484903 on Build GRCh38
rs778219151 Chr.7: 112484834-112484834 on Build GRCh38
rs566776678 Chr.7: 112485019-112485019 on Build GRCh38
rs912601302 Chr.7: 112484790-112484790 on Build GRCh38
rs754005048 Chr.7: 112484774-112484774 on Build GRCh38
rs34322208 Chr.7: 112484889-112484889 on Build GRCh38
rs757785750 Chr.7: 112484953-112484953 on Build GRCh38
rs201282935 Chr.7: 112484976-112484976 on Build GRCh38
rs554612043 Chr.7: 112485069-112485069 on Build GRCh38
rs370303982 Chr.7: 112484778-112484778 on Build GRCh38
rs771990298 Chr.7: 112484966-112484966 on Build GRCh38
rs186898413 Chr.7: 112484912-112484912 on Build GRCh38
rs557067896 Chr.7: 112485036-112485036 on Build GRCh38
rs921803690 Chr.7: 112484756-112484756 on Build GRCh38
rs534126686 Chr.7: 112485098-112485098 on Build GRCh38
rs772523458 Chr.7: 112484830-112484830 on Build GRCh38
rs771766388 Chr.7: 112485119-112485119 on Build GRCh38
rs879883371 Chr.7: 112485031-112485032 on Build GRCh38
rs934616533 Chr.7: 112484962-112484962 on Build GRCh38
rs185117670 Chr.7: 112485022-112485022 on Build GRCh38
rs766365330 Chr.7: 112484972-112484972 on Build GRCh38
rs530055326 Chr.7: 112485191-112485191 on Build GRCh38
rs373176422 Chr.7: 112485208-112485208 on Build GRCh38
rs573138398 Chr.7: 112485071-112485071 on Build GRCh38
rs577012631 Chr.7: 112485149-112485149 on Build GRCh38
rs866929147 Chr.7: 112484811-112484811 on Build GRCh38
rs190550220 Chr.7: 112485056-112485056 on Build GRCh38
rs774125154 Chr.7: 112484965-112484965 on Build GRCh38
rs780241115 Chr.7: 112484770-112484770 on Build GRCh38
rs562538485 Chr.7: 112484981-112484981 on Build GRCh38
rs914564375 Chr.7: 112485218-112485218 on Build GRCh38
rs544719581 Chr.7: 112484980-112484980 on Build GRCh38
rs1007411644 Chr.7: 112485032-112485033 on Build GRCh38
rs573187484 Chr.7: 112484946-112484946 on Build GRCh38
rs769170706 Chr.7: 112484864-112484864 on Build GRCh38
rs997146964 Chr.7: 112484970-112484970 on Build GRCh38
rs779092774 Chr.7: 112484796-112484796 on Build GRCh38
rs758683437 Chr.7: 112484987-112484987 on Build GRCh38
rs759626332 Chr.7: 112484977-112484977 on Build GRCh38
rs949389415 Chr.7: 112485073-112485073 on Build GRCh38
rs764475695 Chr.7: 112484817-112484818 on Build GRCh38
rs199748260 Chr.7: 112484967-112484967 on Build GRCh38
rs796923957 Chr.7: 112485166-112485166 on Build GRCh38
rs3095027 Chr.7: 112485034-112485034 on Build GRCh38
rs748823279 Chr.7: 112484872-112484872 on Build GRCh38
rs200241326 Chr.7: 112484929-112484929 on Build GRCh38
rs1049060680 Chr.7: 112485184-112485184 on Build GRCh38
rs540022698 Chr.7: 112484958-112484958 on Build GRCh38
rs540555148 Chr.7: 112484999-112484999 on Build GRCh38
rs972256628 Chr.7: 112485039-112485039 on Build GRCh38
rs542941761 Chr.7: 112484926-112484926 on Build GRCh38
rs781239794 Chr.7: 112484923-112484923 on Build GRCh38
rs192102447 Chr.7: 112484924-112484924 on Build GRCh38
rs760516049 Chr.7: 112485148-112485148 on Build GRCh38
rs3095028 Chr.7: 112485044-112485044 on Build GRCh38
rs548186414 Chr.7: 112485014-112485014 on Build GRCh38
rs951715605 Chr.7: 112485047-112485047 on Build GRCh38
rs931813188 Chr.7: 112484765-112484765 on Build GRCh38
rs778166066 Chr.7: 112484944-112484944 on Build GRCh38
rs1019072132 Chr.7: 112485033-112485033 on Build GRCh38
rs768368529 Chr.7: 112484882-112484882 on Build GRCh38
rs917904847 Chr.7: 112484876-112484876 on Build GRCh38
rs552461931 Chr.7: 112485024-112485024 on Build GRCh38
rs17159708 Chr.7: 112485127-112485127 on Build GRCh38
rs752952403 Chr.7: 112484932-112484932 on Build GRCh38
rs775130445 Chr.7: 112484865-112484865 on Build GRCh38
rs1045123961 Chr.7: 112484907-112484907 on Build GRCh38
rs758736439 Chr.7: 112484943-112484943 on Build GRCh38

View Full Product Details