Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 112490484-112490994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSMEM1
Entrez Gene ID
286006
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
C7orf53
UniGene
Hs.396189
Gene Chromosome Location
Chr.7: 112480219-112491062 on Build GRCh38
Transcripts
NM_001134468.1 NM_182597.2 XM_011516074.1 XM_011516075.2 XM_011516076.2 XM_017012028.1

SNP Detail

SNP ID Chromosome Location
rs778361765 Chr.7: 112490599-112490599 on Build GRCh38
rs191297180 Chr.7: 112490896-112490896 on Build GRCh38
rs759561921 Chr.7: 112490906-112490906 on Build GRCh38
rs908759601 Chr.7: 112490763-112490763 on Build GRCh38
rs546960757 Chr.7: 112490530-112490530 on Build GRCh38
rs997668457 Chr.7: 112490883-112490883 on Build GRCh38
rs1000736857 Chr.7: 112490608-112490608 on Build GRCh38
rs530316040 Chr.7: 112490715-112490715 on Build GRCh38
rs1032182021 Chr.7: 112490620-112490620 on Build GRCh38
rs1022483128 Chr.7: 112490594-112490594 on Build GRCh38
rs901566562 Chr.7: 112490563-112490563 on Build GRCh38
rs988520397 Chr.7: 112490628-112490628 on Build GRCh38
rs966282651 Chr.7: 112490627-112490627 on Build GRCh38
rs552505120 Chr.7: 112490933-112490933 on Build GRCh38
rs113619391 Chr.7: 112490924-112490924 on Build GRCh38
rs918025013 Chr.7: 112490650-112490650 on Build GRCh38
rs116267413 Chr.7: 112490556-112490556 on Build GRCh38
rs770299886 Chr.7: 112490576-112490576 on Build GRCh38
rs887438642 Chr.7: 112490526-112490526 on Build GRCh38
rs1052217424 Chr.7: 112490570-112490570 on Build GRCh38
rs576941948 Chr.7: 112490557-112490557 on Build GRCh38
rs144330341 Chr.7: 112490843-112490843 on Build GRCh38
rs981398598 Chr.7: 112490855-112490855 on Build GRCh38
rs956807579 Chr.7: 112490934-112490934 on Build GRCh38
rs541535472 Chr.7: 112490853-112490853 on Build GRCh38
rs997383348 Chr.7: 112490575-112490575 on Build GRCh38
rs926768738 Chr.7: 112490704-112490704 on Build GRCh38
rs971199856 Chr.7: 112490660-112490660 on Build GRCh38
rs1036420978 Chr.7: 112490835-112490835 on Build GRCh38
rs1044918737 Chr.7: 112490537-112490537 on Build GRCh38
rs749810969 Chr.7: 112490819-112490819 on Build GRCh38
rs1050262901 Chr.7: 112490891-112490891 on Build GRCh38
rs185067720 Chr.7: 112490621-112490621 on Build GRCh38
rs892317146 Chr.7: 112490571-112490571 on Build GRCh38
rs544174575 Chr.7: 112490601-112490601 on Build GRCh38
rs948746400 Chr.7: 112490532-112490533 on Build GRCh38
rs980990002 Chr.7: 112490673-112490673 on Build GRCh38
rs931591637 Chr.7: 112490713-112490713 on Build GRCh38
rs780739648 Chr.7: 112490545-112490545 on Build GRCh38
rs952324043 Chr.7: 112490765-112490765 on Build GRCh38
rs928677923 Chr.7: 112490888-112490888 on Build GRCh38

View Full Product Details