Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 114538306-114538812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP2
Entrez Gene ID
93986
Gene Name
forkhead box P2
Gene Aliases
CAGH44, SPCH1, TNRC10
UniGene
Hs.282787
Gene Chromosome Location
Chr.7: 114086310-114693772 on Build GRCh38
Transcripts
NM_001172766.2 NM_001172767.2 NM_014491.3 NM_148898.3 NM_148899.3 NM_148900.3 NR_033766.1 NR_033767.1 XM_017012801.1

SNP Detail

SNP ID Chromosome Location
rs988114302 Chr.7: 114538716-114538716 on Build GRCh38
rs905521418 Chr.7: 114538433-114538433 on Build GRCh38
rs918163413 Chr.7: 114538720-114538720 on Build GRCh38
rs935436437 Chr.7: 114538647-114538647 on Build GRCh38
rs1025504044 Chr.7: 114538508-114538508 on Build GRCh38
rs896718324 Chr.7: 114538424-114538424 on Build GRCh38
rs979494955 Chr.7: 114538675-114538675 on Build GRCh38
rs745425231 Chr.7: 114538515-114538515 on Build GRCh38
rs10486023 Chr.7: 114538598-114538598 on Build GRCh38
rs538494977 Chr.7: 114538561-114538561 on Build GRCh38
rs1045196660 Chr.7: 114538453-114538453 on Build GRCh38
rs886762871 Chr.7: 114538543-114538543 on Build GRCh38
rs904948511 Chr.7: 114538467-114538467 on Build GRCh38
rs771443572 Chr.7: 114538562-114538562 on Build GRCh38
rs536323658 Chr.7: 114538682-114538682 on Build GRCh38
rs956703049 Chr.7: 114538686-114538686 on Build GRCh38
rs1024355709 Chr.7: 114538432-114538432 on Build GRCh38
rs949589986 Chr.7: 114538687-114538687 on Build GRCh38
rs979868275 Chr.7: 114538610-114538610 on Build GRCh38
rs905466717 Chr.7: 114538760-114538760 on Build GRCh38
rs931085956 Chr.7: 114538766-114538766 on Build GRCh38
rs953629180 Chr.7: 114538662-114538662 on Build GRCh38
rs1052564885 Chr.7: 114538412-114538412 on Build GRCh38
rs772147806 Chr.7: 114538587-114538587 on Build GRCh38
rs918143213 Chr.7: 114538428-114538428 on Build GRCh38
rs144902541 Chr.7: 114538775-114538775 on Build GRCh38
rs949627731 Chr.7: 114538727-114538727 on Build GRCh38
rs566593208 Chr.7: 114538372-114538372 on Build GRCh38
rs771626382 Chr.7: 114538417-114538417 on Build GRCh38
rs1048145353 Chr.7: 114538786-114538786 on Build GRCh38
rs546566867 Chr.7: 114538637-114538637 on Build GRCh38
rs1045674210 Chr.7: 114538747-114538747 on Build GRCh38
rs1003806292 Chr.7: 114538556-114538556 on Build GRCh38
rs896665846 Chr.7: 114538665-114538665 on Build GRCh38
rs571513044 Chr.7: 114538473-114538473 on Build GRCh38
rs112629574 Chr.7: 114538486-114538486 on Build GRCh38
rs560895211 Chr.7: 114538429-114538429 on Build GRCh38
rs777630100 Chr.7: 114538352-114538352 on Build GRCh38
rs995258211 Chr.7: 114538476-114538476 on Build GRCh38
rs1001648195 Chr.7: 114538460-114538460 on Build GRCh38
rs772828997 Chr.7: 114538413-114538413 on Build GRCh38
rs550752337 Chr.7: 114538575-114538575 on Build GRCh38
rs765422447 Chr.7: 114538654-114538657 on Build GRCh38
rs760576791 Chr.7: 114538606-114538606 on Build GRCh38
rs1019508690 Chr.7: 114538596-114538596 on Build GRCh38
rs986745289 Chr.7: 114538573-114538573 on Build GRCh38
rs771176730 Chr.7: 114538419-114538419 on Build GRCh38
rs566868051 Chr.7: 114538748-114538748 on Build GRCh38

View Full Product Details