Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 114644461-114644966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP2
Entrez Gene ID
93986
Gene Name
forkhead box P2
Gene Aliases
CAGH44, SPCH1, TNRC10
UniGene
Hs.282787
Gene Chromosome Location
Chr.7: 114086310-114693772 on Build GRCh38
Transcripts
NM_001172766.2 NM_001172767.2 NM_014491.3 NM_148898.3 NM_148899.3 NM_148900.3 NR_033766.1 NR_033767.1 XM_017012801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3877403 c.788C>T p.A263V Chr.7: 114644708-114644708 on Build GRCh38
7264267 c.869+5_869+6insT p.? Chr.7: 114644794-114644795 on Build GRCh38
5406263 c.812G>A p.G271E Chr.7: 114644732-114644732 on Build GRCh38
1252733 c.1081A>G p.T361A Chr.7: 114644701-114644701 on Build GRCh38
107926 c.1156G>A p.G386R Chr.7: 114644776-114644776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769128489 Chr.7: 114644642-114644642 on Build GRCh38
rs1029288724 Chr.7: 114644936-114644936 on Build GRCh38
rs775325812 Chr.7: 114644816-114644816 on Build GRCh38
rs776004218 Chr.7: 114644797-114644797 on Build GRCh38
rs1012392115 Chr.7: 114644571-114644571 on Build GRCh38
rs757270050 Chr.7: 114644696-114644696 on Build GRCh38
rs775138791 Chr.7: 114644669-114644669 on Build GRCh38
rs748597582 Chr.7: 114644775-114644775 on Build GRCh38
rs749234223 Chr.7: 114644803-114644803 on Build GRCh38
rs752446566 Chr.7: 114644748-114644748 on Build GRCh38
rs1042552641 Chr.7: 114644495-114644495 on Build GRCh38
rs751555660 Chr.7: 114644559-114644559 on Build GRCh38
rs749710055 Chr.7: 114644641-114644641 on Build GRCh38
rs193095218 Chr.7: 114644572-114644572 on Build GRCh38
rs141431326 Chr.7: 114644776-114644776 on Build GRCh38
rs373965830 Chr.7: 114644667-114644667 on Build GRCh38
rs573566628 Chr.7: 114644718-114644718 on Build GRCh38
rs767548655 Chr.7: 114644823-114644823 on Build GRCh38
rs756276085 Chr.7: 114644703-114644703 on Build GRCh38
rs372704196 Chr.7: 114644693-114644693 on Build GRCh38
rs543528343 Chr.7: 114644796-114644796 on Build GRCh38
rs962838374 Chr.7: 114644590-114644590 on Build GRCh38
rs184212755 Chr.7: 114644806-114644806 on Build GRCh38
rs898533167 Chr.7: 114644920-114644920 on Build GRCh38
rs548176002 Chr.7: 114644864-114644864 on Build GRCh38
rs768388259 Chr.7: 114644814-114644814 on Build GRCh38
rs1015159797 Chr.7: 114644910-114644910 on Build GRCh38
rs922328947 Chr.7: 114644640-114644641 on Build GRCh38
rs895137856 Chr.7: 114644536-114644536 on Build GRCh38
rs1005116903 Chr.7: 114644909-114644909 on Build GRCh38
rs902736096 Chr.7: 114644506-114644506 on Build GRCh38
rs200229512 Chr.7: 114644668-114644668 on Build GRCh38
rs955241027 Chr.7: 114644620-114644620 on Build GRCh38
rs747854322 Chr.7: 114644606-114644606 on Build GRCh38
rs939563150 Chr.7: 114644814-114644814 on Build GRCh38
rs1012753570 Chr.7: 114644863-114644863 on Build GRCh38
rs763921047 Chr.7: 114644820-114644820 on Build GRCh38
rs769311686 Chr.7: 114644807-114644807 on Build GRCh38
rs762715058 Chr.7: 114644819-114644819 on Build GRCh38
rs767291302 Chr.7: 114644707-114644707 on Build GRCh38
rs1056621601 Chr.7: 114644852-114644852 on Build GRCh38
rs761834466 Chr.7: 114644822-114644822 on Build GRCh38
rs887982445 Chr.7: 114644870-114644870 on Build GRCh38
rs746016390 Chr.7: 114644635-114644635 on Build GRCh38
rs368908035 Chr.7: 114644663-114644663 on Build GRCh38
rs1035271062 Chr.7: 114644525-114644525 on Build GRCh38
rs966282020 Chr.7: 114644918-114644918 on Build GRCh38
rs748238991 Chr.7: 114644560-114644560 on Build GRCh38
rs188322711 Chr.7: 114644919-114644919 on Build GRCh38
rs895243512 Chr.7: 114644546-114644546 on Build GRCh38
rs117412029 Chr.7: 114644607-114644607 on Build GRCh38
rs779272638 Chr.7: 114644742-114644742 on Build GRCh38
rs972536154 Chr.7: 114644601-114644601 on Build GRCh38
rs766204783 Chr.7: 114644522-114644522 on Build GRCh38
rs780371962 Chr.7: 114644716-114644716 on Build GRCh38
rs556747037 Chr.7: 114644636-114644636 on Build GRCh38
rs895426281 Chr.7: 114644854-114644854 on Build GRCh38
rs377594557 Chr.7: 114644667-114644669 on Build GRCh38
rs562804094 Chr.7: 114644825-114644825 on Build GRCh38
rs895192939 Chr.7: 114644853-114644853 on Build GRCh38
rs201588163 Chr.7: 114644821-114644821 on Build GRCh38
rs757389374 Chr.7: 114644664-114644666 on Build GRCh38
rs748687515 Chr.7: 114644661-114644661 on Build GRCh38
rs200003519 Chr.7: 114644688-114644688 on Build GRCh38
rs575204161 Chr.7: 114644664-114644664 on Build GRCh38
rs945804979 Chr.7: 114644702-114644702 on Build GRCh38
rs754902556 Chr.7: 114644581-114644581 on Build GRCh38
rs1012270858 Chr.7: 114644539-114644539 on Build GRCh38
rs767426318 Chr.7: 114644700-114644700 on Build GRCh38
rs530040660 Chr.7: 114644826-114644826 on Build GRCh38
rs531363183 Chr.7: 114644681-114644681 on Build GRCh38
rs779895981 Chr.7: 114644802-114644803 on Build GRCh38
rs201819725 Chr.7: 114644800-114644800 on Build GRCh38
rs750582959 Chr.7: 114644701-114644701 on Build GRCh38
rs961053168 Chr.7: 114644621-114644621 on Build GRCh38
rs755297474 Chr.7: 114644723-114644723 on Build GRCh38
rs776218490 Chr.7: 114644665-114644665 on Build GRCh38
rs549337617 Chr.7: 114644654-114644654 on Build GRCh38
rs780277508 Chr.7: 114644637-114644637 on Build GRCh38
rs774876526 Chr.7: 114644643-114644643 on Build GRCh38
rs1056496576 Chr.7: 114644849-114644849 on Build GRCh38
rs868593823 Chr.7: 114644817-114644817 on Build GRCh38

View Full Product Details