Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116525376-116525900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAV1
Entrez Gene ID
857
Gene Name
caveolin 1
Gene Aliases
BSCL3, CGL3, LCCNS, MSTP085, PPH3, VIP21
UniGene
Hs.74034
Gene Chromosome Location
Chr.7: 116524785-116561185 on Build GRCh38
Transcripts
NM_001172895.1 NM_001753.4

SNP Detail

SNP ID Chromosome Location
rs887002099 Chr.7: 116525726-116525726 on Build GRCh38
rs989152912 Chr.7: 116525530-116525530 on Build GRCh38
rs984951777 Chr.7: 116525816-116525816 on Build GRCh38
rs376559796 Chr.7: 116525619-116525619 on Build GRCh38
rs917133495 Chr.7: 116525536-116525536 on Build GRCh38
rs927053877 Chr.7: 116525585-116525585 on Build GRCh38
rs555648389 Chr.7: 116525876-116525876 on Build GRCh38
rs1041035838 Chr.7: 116525687-116525687 on Build GRCh38
rs1010036279 Chr.7: 116525755-116525755 on Build GRCh38
rs796633241 Chr.7: 116525834-116525834 on Build GRCh38
rs914791856 Chr.7: 116525623-116525623 on Build GRCh38
rs1019171784 Chr.7: 116525449-116525449 on Build GRCh38
rs550455588 Chr.7: 116525475-116525475 on Build GRCh38
rs1003053416 Chr.7: 116525431-116525431 on Build GRCh38
rs114523248 Chr.7: 116525872-116525872 on Build GRCh38
rs535081361 Chr.7: 116525610-116525610 on Build GRCh38
rs906931539 Chr.7: 116525419-116525419 on Build GRCh38
rs979763305 Chr.7: 116525569-116525569 on Build GRCh38
rs965266286 Chr.7: 116525517-116525517 on Build GRCh38
rs938890503 Chr.7: 116525703-116525703 on Build GRCh38
rs115877240 Chr.7: 116525455-116525455 on Build GRCh38
rs902087812 Chr.7: 116525707-116525707 on Build GRCh38
rs568633424 Chr.7: 116525512-116525512 on Build GRCh38
rs1033243404 Chr.7: 116525510-116525510 on Build GRCh38
rs928787647 Chr.7: 116525645-116525645 on Build GRCh38
rs1032195700 Chr.7: 116525542-116525542 on Build GRCh38
rs1041844141 Chr.7: 116525629-116525629 on Build GRCh38
rs114917769 Chr.7: 116525515-116525515 on Build GRCh38
rs539136319 Chr.7: 116525806-116525806 on Build GRCh38
rs907585108 Chr.7: 116525825-116525825 on Build GRCh38
rs903974259 Chr.7: 116525712-116525712 on Build GRCh38
rs956069113 Chr.7: 116525754-116525754 on Build GRCh38
rs995548663 Chr.7: 116525717-116525717 on Build GRCh38
rs1031060935 Chr.7: 116525730-116525730 on Build GRCh38
rs901844796 Chr.7: 116525450-116525450 on Build GRCh38
rs996171153 Chr.7: 116525709-116525709 on Build GRCh38
rs951286330 Chr.7: 116525503-116525503 on Build GRCh38
rs930367788 Chr.7: 116525605-116525605 on Build GRCh38
rs1040542452 Chr.7: 116525866-116525866 on Build GRCh38
rs1000916925 Chr.7: 116525725-116525725 on Build GRCh38
rs946314804 Chr.7: 116525627-116525627 on Build GRCh38
rs1005561022 Chr.7: 116525411-116525411 on Build GRCh38
rs914592278 Chr.7: 116525851-116525851 on Build GRCh38
rs926959575 Chr.7: 116525793-116525793 on Build GRCh38
rs968099566 Chr.7: 116525487-116525487 on Build GRCh38
rs976560477 Chr.7: 116525525-116525525 on Build GRCh38
rs978586910 Chr.7: 116525500-116525500 on Build GRCh38
rs940832561 Chr.7: 116525630-116525630 on Build GRCh38
rs1029452814 Chr.7: 116525811-116525811 on Build GRCh38
rs77735289 Chr.7: 116525835-116525835 on Build GRCh38
rs779181151 Chr.7: 116525797-116525797 on Build GRCh38
rs979794195 Chr.7: 116525774-116525774 on Build GRCh38
rs923540720 Chr.7: 116525871-116525871 on Build GRCh38
rs967374000 Chr.7: 116525860-116525860 on Build GRCh38
rs547659828 Chr.7: 116525625-116525625 on Build GRCh38
rs894118214 Chr.7: 116525454-116525454 on Build GRCh38
rs1024682893 Chr.7: 116525768-116525768 on Build GRCh38
rs565465657 Chr.7: 116525770-116525770 on Build GRCh38
rs1050366523 Chr.7: 116525711-116525711 on Build GRCh38
rs557389708 Chr.7: 116525829-116525829 on Build GRCh38
rs949759362 Chr.7: 116525568-116525568 on Build GRCh38
rs922075519 Chr.7: 116525604-116525606 on Build GRCh38
rs1050606376 Chr.7: 116525873-116525873 on Build GRCh38
rs903882493 Chr.7: 116525878-116525878 on Build GRCh38
rs953495643 Chr.7: 116525821-116525821 on Build GRCh38
rs946179112 Chr.7: 116525403-116525403 on Build GRCh38
rs2742125 Chr.7: 116525409-116525409 on Build GRCh38

View Full Product Details