Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116560819-116561326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAV1
Entrez Gene ID
857
Gene Name
caveolin 1
Gene Aliases
BSCL3, CGL3, LCCNS, MSTP085, PPH3, VIP21
UniGene
Hs.74034
Gene Chromosome Location
Chr.7: 116524785-116561185 on Build GRCh38
Transcripts
NM_001172895.1 NM_001172896.1 NM_001172897.1 NM_001753.4

SNP Detail

SNP ID Chromosome Location
rs34397467 Chr.7: 116561156-116561156 on Build GRCh38
rs1050618231 Chr.7: 116561036-116561036 on Build GRCh38
rs543466302 Chr.7: 116561233-116561233 on Build GRCh38
rs1038332070 Chr.7: 116561054-116561054 on Build GRCh38
rs369427253 Chr.7: 116561040-116561040 on Build GRCh38
rs1049375 Chr.7: 116560846-116560846 on Build GRCh38
rs542018638 Chr.7: 116561153-116561153 on Build GRCh38
rs796502851 Chr.7: 116560996-116560996 on Build GRCh38
rs757217243 Chr.7: 116560980-116560980 on Build GRCh38
rs1024973120 Chr.7: 116561100-116561100 on Build GRCh38
rs187498039 Chr.7: 116561174-116561174 on Build GRCh38
rs981241763 Chr.7: 116561213-116561213 on Build GRCh38
rs1015286564 Chr.7: 116561197-116561197 on Build GRCh38
rs534236443 Chr.7: 116560989-116560989 on Build GRCh38
rs556565826 Chr.7: 116561131-116561131 on Build GRCh38
rs926742875 Chr.7: 116561218-116561218 on Build GRCh38
rs931717546 Chr.7: 116561029-116561029 on Build GRCh38
rs538550256 Chr.7: 116561124-116561124 on Build GRCh38
rs1051922020 Chr.7: 116561103-116561103 on Build GRCh38
rs892705378 Chr.7: 116561075-116561075 on Build GRCh38
rs934451100 Chr.7: 116560924-116560924 on Build GRCh38
rs922924798 Chr.7: 116561245-116561245 on Build GRCh38
rs992668412 Chr.7: 116560977-116560977 on Build GRCh38
rs1053129173 Chr.7: 116561052-116561052 on Build GRCh38
rs890194110 Chr.7: 116561106-116561106 on Build GRCh38
rs35103261 Chr.7: 116561258-116561258 on Build GRCh38
rs961972349 Chr.7: 116561207-116561207 on Build GRCh38
rs6868 Chr.7: 116560970-116560970 on Build GRCh38
rs574733654 Chr.7: 116561150-116561150 on Build GRCh38
rs577127115 Chr.7: 116561093-116561093 on Build GRCh38
rs553664696 Chr.7: 116560869-116560869 on Build GRCh38
rs924569077 Chr.7: 116560912-116560912 on Build GRCh38
rs778726620 Chr.7: 116561005-116561005 on Build GRCh38
rs542937271 Chr.7: 116560937-116560937 on Build GRCh38
rs925001891 Chr.7: 116561287-116561287 on Build GRCh38
rs969476489 Chr.7: 116561117-116561117 on Build GRCh38
rs917967683 Chr.7: 116560994-116560994 on Build GRCh38
rs919509091 Chr.7: 116561280-116561280 on Build GRCh38
rs34585220 Chr.7: 116560882-116560882 on Build GRCh38
rs374969893 Chr.7: 116560999-116560999 on Build GRCh38
rs985881114 Chr.7: 116561273-116561273 on Build GRCh38
rs917116576 Chr.7: 116561177-116561177 on Build GRCh38
rs35617054 Chr.7: 116560847-116560847 on Build GRCh38
rs560268745 Chr.7: 116561173-116561173 on Build GRCh38
rs1037713153 Chr.7: 116561262-116561262 on Build GRCh38
rs929592723 Chr.7: 116561284-116561284 on Build GRCh38
rs993589427 Chr.7: 116561079-116561079 on Build GRCh38
rs1007291671 Chr.7: 116561115-116561116 on Build GRCh38
rs577439703 Chr.7: 116561276-116561285 on Build GRCh38
rs907762826 Chr.7: 116560890-116560890 on Build GRCh38
rs951898689 Chr.7: 116561172-116561172 on Build GRCh38
rs1014027835 Chr.7: 116561099-116561099 on Build GRCh38
rs1045060352 Chr.7: 116561038-116561038 on Build GRCh38
rs979109070 Chr.7: 116560894-116560894 on Build GRCh38
rs985754778 Chr.7: 116560875-116560875 on Build GRCh38
rs1000996401 Chr.7: 116561043-116561043 on Build GRCh38

View Full Product Details