Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116892521-116893061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA2
Entrez Gene ID
830
Gene Name
capping actin protein of muscle Z-line alpha subunit 2
Gene Aliases
CAPPA2, CAPZ
UniGene
Hs.446123
Gene Chromosome Location
Chr.7: 116862478-116919261 on Build GRCh38
Transcripts
NM_006136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5414773 c.142G>T p.E48* Chr.7: 116893032-116893032 on Build GRCh38
3257001 c.109C>T p.R37W Chr.7: 116892999-116892999 on Build GRCh38
484663 c.150A>T p.A50A Chr.7: 116893040-116893040 on Build GRCh38
7186649 c.104-2A>G p.? Chr.7: 116892992-116892992 on Build GRCh38
4979840 c.147A>T p.G49G Chr.7: 116893037-116893037 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs186440275 Chr.7: 116892800-116892800 on Build GRCh38
rs797013962 Chr.7: 116892643-116892643 on Build GRCh38
rs372003873 Chr.7: 116892933-116892933 on Build GRCh38
rs953821359 Chr.7: 116892661-116892661 on Build GRCh38
rs377062970 Chr.7: 116892953-116892953 on Build GRCh38
rs748979003 Chr.7: 116892970-116892970 on Build GRCh38
rs540436981 Chr.7: 116892943-116892943 on Build GRCh38
rs1032112589 Chr.7: 116892939-116892939 on Build GRCh38
rs374140633 Chr.7: 116892844-116892844 on Build GRCh38
rs963627196 Chr.7: 116892857-116892857 on Build GRCh38
rs917365357 Chr.7: 116892731-116892731 on Build GRCh38
rs745645847 Chr.7: 116892679-116892679 on Build GRCh38
rs535159777 Chr.7: 116892621-116892621 on Build GRCh38
rs755590842 Chr.7: 116892964-116892964 on Build GRCh38
rs370917046 Chr.7: 116892954-116892954 on Build GRCh38
rs978377566 Chr.7: 116892678-116892678 on Build GRCh38
rs891403783 Chr.7: 116892848-116892851 on Build GRCh38
rs753096627 Chr.7: 116892956-116892956 on Build GRCh38
rs926376313 Chr.7: 116892695-116892695 on Build GRCh38
rs1000634199 Chr.7: 116892627-116892628 on Build GRCh38
rs778897327 Chr.7: 116892978-116892978 on Build GRCh38
rs934904011 Chr.7: 116892684-116892684 on Build GRCh38
rs976875770 Chr.7: 116892567-116892567 on Build GRCh38
rs988036115 Chr.7: 116892660-116892661 on Build GRCh38
rs751850071 Chr.7: 116892981-116892981 on Build GRCh38
rs888405125 Chr.7: 116892845-116892845 on Build GRCh38
rs113053668 Chr.7: 116892780-116892780 on Build GRCh38
rs758994476 Chr.7: 116892959-116892959 on Build GRCh38
rs1024890030 Chr.7: 116892975-116892975 on Build GRCh38
rs766907453 Chr.7: 116892961-116892961 on Build GRCh38
rs775118336 Chr.7: 116892819-116892820 on Build GRCh38
rs950171596 Chr.7: 116892777-116892777 on Build GRCh38
rs953806669 Chr.7: 116892990-116892990 on Build GRCh38
rs531979858 Chr.7: 116892655-116892656 on Build GRCh38
rs199607353 Chr.7: 116892962-116892962 on Build GRCh38
rs74273923 Chr.7: 116892831-116892833 on Build GRCh38
rs994869163 Chr.7: 116892833-116892833 on Build GRCh38
rs1053253576 Chr.7: 116892694-116892694 on Build GRCh38
rs397794781 Chr.7: 116892833-116892834 on Build GRCh38
rs1032162281 Chr.7: 116892629-116892629 on Build GRCh38
rs748001357 Chr.7: 116892981-116892981 on Build GRCh38
rs944048989 Chr.7: 116892850-116892850 on Build GRCh38
rs778624541 Chr.7: 116892628-116892628 on Build GRCh38
rs1000918657 Chr.7: 116892931-116892931 on Build GRCh38
rs920953940 Chr.7: 116892595-116892595 on Build GRCh38
rs754766168 Chr.7: 116892974-116892974 on Build GRCh38
rs59213951 Chr.7: 116892820-116892821 on Build GRCh38
rs925617155 Chr.7: 116893028-116893028 on Build GRCh38
rs899899680 Chr.7: 116892899-116892899 on Build GRCh38
rs566584513 Chr.7: 116892622-116892623 on Build GRCh38
rs772112270 Chr.7: 116892985-116892985 on Build GRCh38
rs941805463 Chr.7: 116892723-116892723 on Build GRCh38
rs1008696047 Chr.7: 116892596-116892596 on Build GRCh38
rs747056452 Chr.7: 116893021-116893021 on Build GRCh38
rs779785865 Chr.7: 116892969-116892969 on Build GRCh38
rs1040130780 Chr.7: 116892895-116892895 on Build GRCh38
rs386411094 Chr.7: 116892832-116892833 on Build GRCh38
rs905880770 Chr.7: 116892801-116892801 on Build GRCh38
rs773154833 Chr.7: 116893005-116893005 on Build GRCh38
rs920112228 Chr.7: 116892786-116892786 on Build GRCh38
rs376828966 Chr.7: 116892965-116892965 on Build GRCh38

View Full Product Details