Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116903940-116904442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA2
Entrez Gene ID
830
Gene Name
capping actin protein of muscle Z-line alpha subunit 2
Gene Aliases
CAPPA2, CAPZ
UniGene
Hs.446123
Gene Chromosome Location
Chr.7: 116862478-116919261 on Build GRCh38
Transcripts
NM_006136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4669218 c.400G>T p.E134* Chr.7: 116904357-116904357 on Build GRCh38
4669217 c.240C>T p.G80G Chr.7: 116904197-116904197 on Build GRCh38
5345245 c.333T>A p.C111* Chr.7: 116904290-116904290 on Build GRCh38
4649770 c.306G>A p.R102R Chr.7: 116904263-116904263 on Build GRCh38
1084418 c.362G>T p.R121I Chr.7: 116904319-116904319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966800661 Chr.7: 116904038-116904038 on Build GRCh38
rs377102966 Chr.7: 116904350-116904350 on Build GRCh38
rs781247938 Chr.7: 116904202-116904202 on Build GRCh38
rs944427862 Chr.7: 116904307-116904307 on Build GRCh38
rs915831392 Chr.7: 116904197-116904197 on Build GRCh38
rs773866358 Chr.7: 116904308-116904308 on Build GRCh38
rs368538215 Chr.7: 116904286-116904286 on Build GRCh38
rs368360127 Chr.7: 116904362-116904362 on Build GRCh38
rs764300633 Chr.7: 116904330-116904330 on Build GRCh38
rs374152545 Chr.7: 116904227-116904227 on Build GRCh38
rs929121556 Chr.7: 116904268-116904268 on Build GRCh38
rs17849974 Chr.7: 116904287-116904287 on Build GRCh38
rs777452136 Chr.7: 116904144-116904144 on Build GRCh38
rs890194325 Chr.7: 116904306-116904306 on Build GRCh38
rs1044570741 Chr.7: 116904156-116904156 on Build GRCh38
rs759052499 Chr.7: 116904320-116904320 on Build GRCh38
rs535780548 Chr.7: 116903977-116903977 on Build GRCh38
rs60417621 Chr.7: 116904047-116904047 on Build GRCh38
rs780042673 Chr.7: 116904417-116904417 on Build GRCh38
rs186849705 Chr.7: 116904410-116904410 on Build GRCh38
rs565312791 Chr.7: 116904103-116904103 on Build GRCh38
rs1056836260 Chr.7: 116904021-116904021 on Build GRCh38
rs199972501 Chr.7: 116904338-116904338 on Build GRCh38
rs780280075 Chr.7: 116904263-116904263 on Build GRCh38
rs754263459 Chr.7: 116904367-116904367 on Build GRCh38
rs1016456262 Chr.7: 116904235-116904235 on Build GRCh38
rs370326853 Chr.7: 116904198-116904198 on Build GRCh38
rs756167030 Chr.7: 116904416-116904416 on Build GRCh38
rs895528114 Chr.7: 116904055-116904055 on Build GRCh38
rs902707523 Chr.7: 116904408-116904408 on Build GRCh38
rs1022490148 Chr.7: 116904377-116904377 on Build GRCh38
rs999682965 Chr.7: 116904412-116904412 on Build GRCh38
rs960727781 Chr.7: 116904078-116904078 on Build GRCh38
rs565970603 Chr.7: 116904345-116904345 on Build GRCh38
rs1035085406 Chr.7: 116904057-116904057 on Build GRCh38
rs934284562 Chr.7: 116904018-116904018 on Build GRCh38
rs973273340 Chr.7: 116904236-116904236 on Build GRCh38
rs34058809 Chr.7: 116904211-116904212 on Build GRCh38
rs751479961 Chr.7: 116904167-116904167 on Build GRCh38
rs772761567 Chr.7: 116904304-116904304 on Build GRCh38
rs754905566 Chr.7: 116904405-116904406 on Build GRCh38
rs753690773 Chr.7: 116904145-116904146 on Build GRCh38
rs769038997 Chr.7: 116904270-116904270 on Build GRCh38
rs762566085 Chr.7: 116904346-116904346 on Build GRCh38
rs144500777 Chr.7: 116904239-116904239 on Build GRCh38
rs749490468 Chr.7: 116904265-116904265 on Build GRCh38
rs372359690 Chr.7: 116904391-116904391 on Build GRCh38

View Full Product Details