Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117368486-117368988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
UniGene
Hs.352412
Gene Chromosome Location
Chr.7: 117363222-117427523 on Build GRCh38
Transcripts
NM_001301821.1 NM_001301822.1 NM_130768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1699237 c.1078C>T p.L360F Chr.7: 117368695-117368695 on Build GRCh38
3770009 c.1096C>T p.Q366* Chr.7: 117368677-117368677 on Build GRCh38
5693803 c.1109T>A p.L370* Chr.7: 117368664-117368664 on Build GRCh38
3831705 c.1132A>T p.I378F Chr.7: 117368641-117368641 on Build GRCh38
5005918 c.1117G>A p.A373T Chr.7: 117368656-117368656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954860569 Chr.7: 117368930-117368930 on Build GRCh38
rs199687267 Chr.7: 117368657-117368657 on Build GRCh38
rs757099237 Chr.7: 117368686-117368686 on Build GRCh38
rs546187042 Chr.7: 117368965-117368965 on Build GRCh38
rs776522125 Chr.7: 117368617-117368617 on Build GRCh38
rs758049899 Chr.7: 117368675-117368675 on Build GRCh38
rs772838344 Chr.7: 117368581-117368584 on Build GRCh38
rs753556409 Chr.7: 117368729-117368732 on Build GRCh38
rs746852807 Chr.7: 117368577-117368577 on Build GRCh38
rs553172954 Chr.7: 117368851-117368851 on Build GRCh38
rs774785077 Chr.7: 117368635-117368635 on Build GRCh38
rs763399527 Chr.7: 117368629-117368629 on Build GRCh38
rs567659937 Chr.7: 117368807-117368807 on Build GRCh38
rs185185735 Chr.7: 117368940-117368940 on Build GRCh38
rs551922470 Chr.7: 117368595-117368595 on Build GRCh38
rs537698945 Chr.7: 117368717-117368717 on Build GRCh38
rs367991079 Chr.7: 117368753-117368753 on Build GRCh38
rs746841209 Chr.7: 117368682-117368682 on Build GRCh38
rs773826471 Chr.7: 117368748-117368748 on Build GRCh38
rs917932606 Chr.7: 117368785-117368785 on Build GRCh38
rs948048665 Chr.7: 117368792-117368792 on Build GRCh38
rs1045562703 Chr.7: 117368835-117368835 on Build GRCh38
rs28513898 Chr.7: 117368655-117368655 on Build GRCh38
rs868027480 Chr.7: 117368704-117368704 on Build GRCh38
rs762270460 Chr.7: 117368743-117368743 on Build GRCh38
rs753247880 Chr.7: 117368663-117368663 on Build GRCh38
rs764623945 Chr.7: 117368630-117368630 on Build GRCh38
rs762540927 Chr.7: 117368766-117368766 on Build GRCh38
rs774531943 Chr.7: 117368730-117368730 on Build GRCh38
rs770060817 Chr.7: 117368710-117368710 on Build GRCh38
rs772428013 Chr.7: 117368746-117368746 on Build GRCh38
rs977278152 Chr.7: 117368632-117368632 on Build GRCh38
rs770105665 Chr.7: 117368626-117368626 on Build GRCh38
rs765950124 Chr.7: 117368723-117368727 on Build GRCh38
rs142005201 Chr.7: 117368625-117368625 on Build GRCh38
rs749458053 Chr.7: 117368720-117368720 on Build GRCh38
rs975581971 Chr.7: 117368524-117368524 on Build GRCh38
rs759041210 Chr.7: 117368749-117368749 on Build GRCh38
rs769015711 Chr.7: 117368726-117368726 on Build GRCh38
rs764856529 Chr.7: 117368670-117368670 on Build GRCh38
rs762171269 Chr.7: 117368648-117368648 on Build GRCh38
rs367695735 Chr.7: 117368759-117368759 on Build GRCh38
rs866514268 Chr.7: 117368952-117368952 on Build GRCh38
rs758246476 Chr.7: 117368569-117368569 on Build GRCh38
rs535158085 Chr.7: 117368847-117368847 on Build GRCh38
rs745916105 Chr.7: 117368696-117368696 on Build GRCh38
rs777682587 Chr.7: 117368574-117368574 on Build GRCh38
rs529769327 Chr.7: 117368604-117368604 on Build GRCh38
rs577799666 Chr.7: 117368904-117368904 on Build GRCh38
rs781226925 Chr.7: 117368688-117368688 on Build GRCh38
rs760477855 Chr.7: 117368709-117368711 on Build GRCh38
rs931509017 Chr.7: 117368869-117368869 on Build GRCh38
rs867756032 Chr.7: 117368668-117368668 on Build GRCh38
rs200577870 Chr.7: 117368681-117368681 on Build GRCh38
rs374127734 Chr.7: 117368667-117368667 on Build GRCh38
rs181146096 Chr.7: 117368905-117368905 on Build GRCh38
rs1047250922 Chr.7: 117368900-117368900 on Build GRCh38
rs1015249134 Chr.7: 117368881-117368881 on Build GRCh38

View Full Product Details