Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117380886-117381412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
UniGene
Hs.352412
Gene Chromosome Location
Chr.7: 117363222-117427523 on Build GRCh38
Transcripts
NM_001301821.1 NM_001301822.1 NM_130768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
380665 c.945G>C p.K315N Chr.7: 117381011-117381011 on Build GRCh38
3922550 c.924G>A p.M308I Chr.7: 117381032-117381032 on Build GRCh38
5886508 c.889G>A p.E297K Chr.7: 117381067-117381067 on Build GRCh38
73756 c.893G>C p.R298T Chr.7: 117381063-117381063 on Build GRCh38
452194 c.913C>G p.L305V Chr.7: 117381043-117381043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943138436 Chr.7: 117381283-117381283 on Build GRCh38
rs915064279 Chr.7: 117381357-117381357 on Build GRCh38
rs1002961038 Chr.7: 117380918-117380918 on Build GRCh38
rs957973369 Chr.7: 117380940-117380940 on Build GRCh38
rs760587484 Chr.7: 117381061-117381061 on Build GRCh38
rs77394880 Chr.7: 117381361-117381361 on Build GRCh38
rs909358082 Chr.7: 117381000-117381000 on Build GRCh38
rs1048600101 Chr.7: 117381382-117381382 on Build GRCh38
rs963537413 Chr.7: 117381203-117381203 on Build GRCh38
rs927827859 Chr.7: 117381276-117381276 on Build GRCh38
rs954404060 Chr.7: 117381238-117381238 on Build GRCh38
rs780365468 Chr.7: 117381139-117381139 on Build GRCh38
rs570934754 Chr.7: 117381162-117381162 on Build GRCh38
rs371414120 Chr.7: 117380978-117380978 on Build GRCh38
rs1042268742 Chr.7: 117381120-117381120 on Build GRCh38
rs188157077 Chr.7: 117381327-117381327 on Build GRCh38
rs920237362 Chr.7: 117381297-117381297 on Build GRCh38
rs1035775072 Chr.7: 117380921-117380921 on Build GRCh38
rs772101314 Chr.7: 117381133-117381133 on Build GRCh38
rs186067529 Chr.7: 117381175-117381175 on Build GRCh38
rs751810129 Chr.7: 117381118-117381120 on Build GRCh38
rs558804527 Chr.7: 117381243-117381243 on Build GRCh38
rs777168123 Chr.7: 117380988-117380988 on Build GRCh38
rs937783251 Chr.7: 117381071-117381071 on Build GRCh38
rs966644931 Chr.7: 117381273-117381273 on Build GRCh38
rs758791076 Chr.7: 117381080-117381081 on Build GRCh38
rs765302267 Chr.7: 117381082-117381082 on Build GRCh38
rs113269630 Chr.7: 117381115-117381117 on Build GRCh38
rs537863791 Chr.7: 117381149-117381149 on Build GRCh38
rs570489423 Chr.7: 117381142-117381142 on Build GRCh38
rs555195143 Chr.7: 117381194-117381194 on Build GRCh38
rs757672074 Chr.7: 117381098-117381098 on Build GRCh38
rs989212538 Chr.7: 117381340-117381340 on Build GRCh38
rs771393711 Chr.7: 117380986-117380986 on Build GRCh38
rs544206783 Chr.7: 117381274-117381274 on Build GRCh38
rs768282961 Chr.7: 117381045-117381045 on Build GRCh38
rs142166886 Chr.7: 117381052-117381052 on Build GRCh38
rs569910762 Chr.7: 117381293-117381293 on Build GRCh38
rs142082886 Chr.7: 117381053-117381053 on Build GRCh38
rs535157480 Chr.7: 117381188-117381188 on Build GRCh38
rs753215409 Chr.7: 117381079-117381080 on Build GRCh38
rs112516673 Chr.7: 117381092-117381092 on Build GRCh38
rs539593129 Chr.7: 117381166-117381166 on Build GRCh38
rs767169338 Chr.7: 117381067-117381067 on Build GRCh38
rs769755144 Chr.7: 117381259-117381259 on Build GRCh38
rs747403334 Chr.7: 117380985-117380985 on Build GRCh38
rs893613425 Chr.7: 117381107-117381107 on Build GRCh38
rs17139765 Chr.7: 117380927-117380927 on Build GRCh38
rs753884212 Chr.7: 117381062-117381062 on Build GRCh38
rs374642066 Chr.7: 117381054-117381054 on Build GRCh38
rs182897927 Chr.7: 117381265-117381265 on Build GRCh38
rs531920315 Chr.7: 117381177-117381177 on Build GRCh38
rs778060086 Chr.7: 117380960-117380960 on Build GRCh38
rs987215048 Chr.7: 117381077-117381077 on Build GRCh38
rs190383508 Chr.7: 117381225-117381225 on Build GRCh38
rs748730818 Chr.7: 117381027-117381027 on Build GRCh38
rs987076337 Chr.7: 117381241-117381241 on Build GRCh38
rs990984953 Chr.7: 117381290-117381290 on Build GRCh38
rs201641418 Chr.7: 117381069-117381069 on Build GRCh38
rs989404874 Chr.7: 117381004-117381004 on Build GRCh38
rs943231734 Chr.7: 117381089-117381089 on Build GRCh38
rs932983755 Chr.7: 117381064-117381064 on Build GRCh38
rs754494220 Chr.7: 117381148-117381148 on Build GRCh38
rs958278927 Chr.7: 117381251-117381251 on Build GRCh38
rs758923931 Chr.7: 117381068-117381068 on Build GRCh38
rs528148133 Chr.7: 117381187-117381187 on Build GRCh38
rs758604779 Chr.7: 117381087-117381087 on Build GRCh38
rs78018733 Chr.7: 117381206-117381206 on Build GRCh38
rs747350199 Chr.7: 117381097-117381097 on Build GRCh38
rs746968339 Chr.7: 117380930-117380930 on Build GRCh38
rs773103344 Chr.7: 117381058-117381058 on Build GRCh38
rs770220973 Chr.7: 117381119-117381119 on Build GRCh38
rs201128524 Chr.7: 117381113-117381113 on Build GRCh38
rs370147977 Chr.7: 117381086-117381086 on Build GRCh38
rs373768852 Chr.7: 117381189-117381189 on Build GRCh38
rs754875854 Chr.7: 117381070-117381070 on Build GRCh38
rs746322426 Chr.7: 117381118-117381118 on Build GRCh38
rs866777738 Chr.7: 117381226-117381226 on Build GRCh38

View Full Product Details