Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117539709-117540219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFTR
Entrez Gene ID
1080
Gene Name
cystic fibrosis transmembrane conductance regulator
Gene Aliases
ABC35, ABCC7, CF, CFTR/MRP, MRP7, TNR-CFTR, dJ760C5.1
UniGene
Hs.489786
Gene Chromosome Location
Chr.7: 117478367-117668665 on Build GRCh38
Transcripts
NM_000492.3 XM_011515751.2 XM_011515753.1 XM_011515754.2 XM_017011699.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5871654 c.970C>T p.P324S Chr.7: 117540200-117540200 on Build GRCh38
6627588 c.870-10delT p.? Chr.7: 117540090-117540090 on Build GRCh38
4882948 c.927C>G p.A309A Chr.7: 117540157-117540157 on Build GRCh38
1084465 c.874G>T p.E292* Chr.7: 117540104-117540104 on Build GRCh38
7196289 c.960A>G p.L320L Chr.7: 117540190-117540190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575984390 Chr.7: 117540021-117540021 on Build GRCh38
rs972944408 Chr.7: 117539746-117539746 on Build GRCh38
rs760683078 Chr.7: 117540078-117540078 on Build GRCh38
rs397508817 Chr.7: 117540150-117540150 on Build GRCh38
rs201885470 Chr.7: 117540143-117540143 on Build GRCh38
rs765466169 Chr.7: 117540115-117540116 on Build GRCh38
rs536748954 Chr.7: 117540006-117540006 on Build GRCh38
rs750145040 Chr.7: 117540066-117540066 on Build GRCh38
rs397508814 Chr.7: 117540119-117540119 on Build GRCh38
rs532798115 Chr.7: 117539830-117539830 on Build GRCh38
rs121909016 Chr.7: 117540163-117540163 on Build GRCh38
rs961403023 Chr.7: 117539738-117539738 on Build GRCh38
rs142134579 Chr.7: 117540129-117540129 on Build GRCh38
rs746375310 Chr.7: 117540147-117540147 on Build GRCh38
rs555127124 Chr.7: 117540020-117540020 on Build GRCh38
rs1800083 Chr.7: 117540145-117540145 on Build GRCh38
rs150691494 Chr.7: 117540132-117540132 on Build GRCh38
rs397508818 Chr.7: 117540156-117540156 on Build GRCh38
rs955701492 Chr.7: 117539811-117539811 on Build GRCh38
rs911405811 Chr.7: 117539870-117539870 on Build GRCh38
rs121908768 Chr.7: 117540163-117540165 on Build GRCh38
rs199926595 Chr.7: 117540127-117540127 on Build GRCh38
rs779120165 Chr.7: 117540102-117540102 on Build GRCh38
rs907131618 Chr.7: 117539967-117539967 on Build GRCh38
rs1057516387 Chr.7: 117540119-117540120 on Build GRCh38
rs368890501 Chr.7: 117540087-117540087 on Build GRCh38
rs397508813 Chr.7: 117540111-117540112 on Build GRCh38
rs760319837 Chr.7: 117540174-117540174 on Build GRCh38
rs977698733 Chr.7: 117539760-117539760 on Build GRCh38
rs397508812 Chr.7: 117540107-117540107 on Build GRCh38
rs148013312 Chr.7: 117540155-117540155 on Build GRCh38
rs565615211 Chr.7: 117539777-117539777 on Build GRCh38
rs928854256 Chr.7: 117539864-117539864 on Build GRCh38
rs1800085 Chr.7: 117540194-117540194 on Build GRCh38
rs935752254 Chr.7: 117539876-117539876 on Build GRCh38
rs75528968 Chr.7: 117540176-117540177 on Build GRCh38
rs759762840 Chr.7: 117540090-117540092 on Build GRCh38
rs765443528 Chr.7: 117540191-117540191 on Build GRCh38
rs397508810 Chr.7: 117540097-117540097 on Build GRCh38
rs1001886734 Chr.7: 117540016-117540016 on Build GRCh38
rs121908744 Chr.7: 117540178-117540178 on Build GRCh38
rs766189605 Chr.7: 117540184-117540184 on Build GRCh38
rs755173693 Chr.7: 117540096-117540096 on Build GRCh38
rs397508816 Chr.7: 117540142-117540142 on Build GRCh38
rs144476686 Chr.7: 117540188-117540188 on Build GRCh38
rs78742051 Chr.7: 117540178-117540178 on Build GRCh38
rs779201407 Chr.7: 117540177-117540177 on Build GRCh38
rs1048951787 Chr.7: 117540004-117540004 on Build GRCh38
rs397508811 Chr.7: 117540104-117540104 on Build GRCh38
rs75763344 Chr.7: 117540171-117540171 on Build GRCh38
rs62469438 Chr.7: 117539922-117539922 on Build GRCh38
rs397508819 Chr.7: 117540170-117540170 on Build GRCh38
rs752715236 Chr.7: 117540117-117540117 on Build GRCh38
rs587777924 Chr.7: 117540165-117540167 on Build GRCh38
rs200046355 Chr.7: 117540148-117540148 on Build GRCh38
rs922879735 Chr.7: 117539778-117539778 on Build GRCh38
rs397508820 Chr.7: 117540189-117540189 on Build GRCh38
rs752781052 Chr.7: 117540193-117540193 on Build GRCh38
rs766169135 Chr.7: 117540079-117540079 on Build GRCh38
rs377686676 Chr.7: 117540087-117540094 on Build GRCh38
rs56093012 Chr.7: 117540190-117540190 on Build GRCh38
rs550951300 Chr.7: 117539877-117539877 on Build GRCh38
rs1800084 Chr.7: 117540157-117540157 on Build GRCh38
rs769325089 Chr.7: 117539745-117539748 on Build GRCh38
rs146652541 Chr.7: 117540130-117540130 on Build GRCh38
rs887147551 Chr.7: 117540089-117540090 on Build GRCh38
rs143486492 Chr.7: 117540120-117540120 on Build GRCh38
rs115770955 Chr.7: 117539932-117539932 on Build GRCh38
rs201465561 Chr.7: 117540118-117540118 on Build GRCh38
rs1046037350 Chr.7: 117539954-117539954 on Build GRCh38
rs202168465 Chr.7: 117540169-117540169 on Build GRCh38

View Full Product Details