Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 120989300-120989796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPED1
Entrez Gene ID
79974
Gene Name
cadherin like and PC-esterase domain containing 1
Gene Aliases
C7orf58
UniGene
Hs.189652
Gene Chromosome Location
Chr.7: 120988697-121297444 on Build GRCh38
Transcripts
NM_001105533.1 NM_024913.4 XM_017012649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
266148 c.34C>T p.R12* Chr.7: 120989655-120989655 on Build GRCh38
5173912 c.41G>A p.C14Y Chr.7: 120989662-120989662 on Build GRCh38
5765881 c.165A>G p.E55E Chr.7: 120989786-120989786 on Build GRCh38
5628974 c.140G>A p.G47E Chr.7: 120989761-120989761 on Build GRCh38
3995307 c.102C>G p.T34T Chr.7: 120989723-120989723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867091470 Chr.7: 120989740-120989740 on Build GRCh38
rs921530473 Chr.7: 120989467-120989467 on Build GRCh38
rs970341929 Chr.7: 120989510-120989510 on Build GRCh38
rs1004684670 Chr.7: 120989723-120989723 on Build GRCh38
rs777840717 Chr.7: 120989605-120989605 on Build GRCh38
rs746198196 Chr.7: 120989716-120989716 on Build GRCh38
rs374427830 Chr.7: 120989644-120989644 on Build GRCh38
rs990596912 Chr.7: 120989491-120989491 on Build GRCh38
rs2525740 Chr.7: 120989427-120989427 on Build GRCh38
rs200094818 Chr.7: 120989743-120989743 on Build GRCh38
rs752887386 Chr.7: 120989678-120989678 on Build GRCh38
rs376695432 Chr.7: 120989763-120989763 on Build GRCh38
rs551515211 Chr.7: 120989518-120989518 on Build GRCh38
rs767046266 Chr.7: 120989577-120989577 on Build GRCh38
rs772408709 Chr.7: 120989601-120989601 on Build GRCh38
rs772674831 Chr.7: 120989655-120989655 on Build GRCh38
rs983950532 Chr.7: 120989587-120989587 on Build GRCh38
rs760551767 Chr.7: 120989656-120989656 on Build GRCh38
rs779185603 Chr.7: 120989595-120989595 on Build GRCh38
rs902770295 Chr.7: 120989429-120989429 on Build GRCh38
rs759985372 Chr.7: 120989640-120989640 on Build GRCh38
rs140850174 Chr.7: 120989726-120989726 on Build GRCh38
rs747156936 Chr.7: 120989688-120989688 on Build GRCh38
rs144976107 Chr.7: 120989741-120989741 on Build GRCh38
rs1034300026 Chr.7: 120989349-120989349 on Build GRCh38
rs1055698407 Chr.7: 120989637-120989637 on Build GRCh38
rs760297310 Chr.7: 120989736-120989736 on Build GRCh38
rs962931614 Chr.7: 120989453-120989453 on Build GRCh38
rs778056931 Chr.7: 120989684-120989684 on Build GRCh38
rs377692691 Chr.7: 120989687-120989687 on Build GRCh38
rs931240963 Chr.7: 120989576-120989576 on Build GRCh38
rs189461125 Chr.7: 120989382-120989382 on Build GRCh38
rs756753784 Chr.7: 120989610-120989610 on Build GRCh38
rs755134687 Chr.7: 120989590-120989590 on Build GRCh38
rs763708606 Chr.7: 120989463-120989463 on Build GRCh38
rs1024303974 Chr.7: 120989667-120989667 on Build GRCh38
rs776638026 Chr.7: 120989750-120989750 on Build GRCh38
rs748212252 Chr.7: 120989600-120989600 on Build GRCh38
rs778630027 Chr.7: 120989622-120989623 on Build GRCh38
rs997718765 Chr.7: 120989423-120989423 on Build GRCh38
rs567416903 Chr.7: 120989665-120989665 on Build GRCh38
rs945786673 Chr.7: 120989502-120989502 on Build GRCh38
rs144547146 Chr.7: 120989442-120989442 on Build GRCh38
rs755008351 Chr.7: 120989671-120989671 on Build GRCh38
rs768507297 Chr.7: 120989588-120989593 on Build GRCh38
rs565305465 Chr.7: 120989589-120989589 on Build GRCh38
rs368713267 Chr.7: 120989498-120989498 on Build GRCh38
rs753666660 Chr.7: 120989668-120989668 on Build GRCh38
rs781584083 Chr.7: 120989690-120989690 on Build GRCh38
rs902741757 Chr.7: 120989711-120989711 on Build GRCh38
rs560130076 Chr.7: 120989474-120989474 on Build GRCh38
rs758347698 Chr.7: 120989683-120989683 on Build GRCh38
rs770351504 Chr.7: 120989718-120989718 on Build GRCh38
rs765274413 Chr.7: 120989768-120989768 on Build GRCh38
rs149960034 Chr.7: 120989734-120989734 on Build GRCh38
rs918481425 Chr.7: 120989525-120989525 on Build GRCh38
rs1017517223 Chr.7: 120989449-120989449 on Build GRCh38
rs770261755 Chr.7: 120989643-120989643 on Build GRCh38
rs768567827 Chr.7: 120989341-120989341 on Build GRCh38
rs200553334 Chr.7: 120989608-120989608 on Build GRCh38
rs374473383 Chr.7: 120989634-120989634 on Build GRCh38
rs767213299 Chr.7: 120989653-120989653 on Build GRCh38
rs146633346 Chr.7: 120989456-120989456 on Build GRCh38
rs202149555 Chr.7: 120989585-120989585 on Build GRCh38
rs753364642 Chr.7: 120989473-120989473 on Build GRCh38
rs570134352 Chr.7: 120989375-120989375 on Build GRCh38
rs139400449 Chr.7: 120989730-120989730 on Build GRCh38
rs901889040 Chr.7: 120989417-120989417 on Build GRCh38
rs377528126 Chr.7: 120989370-120989370 on Build GRCh38
rs777151635 Chr.7: 120989625-120989625 on Build GRCh38
rs199634821 Chr.7: 120989648-120989648 on Build GRCh38
rs943482082 Chr.7: 120989338-120989338 on Build GRCh38
rs1042797422 Chr.7: 120989697-120989697 on Build GRCh38
rs747941752 Chr.7: 120989668-120989668 on Build GRCh38
rs1024355031 Chr.7: 120989413-120989413 on Build GRCh38
rs375547307 Chr.7: 120989752-120989752 on Build GRCh38
rs771443560 Chr.7: 120989731-120989731 on Build GRCh38
rs895350305 Chr.7: 120989739-120989739 on Build GRCh38

View Full Product Details