Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121125820-121126369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPED1
Entrez Gene ID
79974
Gene Name
cadherin like and PC-esterase domain containing 1
Gene Aliases
C7orf58
UniGene
Hs.189652
Gene Chromosome Location
Chr.7: 120988697-121297444 on Build GRCh38
Transcripts
NM_001105533.1 NM_024913.4 XM_017012649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6390703 c.1065C>A p.C355* Chr.7: 121125823-121125823 on Build GRCh38
6390702 c.1065C>A p.C355* Chr.7: 121125823-121125823 on Build GRCh38
6108401 c.1126G>T p.V376L Chr.7: 121125884-121125884 on Build GRCh38
1084558 c.1090T>G p.F364V Chr.7: 121125848-121125848 on Build GRCh38
7135300 c.1128G>T p.V376V Chr.7: 121125886-121125886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113289869 Chr.7: 121126163-121126163 on Build GRCh38
rs1009297092 Chr.7: 121126264-121126264 on Build GRCh38
rs755212160 Chr.7: 121125890-121125890 on Build GRCh38
rs761866836 Chr.7: 121126221-121126221 on Build GRCh38
rs753004596 Chr.7: 121125895-121125895 on Build GRCh38
rs147828421 Chr.7: 121125867-121125868 on Build GRCh38
rs12673770 Chr.7: 121126268-121126268 on Build GRCh38
rs557976074 Chr.7: 121125866-121125866 on Build GRCh38
rs960675823 Chr.7: 121126326-121126326 on Build GRCh38
rs772474053 Chr.7: 121125914-121125914 on Build GRCh38
rs1039660277 Chr.7: 121126181-121126181 on Build GRCh38
rs577164886 Chr.7: 121125919-121125919 on Build GRCh38
rs750348141 Chr.7: 121125873-121125873 on Build GRCh38
rs750513301 Chr.7: 121125920-121125922 on Build GRCh38
rs933302387 Chr.7: 121126240-121126240 on Build GRCh38
rs561388220 Chr.7: 121126108-121126108 on Build GRCh38
rs936870701 Chr.7: 121126263-121126263 on Build GRCh38
rs933588019 Chr.7: 121125844-121125844 on Build GRCh38
rs1025520790 Chr.7: 121125982-121125982 on Build GRCh38
rs188718968 Chr.7: 121126313-121126313 on Build GRCh38
rs992541512 Chr.7: 121126328-121126328 on Build GRCh38
rs369529110 Chr.7: 121125910-121125910 on Build GRCh38
rs798949 Chr.7: 121125900-121125900 on Build GRCh38
rs772039513 Chr.7: 121125913-121125913 on Build GRCh38
rs967891215 Chr.7: 121126301-121126302 on Build GRCh38
rs766604756 Chr.7: 121125878-121125878 on Build GRCh38
rs902688535 Chr.7: 121125885-121125885 on Build GRCh38
rs556529793 Chr.7: 121126060-121126060 on Build GRCh38
rs767568660 Chr.7: 121125872-121125872 on Build GRCh38
rs776760561 Chr.7: 121126220-121126220 on Build GRCh38
rs1024613051 Chr.7: 121125918-121125918 on Build GRCh38
rs202194529 Chr.7: 121125877-121125877 on Build GRCh38
rs983946125 Chr.7: 121126162-121126162 on Build GRCh38
rs745597995 Chr.7: 121125998-121125998 on Build GRCh38
rs1049240389 Chr.7: 121126094-121126095 on Build GRCh38
rs773109358 Chr.7: 121125920-121125920 on Build GRCh38
rs970388833 Chr.7: 121125939-121125939 on Build GRCh38
rs778607187 Chr.7: 121125953-121125955 on Build GRCh38
rs756206500 Chr.7: 121125944-121125945 on Build GRCh38
rs765139689 Chr.7: 121125943-121125944 on Build GRCh38
rs1011962888 Chr.7: 121126331-121126331 on Build GRCh38
rs1043036955 Chr.7: 121126247-121126247 on Build GRCh38
rs372612526 Chr.7: 121125957-121125957 on Build GRCh38
rs547322606 Chr.7: 121126165-121126165 on Build GRCh38
rs545698348 Chr.7: 121125888-121125888 on Build GRCh38
rs779135080 Chr.7: 121125892-121125892 on Build GRCh38
rs895293352 Chr.7: 121126286-121126286 on Build GRCh38
rs750012006 Chr.7: 121125953-121125953 on Build GRCh38
rs761734298 Chr.7: 121125911-121125911 on Build GRCh38
rs771399942 Chr.7: 121125901-121125901 on Build GRCh38
rs760761011 Chr.7: 121125930-121125930 on Build GRCh38
rs1029786266 Chr.7: 121125893-121125893 on Build GRCh38
rs79224464 Chr.7: 121125868-121125868 on Build GRCh38
rs779500438 Chr.7: 121125907-121125907 on Build GRCh38
rs758719880 Chr.7: 121125898-121125898 on Build GRCh38
rs146282574 Chr.7: 121126251-121126251 on Build GRCh38
rs181111347 Chr.7: 121126327-121126327 on Build GRCh38
rs1001002574 Chr.7: 121125896-121125896 on Build GRCh38
rs980583477 Chr.7: 121126309-121126309 on Build GRCh38
rs201765489 Chr.7: 121125908-121125908 on Build GRCh38
rs1056472350 Chr.7: 121125990-121125990 on Build GRCh38
rs868106310 Chr.7: 121125932-121125932 on Build GRCh38
rs1033975506 Chr.7: 121126315-121126315 on Build GRCh38
rs746939955 Chr.7: 121125845-121125845 on Build GRCh38
rs768354371 Chr.7: 121125852-121125852 on Build GRCh38
rs17537827 Chr.7: 121126186-121126186 on Build GRCh38
rs919310742 Chr.7: 121126058-121126058 on Build GRCh38

View Full Product Details