Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121350945-121351461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3C
Entrez Gene ID
10447
Gene Name
family with sequence similarity 3 member C
Gene Aliases
GS3786, ILEI
UniGene
Hs.434053
Gene Chromosome Location
Chr.7: 121348851-121396369 on Build GRCh38
Transcripts
NM_001040020.1 NM_014888.2 XM_011515736.2 XM_011515737.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6175907 c.561G>T p.G187G Chr.7: 121351176-121351176 on Build GRCh38
6175906 c.559G>T p.G187W Chr.7: 121351178-121351178 on Build GRCh38
2766460 c.487C>T p.R163W Chr.7: 121351250-121351250 on Build GRCh38
1736455 c.587T>G p.F196C Chr.7: 121351150-121351150 on Build GRCh38
6405391 c.575C>A p.T192K Chr.7: 121351162-121351162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902130189 Chr.7: 121350996-121350996 on Build GRCh38
rs749458653 Chr.7: 121351096-121351096 on Build GRCh38
rs760240079 Chr.7: 121351250-121351250 on Build GRCh38
rs752147998 Chr.7: 121351109-121351110 on Build GRCh38
rs766575834 Chr.7: 121350983-121350983 on Build GRCh38
rs759490654 Chr.7: 121351272-121351272 on Build GRCh38
rs746198821 Chr.7: 121351316-121351316 on Build GRCh38
rs747185717 Chr.7: 121351189-121351189 on Build GRCh38
rs752610999 Chr.7: 121351282-121351282 on Build GRCh38
rs774033092 Chr.7: 121351239-121351239 on Build GRCh38
rs756512268 Chr.7: 121351317-121351317 on Build GRCh38
rs917831381 Chr.7: 121351174-121351174 on Build GRCh38
rs764077045 Chr.7: 121351306-121351306 on Build GRCh38
rs766201371 Chr.7: 121351256-121351256 on Build GRCh38
rs1020990226 Chr.7: 121351026-121351026 on Build GRCh38
rs910969609 Chr.7: 121351308-121351308 on Build GRCh38
rs751763318 Chr.7: 121351213-121351213 on Build GRCh38
rs200009932 Chr.7: 121351169-121351169 on Build GRCh38
rs754842648 Chr.7: 121351162-121351162 on Build GRCh38
rs1046044727 Chr.7: 121351172-121351172 on Build GRCh38
rs184947477 Chr.7: 121350982-121350982 on Build GRCh38
rs776350244 Chr.7: 121351264-121351264 on Build GRCh38
rs377300911 Chr.7: 121351311-121351311 on Build GRCh38
rs189617673 Chr.7: 121351021-121351021 on Build GRCh38
rs772971347 Chr.7: 121351249-121351249 on Build GRCh38
rs979510193 Chr.7: 121351048-121351048 on Build GRCh38
rs1024343592 Chr.7: 121351211-121351211 on Build GRCh38
rs997513989 Chr.7: 121351023-121351023 on Build GRCh38
rs758833533 Chr.7: 121351143-121351144 on Build GRCh38
rs766071595 Chr.7: 121351118-121351118 on Build GRCh38
rs377173412 Chr.7: 121351165-121351165 on Build GRCh38
rs753878994 Chr.7: 121351128-121351128 on Build GRCh38
rs771457946 Chr.7: 121351245-121351245 on Build GRCh38
rs201667344 Chr.7: 121351398-121351399 on Build GRCh38
rs201961854 Chr.7: 121351278-121351278 on Build GRCh38
rs368786295 Chr.7: 121351163-121351163 on Build GRCh38
rs1018696879 Chr.7: 121351401-121351401 on Build GRCh38
rs781638542 Chr.7: 121351314-121351314 on Build GRCh38
rs113441026 Chr.7: 121351265-121351265 on Build GRCh38
rs538639884 Chr.7: 121351381-121351381 on Build GRCh38
rs1004428720 Chr.7: 121351209-121351209 on Build GRCh38
rs757367885 Chr.7: 121351313-121351313 on Build GRCh38
rs987263144 Chr.7: 121351396-121351396 on Build GRCh38
rs770377739 Chr.7: 121351236-121351236 on Build GRCh38
rs972360315 Chr.7: 121351182-121351182 on Build GRCh38
rs186189838 Chr.7: 121351075-121351075 on Build GRCh38
rs553410809 Chr.7: 121351341-121351341 on Build GRCh38
rs967277401 Chr.7: 121351421-121351421 on Build GRCh38
rs181911701 Chr.7: 121351047-121351047 on Build GRCh38
rs536321056 Chr.7: 121351090-121351090 on Build GRCh38
rs757708170 Chr.7: 121351195-121351195 on Build GRCh38
rs201572924 Chr.7: 121351089-121351089 on Build GRCh38
rs11768996 Chr.7: 121351435-121351435 on Build GRCh38
rs201209008 Chr.7: 121351253-121351253 on Build GRCh38
rs149263942 Chr.7: 121351179-121351179 on Build GRCh38
rs746395241 Chr.7: 121351216-121351216 on Build GRCh38
rs758531201 Chr.7: 121351284-121351284 on Build GRCh38
rs573035986 Chr.7: 121351359-121351359 on Build GRCh38
rs1041705482 Chr.7: 121350985-121350985 on Build GRCh38

View Full Product Details