Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121395990-121396529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3C
Entrez Gene ID
10447
Gene Name
family with sequence similarity 3 member C
Gene Aliases
GS3786, ILEI
UniGene
Hs.434053
Gene Chromosome Location
Chr.7: 121348851-121396369 on Build GRCh38
Transcripts
NM_001040020.1 NM_014888.2 XM_011515736.2

SNP Detail

SNP ID Chromosome Location
rs540665275 Chr.7: 121396032-121396032 on Build GRCh38
rs924333190 Chr.7: 121396364-121396364 on Build GRCh38
rs1043006308 Chr.7: 121396332-121396332 on Build GRCh38
rs922391893 Chr.7: 121396480-121396480 on Build GRCh38
rs972680687 Chr.7: 121396065-121396065 on Build GRCh38
rs889562762 Chr.7: 121396100-121396100 on Build GRCh38
rs889787795 Chr.7: 121396493-121396493 on Build GRCh38
rs970587033 Chr.7: 121396212-121396212 on Build GRCh38
rs191893545 Chr.7: 121396458-121396458 on Build GRCh38
rs1036886293 Chr.7: 121396469-121396469 on Build GRCh38
rs116965753 Chr.7: 121396063-121396063 on Build GRCh38
rs574359167 Chr.7: 121396498-121396498 on Build GRCh38
rs930995899 Chr.7: 121396078-121396078 on Build GRCh38
rs1012672298 Chr.7: 121396043-121396043 on Build GRCh38
rs983532530 Chr.7: 121396143-121396144 on Build GRCh38
rs1032408573 Chr.7: 121396170-121396170 on Build GRCh38
rs547309889 Chr.7: 121396454-121396454 on Build GRCh38
rs959481844 Chr.7: 121396181-121396181 on Build GRCh38
rs895155621 Chr.7: 121396411-121396411 on Build GRCh38
rs530387048 Chr.7: 121396331-121396331 on Build GRCh38
rs1039218493 Chr.7: 121396119-121396119 on Build GRCh38
rs976887028 Chr.7: 121396461-121396461 on Build GRCh38
rs10244034 Chr.7: 121396492-121396492 on Build GRCh38
rs1021210361 Chr.7: 121396140-121396140 on Build GRCh38
rs942489124 Chr.7: 121396296-121396296 on Build GRCh38
rs781073658 Chr.7: 121396436-121396436 on Build GRCh38
rs138090420 Chr.7: 121396112-121396112 on Build GRCh38
rs1012796891 Chr.7: 121396233-121396233 on Build GRCh38
rs1040807574 Chr.7: 121396303-121396303 on Build GRCh38
rs1003092452 Chr.7: 121396467-121396467 on Build GRCh38
rs187782127 Chr.7: 121396185-121396185 on Build GRCh38
rs918205910 Chr.7: 121396077-121396077 on Build GRCh38
rs983745076 Chr.7: 121396439-121396439 on Build GRCh38
rs952671066 Chr.7: 121396438-121396438 on Build GRCh38
rs60492718 Chr.7: 121396187-121396187 on Build GRCh38
rs954830146 Chr.7: 121396160-121396160 on Build GRCh38
rs1054060951 Chr.7: 121396204-121396204 on Build GRCh38
rs973036257 Chr.7: 121396230-121396230 on Build GRCh38
rs970936941 Chr.7: 121396434-121396434 on Build GRCh38
rs563478022 Chr.7: 121396075-121396077 on Build GRCh38
rs1054008800 Chr.7: 121396424-121396424 on Build GRCh38
rs4731011 Chr.7: 121396050-121396050 on Build GRCh38
rs1044356511 Chr.7: 121396239-121396239 on Build GRCh38
rs867504710 Chr.7: 121396317-121396317 on Build GRCh38
rs143597774 Chr.7: 121396291-121396291 on Build GRCh38
rs1015719599 Chr.7: 121396079-121396079 on Build GRCh38
rs902088029 Chr.7: 121396134-121396134 on Build GRCh38
rs901969080 Chr.7: 121396304-121396304 on Build GRCh38
rs539223233 Chr.7: 121396489-121396489 on Build GRCh38
rs930966525 Chr.7: 121396271-121396271 on Build GRCh38
rs918522370 Chr.7: 121396247-121396247 on Build GRCh38
rs933523203 Chr.7: 121396328-121396328 on Build GRCh38
rs1000882479 Chr.7: 121396164-121396164 on Build GRCh38
rs568938919 Chr.7: 121396337-121396337 on Build GRCh38
rs971474088 Chr.7: 121396055-121396055 on Build GRCh38
rs902489344 Chr.7: 121396144-121396144 on Build GRCh38
rs1032377372 Chr.7: 121396382-121396382 on Build GRCh38
rs559339642 Chr.7: 121396082-121396082 on Build GRCh38
rs561221425 Chr.7: 121396435-121396435 on Build GRCh38
rs943524414 Chr.7: 121396101-121396101 on Build GRCh38
rs530310459 Chr.7: 121396446-121396446 on Build GRCh38
rs907029893 Chr.7: 121396265-121396265 on Build GRCh38
rs759213247 Chr.7: 121396135-121396135 on Build GRCh38
rs1025074159 Chr.7: 121396422-121396422 on Build GRCh38
rs954243257 Chr.7: 121396315-121396315 on Build GRCh38
rs1012295853 Chr.7: 121396418-121396418 on Build GRCh38
rs550178528 Chr.7: 121396372-121396372 on Build GRCh38
rs1002735044 Chr.7: 121396288-121396288 on Build GRCh38
rs933473670 Chr.7: 121396487-121396487 on Build GRCh38
rs983776365 Chr.7: 121396290-121396290 on Build GRCh38
rs954296036 Chr.7: 121396127-121396127 on Build GRCh38

View Full Product Details