Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122301607-122302124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZF1
Entrez Gene ID
389549
Gene Name
FEZ family zinc finger 1
Gene Aliases
FEZ, HH22, ZNF312B
UniGene
Hs.553970
Gene Chromosome Location
Chr.7: 122300939-122311855 on Build GRCh38
Transcripts
NM_001024613.3 NM_001160264.2 XM_005250337.3 XM_011516202.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
744830 c.1304G>C p.G435A Chr.7: 122302121-122302121 on Build GRCh38
6222384 c.1373C>A p.P458Q Chr.7: 122302052-122302052 on Build GRCh38
3831745 c.1346C>T p.P449L Chr.7: 122302079-122302079 on Build GRCh38
1084774 c.1381C>T p.P461S Chr.7: 122302044-122302044 on Build GRCh38
6663263 c.1326G>A p.P442P Chr.7: 122302099-122302099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772257320 Chr.7: 122302012-122302012 on Build GRCh38
rs776136247 Chr.7: 122302058-122302058 on Build GRCh38
rs923234002 Chr.7: 122301797-122301797 on Build GRCh38
rs778049708 Chr.7: 122302021-122302062 on Build GRCh38
rs553922407 Chr.7: 122301913-122301913 on Build GRCh38
rs754858674 Chr.7: 122302077-122302077 on Build GRCh38
rs753092204 Chr.7: 122301967-122301967 on Build GRCh38
rs759943207 Chr.7: 122301955-122301955 on Build GRCh38
rs940218019 Chr.7: 122301943-122301943 on Build GRCh38
rs950214802 Chr.7: 122301672-122301672 on Build GRCh38
rs749959110 Chr.7: 122302065-122302065 on Build GRCh38
rs1055674717 Chr.7: 122301875-122301875 on Build GRCh38
rs1042516702 Chr.7: 122301836-122301836 on Build GRCh38
rs1040327488 Chr.7: 122302029-122302029 on Build GRCh38
rs867655977 Chr.7: 122302089-122302089 on Build GRCh38
rs998981964 Chr.7: 122301811-122301811 on Build GRCh38
rs957221734 Chr.7: 122301881-122301881 on Build GRCh38
rs778923284 Chr.7: 122302008-122302008 on Build GRCh38
rs756895628 Chr.7: 122301890-122301890 on Build GRCh38
rs1050295609 Chr.7: 122301750-122301751 on Build GRCh38
rs923181791 Chr.7: 122302035-122302035 on Build GRCh38
rs779633991 Chr.7: 122301990-122301990 on Build GRCh38
rs373295088 Chr.7: 122302010-122302010 on Build GRCh38
rs940125419 Chr.7: 122301728-122301731 on Build GRCh38
rs561681709 Chr.7: 122302082-122302082 on Build GRCh38
rs1018819504 Chr.7: 122301765-122301765 on Build GRCh38
rs758243120 Chr.7: 122302088-122302088 on Build GRCh38
rs558220470 Chr.7: 122301750-122301750 on Build GRCh38
rs993103368 Chr.7: 122301931-122301931 on Build GRCh38
rs1040401142 Chr.7: 122301756-122301756 on Build GRCh38
rs201400828 Chr.7: 122301727-122301728 on Build GRCh38
rs908693905 Chr.7: 122301929-122301929 on Build GRCh38
rs765509411 Chr.7: 122302053-122302053 on Build GRCh38
rs957073789 Chr.7: 122301632-122301632 on Build GRCh38
rs759996308 Chr.7: 122302051-122302051 on Build GRCh38
rs920024848 Chr.7: 122301947-122301947 on Build GRCh38
rs971075155 Chr.7: 122301909-122301909 on Build GRCh38
rs754661273 Chr.7: 122302003-122302003 on Build GRCh38
rs908618608 Chr.7: 122301717-122301717 on Build GRCh38
rs747114666 Chr.7: 122302094-122302094 on Build GRCh38
rs575879164 Chr.7: 122301647-122301647 on Build GRCh38
rs967220929 Chr.7: 122301796-122301796 on Build GRCh38
rs1013754348 Chr.7: 122301888-122301888 on Build GRCh38
rs556444210 Chr.7: 122302086-122302086 on Build GRCh38
rs1026091732 Chr.7: 122301946-122301946 on Build GRCh38
rs775695735 Chr.7: 122302102-122302102 on Build GRCh38
rs777800892 Chr.7: 122302091-122302091 on Build GRCh38
rs537852878 Chr.7: 122301709-122301709 on Build GRCh38
rs895419818 Chr.7: 122301885-122301885 on Build GRCh38
rs749111526 Chr.7: 122302000-122302000 on Build GRCh38
rs971820357 Chr.7: 122301636-122301636 on Build GRCh38
rs766932093 Chr.7: 122301985-122301985 on Build GRCh38
rs747280440 Chr.7: 122302031-122302031 on Build GRCh38
rs916153286 Chr.7: 122301907-122301907 on Build GRCh38
rs755742704 Chr.7: 122301987-122301987 on Build GRCh38
rs985016683 Chr.7: 122301702-122301702 on Build GRCh38
rs1001070986 Chr.7: 122301864-122301864 on Build GRCh38
rs771299112 Chr.7: 122302036-122302036 on Build GRCh38
rs766021955 Chr.7: 122302076-122302076 on Build GRCh38
rs756852930 Chr.7: 122301983-122301983 on Build GRCh38
rs1025171170 Chr.7: 122301891-122301891 on Build GRCh38
rs386717352 Chr.7: 122301748-122301750 on Build GRCh38
rs577039958 Chr.7: 122302028-122302028 on Build GRCh38
rs777111886 Chr.7: 122301951-122301951 on Build GRCh38
rs1055303695 Chr.7: 122302087-122302087 on Build GRCh38
rs767104973 Chr.7: 122302062-122302062 on Build GRCh38
rs776929932 Chr.7: 122302039-122302039 on Build GRCh38
rs750116742 Chr.7: 122301986-122301986 on Build GRCh38
rs1045909958 Chr.7: 122301699-122301699 on Build GRCh38
rs534384756 Chr.7: 122301874-122301874 on Build GRCh38
rs747374890 Chr.7: 122302047-122302048 on Build GRCh38
rs951935623 Chr.7: 122301653-122301653 on Build GRCh38
rs746077863 Chr.7: 122302097-122302097 on Build GRCh38
rs1038473968 Chr.7: 122301732-122301737 on Build GRCh38
rs765781031 Chr.7: 122301957-122301957 on Build GRCh38
rs550112024 Chr.7: 122301651-122301651 on Build GRCh38
rs945352684 Chr.7: 122301835-122301835 on Build GRCh38
rs375261549 Chr.7: 122301758-122301758 on Build GRCh38
rs537162595 Chr.7: 122301748-122301748 on Build GRCh38
rs144474907 Chr.7: 122302103-122302103 on Build GRCh38
rs1038830409 Chr.7: 122301945-122301945 on Build GRCh38
rs763337621 Chr.7: 122302061-122302061 on Build GRCh38
rs569233836 Chr.7: 122301691-122301691 on Build GRCh38
rs112066691 Chr.7: 122302075-122302075 on Build GRCh38
rs898464159 Chr.7: 122301738-122301738 on Build GRCh38
rs753403078 Chr.7: 122301645-122301645 on Build GRCh38
rs964630833 Chr.7: 122302018-122302018 on Build GRCh38
rs903919438 Chr.7: 122301842-122301842 on Build GRCh38
rs981555542 Chr.7: 122301917-122301917 on Build GRCh38

View Full Product Details