Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 123629191-123629704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB15
Entrez Gene ID
142685
Gene Name
ankyrin repeat and SOCS box containing 15
Gene Aliases
-
UniGene
Hs.83916
Gene Chromosome Location
Chr.7: 123567003-123637900 on Build GRCh38
Transcripts
NM_001290258.1 NM_080928.4 XM_005250146.4 XM_011515816.2 XM_011515817.2 XM_011515818.2 XM_011515819.2 XM_011515820.2 XM_011515821.2 XM_017011756.1 XM_017011757.1 XM_017011758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6023089 c.1256C>A p.P419H Chr.7: 123629250-123629250 on Build GRCh38
345896 c.1432G>C p.D478H Chr.7: 123629426-123629426 on Build GRCh38
452250 c.1315G>A p.G439S Chr.7: 123629309-123629309 on Build GRCh38
7129154 c.1296G>C p.L432L Chr.7: 123629290-123629290 on Build GRCh38
7134065 c.1208G>C p.G403A Chr.7: 123629202-123629202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184541144 Chr.7: 123629641-123629641 on Build GRCh38
rs750793743 Chr.7: 123629230-123629230 on Build GRCh38
rs770056947 Chr.7: 123629369-123629369 on Build GRCh38
rs199686253 Chr.7: 123629372-123629372 on Build GRCh38
rs755684103 Chr.7: 123629457-123629457 on Build GRCh38
rs779941765 Chr.7: 123629459-123629459 on Build GRCh38
rs774628678 Chr.7: 123629398-123629398 on Build GRCh38
rs770541778 Chr.7: 123629262-123629262 on Build GRCh38
rs111961710 Chr.7: 123629619-123629619 on Build GRCh38
rs757268616 Chr.7: 123629601-123629601 on Build GRCh38
rs917372335 Chr.7: 123629549-123629549 on Build GRCh38
rs897811731 Chr.7: 123629646-123629646 on Build GRCh38
rs1046957787 Chr.7: 123629480-123629480 on Build GRCh38
rs192721201 Chr.7: 123629505-123629505 on Build GRCh38
rs756964826 Chr.7: 123629363-123629363 on Build GRCh38
rs368938159 Chr.7: 123629439-123629439 on Build GRCh38
rs766591373 Chr.7: 123629324-123629324 on Build GRCh38
rs201469071 Chr.7: 123629318-123629318 on Build GRCh38
rs114514161 Chr.7: 123629504-123629504 on Build GRCh38
rs532087319 Chr.7: 123629592-123629592 on Build GRCh38
rs376348325 Chr.7: 123629382-123629382 on Build GRCh38
rs997927896 Chr.7: 123629546-123629546 on Build GRCh38
rs750098681 Chr.7: 123629454-123629454 on Build GRCh38
rs774257065 Chr.7: 123629288-123629288 on Build GRCh38
rs531057086 Chr.7: 123629404-123629404 on Build GRCh38
rs777095284 Chr.7: 123629251-123629251 on Build GRCh38
rs115142564 Chr.7: 123629472-123629472 on Build GRCh38
rs773581932 Chr.7: 123629407-123629407 on Build GRCh38
rs950158639 Chr.7: 123629566-123629566 on Build GRCh38
rs762741345 Chr.7: 123629278-123629278 on Build GRCh38
rs374788183 Chr.7: 123629297-123629297 on Build GRCh38
rs144320147 Chr.7: 123629367-123629367 on Build GRCh38
rs200113553 Chr.7: 123629248-123629248 on Build GRCh38
rs372451951 Chr.7: 123629341-123629341 on Build GRCh38
rs376758380 Chr.7: 123629263-123629263 on Build GRCh38
rs370567182 Chr.7: 123629387-123629387 on Build GRCh38
rs774944187 Chr.7: 123629434-123629434 on Build GRCh38
rs780992291 Chr.7: 123629395-123629395 on Build GRCh38
rs754119292 Chr.7: 123629425-123629425 on Build GRCh38
rs767412628 Chr.7: 123629267-123629267 on Build GRCh38
rs750883849 Chr.7: 123629308-123629308 on Build GRCh38
rs149605351 Chr.7: 123629279-123629279 on Build GRCh38
rs769692009 Chr.7: 123629275-123629275 on Build GRCh38
rs888120681 Chr.7: 123629498-123629498 on Build GRCh38
rs749699178 Chr.7: 123629424-123629424 on Build GRCh38
rs745652824 Chr.7: 123629265-123629265 on Build GRCh38
rs148775577 Chr.7: 123629431-123629431 on Build GRCh38
rs1030958063 Chr.7: 123629628-123629628 on Build GRCh38
rs760926805 Chr.7: 123629421-123629421 on Build GRCh38
rs923451347 Chr.7: 123629248-123629248 on Build GRCh38
rs372138258 Chr.7: 123629218-123629218 on Build GRCh38
rs754388714 Chr.7: 123629340-123629340 on Build GRCh38
rs1006657131 Chr.7: 123629412-123629412 on Build GRCh38
rs775502718 Chr.7: 123629276-123629276 on Build GRCh38
rs761979517 Chr.7: 123629399-123629399 on Build GRCh38
rs1055938615 Chr.7: 123629523-123629523 on Build GRCh38
rs369173243 Chr.7: 123629287-123629287 on Build GRCh38
rs865846460 Chr.7: 123629432-123629432 on Build GRCh38
rs73718442 Chr.7: 123629226-123629226 on Build GRCh38
rs143332776 Chr.7: 123629502-123629502 on Build GRCh38
rs1026801680 Chr.7: 123629373-123629373 on Build GRCh38
rs767837021 Chr.7: 123629300-123629300 on Build GRCh38
rs371037323 Chr.7: 123629465-123629465 on Build GRCh38
rs756768568 Chr.7: 123629436-123629436 on Build GRCh38
rs746692691 Chr.7: 123629257-123629257 on Build GRCh38
rs937201875 Chr.7: 123629455-123629455 on Build GRCh38
rs149642706 Chr.7: 123629244-123629244 on Build GRCh38
rs780822240 Chr.7: 123629364-123629364 on Build GRCh38
rs373669636 Chr.7: 123629433-123629433 on Build GRCh38
rs545385636 Chr.7: 123629296-123629296 on Build GRCh38
rs751050300 Chr.7: 123629361-123629361 on Build GRCh38

View Full Product Details