Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151218483-151218983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF2
Entrez Gene ID
10061
Gene Name
ATP binding cassette subfamily F member 2
Gene Aliases
ABC28, EST133090, HUSSY-18, HUSSY18
UniGene
Hs.654958
Gene Chromosome Location
Chr.7: 151207837-151227374 on Build GRCh38
Transcripts
NM_005692.4 NM_007189.2 XM_017011639.1 XM_017011640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3411817 c.1069C>T p.Q357* Chr.7: 151218822-151218822 on Build GRCh38
6439523 c.1073G>A p.S358N Chr.7: 151218818-151218818 on Build GRCh38
5407014 c.1160C>T p.P387L Chr.7: 151218628-151218628 on Build GRCh38
6439522 c.1073G>A p.S358N Chr.7: 151218818-151218818 on Build GRCh38
4813504 c.1052A>G p.K351R Chr.7: 151218839-151218839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748329981 Chr.7: 151218542-151218542 on Build GRCh38
rs528203375 Chr.7: 151218663-151218663 on Build GRCh38
rs970235812 Chr.7: 151218678-151218678 on Build GRCh38
rs376119610 Chr.7: 151218654-151218654 on Build GRCh38
rs769522970 Chr.7: 151218536-151218536 on Build GRCh38
rs377001127 Chr.7: 151218793-151218793 on Build GRCh38
rs761396504 Chr.7: 151218772-151218772 on Build GRCh38
rs775988044 Chr.7: 151218765-151218765 on Build GRCh38
rs139539137 Chr.7: 151218766-151218766 on Build GRCh38
rs761498247 Chr.7: 151218892-151218892 on Build GRCh38
rs573905461 Chr.7: 151218930-151218930 on Build GRCh38
rs780645129 Chr.7: 151218871-151218871 on Build GRCh38
rs765884970 Chr.7: 151218907-151218907 on Build GRCh38
rs1040351411 Chr.7: 151218847-151218847 on Build GRCh38
rs777983145 Chr.7: 151218734-151218734 on Build GRCh38
rs776602190 Chr.7: 151218757-151218757 on Build GRCh38
rs747208202 Chr.7: 151218739-151218739 on Build GRCh38
rs1021014683 Chr.7: 151218615-151218615 on Build GRCh38
rs768386601 Chr.7: 151218687-151218687 on Build GRCh38
rs770918183 Chr.7: 151218623-151218623 on Build GRCh38
rs764103686 Chr.7: 151218863-151218863 on Build GRCh38
rs769018927 Chr.7: 151218884-151218884 on Build GRCh38
rs528097798 Chr.7: 151218949-151218949 on Build GRCh38
rs572488243 Chr.7: 151218832-151218832 on Build GRCh38
rs778572651 Chr.7: 151218717-151218731 on Build GRCh38
rs12113924 Chr.7: 151218947-151218947 on Build GRCh38
rs200508522 Chr.7: 151218868-151218868 on Build GRCh38
rs759497008 Chr.7: 151218911-151218911 on Build GRCh38
rs781305827 Chr.7: 151218853-151218853 on Build GRCh38
rs78511279 Chr.7: 151218573-151218573 on Build GRCh38
rs756722846 Chr.7: 151218675-151218675 on Build GRCh38
rs773150864 Chr.7: 151218594-151218594 on Build GRCh38
rs893271135 Chr.7: 151218934-151218934 on Build GRCh38
rs747715357 Chr.7: 151218851-151218852 on Build GRCh38
rs528875227 Chr.7: 151218505-151218505 on Build GRCh38
rs753724963 Chr.7: 151218909-151218909 on Build GRCh38
rs1031762016 Chr.7: 151218535-151218535 on Build GRCh38
rs370361417 Chr.7: 151218805-151218805 on Build GRCh38
rs931173158 Chr.7: 151218780-151218780 on Build GRCh38
rs760110218 Chr.7: 151218896-151218896 on Build GRCh38
rs919719033 Chr.7: 151218775-151218775 on Build GRCh38
rs1050051334 Chr.7: 151218781-151218781 on Build GRCh38
rs968078037 Chr.7: 151218628-151218628 on Build GRCh38
rs758614030 Chr.7: 151218733-151218733 on Build GRCh38
rs774611657 Chr.7: 151218891-151218891 on Build GRCh38
rs1000831940 Chr.7: 151218638-151218638 on Build GRCh38
rs145160370 Chr.7: 151218806-151218806 on Build GRCh38
rs779848869 Chr.7: 151218537-151218537 on Build GRCh38
rs550165555 Chr.7: 151218760-151218760 on Build GRCh38
rs535828109 Chr.7: 151218778-151218778 on Build GRCh38
rs542198209 Chr.7: 151218935-151218935 on Build GRCh38
rs770469392 Chr.7: 151218762-151218762 on Build GRCh38
rs368530879 Chr.7: 151218534-151218534 on Build GRCh38
rs142338230 Chr.7: 151218725-151218725 on Build GRCh38
rs559000292 Chr.7: 151218660-151218660 on Build GRCh38
rs7782699 Chr.7: 151218862-151218862 on Build GRCh38
rs146657660 Chr.7: 151218912-151218912 on Build GRCh38
rs971865980 Chr.7: 151218722-151218722 on Build GRCh38
rs959666710 Chr.7: 151218953-151218953 on Build GRCh38
rs543644640 Chr.7: 151218872-151218872 on Build GRCh38
rs759980570 Chr.7: 151218646-151218646 on Build GRCh38
rs749941180 Chr.7: 151218787-151218787 on Build GRCh38
rs376560255 Chr.7: 151218518-151218518 on Build GRCh38
rs372876973 Chr.7: 151218598-151218598 on Build GRCh38
rs776033270 Chr.7: 151218655-151218655 on Build GRCh38
rs959398086 Chr.7: 151218656-151218660 on Build GRCh38
rs943040588 Chr.7: 151218829-151218829 on Build GRCh38
rs778593929 Chr.7: 151218830-151218830 on Build GRCh38
rs772696955 Chr.7: 151218895-151218895 on Build GRCh38
rs375328067 Chr.7: 151218893-151218893 on Build GRCh38
rs201524586 Chr.7: 151218812-151218812 on Build GRCh38
rs766952382 Chr.7: 151218682-151218682 on Build GRCh38
rs770990449 Chr.7: 151218754-151218754 on Build GRCh38
rs71196740 Chr.7: 151218717-151218718 on Build GRCh38
rs1023583718 Chr.7: 151218959-151218959 on Build GRCh38
rs774000810 Chr.7: 151218769-151218769 on Build GRCh38
rs373325837 Chr.7: 151218719-151218719 on Build GRCh38
rs35102612 Chr.7: 151218668-151218668 on Build GRCh38
rs771603542 Chr.7: 151218886-151218887 on Build GRCh38
rs200403284 Chr.7: 151218702-151218702 on Build GRCh38
rs373824471 Chr.7: 151218731-151218731 on Build GRCh38
rs754707891 Chr.7: 151218826-151218826 on Build GRCh38
rs535093235 Chr.7: 151218839-151218839 on Build GRCh38
rs777136951 Chr.7: 151218629-151218629 on Build GRCh38

View Full Product Details