Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 154669246-154669750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP6
Entrez Gene ID
1804
Gene Name
dipeptidyl peptidase like 6
Gene Aliases
DPL1, DPPX, MRD33, VF2
UniGene
Hs.490684
Gene Chromosome Location
Chr.7: 153747986-154894290 on Build GRCh38
Transcripts
NM_001039350.2 NM_001290252.1 NM_001936.4 NM_130797.3 XM_017011808.1 XM_017011809.1 XM_017011810.1 XM_017011811.1 XM_017011812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950233 c.508C>A p.P170T Chr.7: 154669379-154669379 on Build GRCh38
7230480 c.499C>T p.P167S Chr.7: 154669364-154669364 on Build GRCh38
1449519 c.565G>T p.G189C Chr.7: 154669430-154669430 on Build GRCh38
3685119 c.537A>T p.K179N Chr.7: 154669402-154669402 on Build GRCh38
5925345 c.503A>C p.Q168P Chr.7: 154669368-154669368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919437758 Chr.7: 154669543-154669543 on Build GRCh38
rs3807218 Chr.7: 154669402-154669402 on Build GRCh38
rs3220067 Chr.7: 154669564-154669567 on Build GRCh38
rs538699484 Chr.7: 154669371-154669371 on Build GRCh38
rs59801709 Chr.7: 154669566-154669567 on Build GRCh38
rs143184362 Chr.7: 154669698-154669698 on Build GRCh38
rs72056937 Chr.7: 154669539-154669540 on Build GRCh38
rs888872472 Chr.7: 154669350-154669350 on Build GRCh38
rs1032496284 Chr.7: 154669643-154669643 on Build GRCh38
rs761485998 Chr.7: 154669543-154669544 on Build GRCh38
rs998963409 Chr.7: 154669330-154669330 on Build GRCh38
rs893705796 Chr.7: 154669646-154669646 on Build GRCh38
rs181488953 Chr.7: 154669685-154669685 on Build GRCh38
rs542967433 Chr.7: 154669452-154669452 on Build GRCh38
rs781499961 Chr.7: 154669400-154669400 on Build GRCh38
rs543794303 Chr.7: 154669585-154669585 on Build GRCh38
rs774687375 Chr.7: 154669279-154669279 on Build GRCh38
rs1035622189 Chr.7: 154669518-154669518 on Build GRCh38
rs780817339 Chr.7: 154669539-154669544 on Build GRCh38
rs993711027 Chr.7: 154669506-154669506 on Build GRCh38
rs561610953 Chr.7: 154669472-154669472 on Build GRCh38
rs902362922 Chr.7: 154669684-154669684 on Build GRCh38
rs756505789 Chr.7: 154669683-154669683 on Build GRCh38
rs775378449 Chr.7: 154669407-154669407 on Build GRCh38
rs763515808 Chr.7: 154669421-154669421 on Build GRCh38
rs778919869 Chr.7: 154669392-154669392 on Build GRCh38
rs190997653 Chr.7: 154669669-154669669 on Build GRCh38
rs750773891 Chr.7: 154669541-154669544 on Build GRCh38
rs1054756775 Chr.7: 154669632-154669632 on Build GRCh38
rs1010808256 Chr.7: 154669660-154669660 on Build GRCh38
rs1050055966 Chr.7: 154669349-154669349 on Build GRCh38
rs368464051 Chr.7: 154669310-154669310 on Build GRCh38
rs963665075 Chr.7: 154669495-154669495 on Build GRCh38
rs769925137 Chr.7: 154669405-154669405 on Build GRCh38
rs565077306 Chr.7: 154669626-154669626 on Build GRCh38
rs755139442 Chr.7: 154669700-154669700 on Build GRCh38
rs973552345 Chr.7: 154669505-154669505 on Build GRCh38
rs371506958 Chr.7: 154669565-154669566 on Build GRCh38
rs554408118 Chr.7: 154669384-154669384 on Build GRCh38
rs752177161 Chr.7: 154669328-154669328 on Build GRCh38
rs763499060 Chr.7: 154669323-154669323 on Build GRCh38
rs954737131 Chr.7: 154669364-154669364 on Build GRCh38
rs754951037 Chr.7: 154669366-154669366 on Build GRCh38
rs575480581 Chr.7: 154669636-154669636 on Build GRCh38
rs113346453 Chr.7: 154669510-154669683 on Build GRCh38
rs779783497 Chr.7: 154669466-154669466 on Build GRCh38
rs879183486 Chr.7: 154669541-154669542 on Build GRCh38
rs750838697 Chr.7: 154669608-154669608 on Build GRCh38
rs1030724179 Chr.7: 154669339-154669339 on Build GRCh38
rs927584422 Chr.7: 154669615-154669615 on Build GRCh38
rs866615121 Chr.7: 154669561-154669567 on Build GRCh38
rs929021883 Chr.7: 154669560-154669560 on Build GRCh38
rs771327985 Chr.7: 154669393-154669393 on Build GRCh38
rs753825686 Chr.7: 154669329-154669329 on Build GRCh38
rs910801204 Chr.7: 154669466-154669466 on Build GRCh38
rs1023588356 Chr.7: 154669613-154669613 on Build GRCh38
rs372750485 Chr.7: 154669314-154669314 on Build GRCh38
rs755946605 Chr.7: 154669567-154669568 on Build GRCh38
rs59816725 Chr.7: 154669562-154669567 on Build GRCh38
rs35719981 Chr.7: 154669539-154669542 on Build GRCh38
rs779363142 Chr.7: 154669313-154669313 on Build GRCh38
rs4960721 Chr.7: 154669581-154669581 on Build GRCh38

View Full Product Details