Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157415938-157416448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB6
Entrez Gene ID
10049
Gene Name
DnaJ heat shock protein family (Hsp40) member B6
Gene Aliases
DJ4, DnaJ, HHDJ1, HSJ-2, HSJ2, LGMD1D, LGMD1E, MRJ, MSJ-1
UniGene
Hs.490745
Gene Chromosome Location
Chr.7: 157337004-157417439 on Build GRCh38
Transcripts
NM_058246.3 XM_006715823.1 XM_017011633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582715 c.908A>G p.E303G Chr.7: 157416025-157416025 on Build GRCh38
4939769 c.918G>A p.K306K Chr.7: 157416035-157416035 on Build GRCh38
5985277 c.962C>T p.S321L Chr.7: 157416079-157416079 on Build GRCh38
1449677 c.948_950delGAA p.K320delK Chr.7: 157416065-157416067 on Build GRCh38
255896 c.958_960delAAG p.K320delK Chr.7: 157416075-157416077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965557122 Chr.7: 157416159-157416159 on Build GRCh38
rs1030976259 Chr.7: 157416185-157416185 on Build GRCh38
rs776962478 Chr.7: 157416027-157416027 on Build GRCh38
rs554220388 Chr.7: 157416368-157416368 on Build GRCh38
rs542232099 Chr.7: 157416349-157416349 on Build GRCh38
rs1015881680 Chr.7: 157416315-157416315 on Build GRCh38
rs576133569 Chr.7: 157416404-157416404 on Build GRCh38
rs763062645 Chr.7: 157416132-157416132 on Build GRCh38
rs953620507 Chr.7: 157416313-157416313 on Build GRCh38
rs372172001 Chr.7: 157416120-157416120 on Build GRCh38
rs377063562 Chr.7: 157416126-157416126 on Build GRCh38
rs1011792317 Chr.7: 157416158-157416158 on Build GRCh38
rs749767817 Chr.7: 157415987-157415988 on Build GRCh38
rs886042962 Chr.7: 157416075-157416076 on Build GRCh38
rs775708476 Chr.7: 157416049-157416049 on Build GRCh38
rs192037366 Chr.7: 157416121-157416121 on Build GRCh38
rs369221072 Chr.7: 157416141-157416141 on Build GRCh38
rs886062131 Chr.7: 157416387-157416387 on Build GRCh38
rs371304535 Chr.7: 157416064-157416064 on Build GRCh38
rs544983219 Chr.7: 157416287-157416287 on Build GRCh38
rs764933589 Chr.7: 157416215-157416215 on Build GRCh38
rs140445215 Chr.7: 157416101-157416101 on Build GRCh38
rs879110420 Chr.7: 157416193-157416193 on Build GRCh38
rs140516420 Chr.7: 157416330-157416330 on Build GRCh38
rs949220233 Chr.7: 157416244-157416244 on Build GRCh38
rs556372247 Chr.7: 157416286-157416286 on Build GRCh38
rs567762107 Chr.7: 157416238-157416238 on Build GRCh38
rs1028887540 Chr.7: 157416311-157416311 on Build GRCh38
rs534203597 Chr.7: 157416203-157416203 on Build GRCh38
rs1046335065 Chr.7: 157416262-157416262 on Build GRCh38
rs990304178 Chr.7: 157416018-157416018 on Build GRCh38
rs767898412 Chr.7: 157416076-157416076 on Build GRCh38
rs759467623 Chr.7: 157416314-157416314 on Build GRCh38
rs142165399 Chr.7: 157416145-157416145 on Build GRCh38
rs374432835 Chr.7: 157416003-157416003 on Build GRCh38
rs939042113 Chr.7: 157416397-157416397 on Build GRCh38
rs551156852 Chr.7: 157416156-157416156 on Build GRCh38
rs773534306 Chr.7: 157416219-157416219 on Build GRCh38
rs779513290 Chr.7: 157415998-157415998 on Build GRCh38
rs779492467 Chr.7: 157416004-157416004 on Build GRCh38
rs765886234 Chr.7: 157416045-157416045 on Build GRCh38
rs139193261 Chr.7: 157416186-157416186 on Build GRCh38
rs926566285 Chr.7: 157416387-157416387 on Build GRCh38
rs1017166941 Chr.7: 157416333-157416333 on Build GRCh38
rs768816202 Chr.7: 157416134-157416134 on Build GRCh38
rs1003356872 Chr.7: 157416125-157416125 on Build GRCh38
rs377683714 Chr.7: 157416151-157416151 on Build GRCh38
rs538057079 Chr.7: 157416102-157416102 on Build GRCh38
rs767372593 Chr.7: 157416082-157416082 on Build GRCh38
rs926520826 Chr.7: 157416265-157416265 on Build GRCh38
rs893103166 Chr.7: 157416278-157416278 on Build GRCh38
rs746303138 Chr.7: 157416124-157416124 on Build GRCh38
rs1044074680 Chr.7: 157416415-157416415 on Build GRCh38
rs187742799 Chr.7: 157415985-157415985 on Build GRCh38
rs375911119 Chr.7: 157416091-157416091 on Build GRCh38
rs982347227 Chr.7: 157416375-157416375 on Build GRCh38
rs561059767 Chr.7: 157416144-157416144 on Build GRCh38
rs886062130 Chr.7: 157416216-157416216 on Build GRCh38
rs778749161 Chr.7: 157416065-157416067 on Build GRCh38
rs886043804 Chr.7: 157416075-157416077 on Build GRCh38
rs771151320 Chr.7: 157416117-157416117 on Build GRCh38
rs756598628 Chr.7: 157416274-157416274 on Build GRCh38
rs748544225 Chr.7: 157416093-157416094 on Build GRCh38
rs953132658 Chr.7: 157416081-157416081 on Build GRCh38
rs755524240 Chr.7: 157416042-157416042 on Build GRCh38
rs1042165524 Chr.7: 157416280-157416280 on Build GRCh38
rs541259018 Chr.7: 157416188-157416188 on Build GRCh38
rs866911588 Chr.7: 157416367-157416367 on Build GRCh38
rs565527346 Chr.7: 157416065-157416065 on Build GRCh38
rs571981993 Chr.7: 157416340-157416340 on Build GRCh38
rs146779424 Chr.7: 157416296-157416296 on Build GRCh38
rs972498518 Chr.7: 157416192-157416192 on Build GRCh38
rs759872211 Chr.7: 157416075-157416075 on Build GRCh38
rs755568589 Chr.7: 157415992-157415992 on Build GRCh38
rs989362447 Chr.7: 157416407-157416407 on Build GRCh38
rs767669570 Chr.7: 157416138-157416138 on Build GRCh38
rs946554053 Chr.7: 157416411-157416411 on Build GRCh38
rs754496495 Chr.7: 157416114-157416114 on Build GRCh38
rs972382657 Chr.7: 157416361-157416361 on Build GRCh38
rs577875381 Chr.7: 157416116-157416116 on Build GRCh38
rs549360686 Chr.7: 157416220-157416220 on Build GRCh38
rs748781499 Chr.7: 157416103-157416103 on Build GRCh38
rs150779247 Chr.7: 157416171-157416171 on Build GRCh38
rs369235805 Chr.7: 157416115-157416115 on Build GRCh38
rs754405567 Chr.7: 157416146-157416146 on Build GRCh38
rs116630040 Chr.7: 157416170-157416170 on Build GRCh38
rs764284901 Chr.7: 157416279-157416279 on Build GRCh38
rs998313618 Chr.7: 157416297-157416297 on Build GRCh38
rs147168661 Chr.7: 157416078-157416078 on Build GRCh38
rs946600853 Chr.7: 157416276-157416276 on Build GRCh38
rs369741728 Chr.7: 157416131-157416131 on Build GRCh38
rs971860158 Chr.7: 157416341-157416341 on Build GRCh38
rs753180096 Chr.7: 157416256-157416256 on Build GRCh38
rs550867169 Chr.7: 157415984-157415984 on Build GRCh38
rs760098587 Chr.7: 157416040-157416040 on Build GRCh38
rs1026466800 Chr.7: 157416354-157416354 on Build GRCh38
rs954402824 Chr.7: 157416166-157416167 on Build GRCh38
rs537908243 Chr.7: 157416275-157416275 on Build GRCh38
rs776254454 Chr.7: 157416173-157416173 on Build GRCh38
rs562936080 Chr.7: 157416162-157416162 on Build GRCh38
rs142974468 Chr.7: 157416079-157416079 on Build GRCh38
rs542342637 Chr.7: 157416137-157416137 on Build GRCh38
rs774391788 Chr.7: 157416135-157416135 on Build GRCh38
rs78337193 Chr.7: 157416010-157416010 on Build GRCh38

View Full Product Details