Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 467628-468126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO25
Entrez Gene ID
26260
Gene Name
F-box protein 25
Gene Aliases
FBX25
UniGene
Hs.438454
Gene Chromosome Location
Chr.8: 406808-477967 on Build GRCh38
Transcripts
NM_012173.3 NM_183420.1 NM_183421.1 XM_011534748.2 XM_011534749.1 XM_017013313.1 XM_017013314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834734 c.988G>T p.D330Y Chr.8: 467720-467720 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs989211801 Chr.8: 467794-467794 on Build GRCh38
rs919187296 Chr.8: 468091-468091 on Build GRCh38
rs56125068 Chr.8: 468000-468000 on Build GRCh38
rs1021402869 Chr.8: 467897-467897 on Build GRCh38
rs1000881585 Chr.8: 468042-468042 on Build GRCh38
rs751125069 Chr.8: 467743-467743 on Build GRCh38
rs928987461 Chr.8: 467988-467988 on Build GRCh38
rs1032733347 Chr.8: 468053-468053 on Build GRCh38
rs577737681 Chr.8: 468012-468012 on Build GRCh38
rs201672813 Chr.8: 467757-467757 on Build GRCh38
rs571386119 Chr.8: 467887-467887 on Build GRCh38
rs944601998 Chr.8: 467828-467828 on Build GRCh38
rs902725903 Chr.8: 468041-468041 on Build GRCh38
rs1042642675 Chr.8: 468040-468040 on Build GRCh38
rs186553111 Chr.8: 467896-467896 on Build GRCh38
rs753248273 Chr.8: 467766-467766 on Build GRCh38
rs763692638 Chr.8: 467734-467734 on Build GRCh38
rs17745485 Chr.8: 467836-467836 on Build GRCh38
rs191346011 Chr.8: 467773-467773 on Build GRCh38
rs377525243 Chr.8: 467764-467764 on Build GRCh38
rs912124353 Chr.8: 467819-467819 on Build GRCh38
rs1028842750 Chr.8: 468098-468098 on Build GRCh38
rs373102655 Chr.8: 467673-467673 on Build GRCh38
rs878912257 Chr.8: 467968-467968 on Build GRCh38
rs1032430280 Chr.8: 467920-467920 on Build GRCh38
rs540279334 Chr.8: 468081-468081 on Build GRCh38
rs984494139 Chr.8: 468103-468103 on Build GRCh38
rs576494406 Chr.8: 468004-468004 on Build GRCh38
rs958142123 Chr.8: 467791-467791 on Build GRCh38
rs965846270 Chr.8: 468071-468071 on Build GRCh38
rs975962236 Chr.8: 467953-467953 on Build GRCh38
rs1001397580 Chr.8: 467908-467908 on Build GRCh38
rs773238558 Chr.8: 467701-467701 on Build GRCh38
rs369351258 Chr.8: 468002-468002 on Build GRCh38
rs377588678 Chr.8: 467799-467799 on Build GRCh38
rs1002379508 Chr.8: 467846-467846 on Build GRCh38
rs370757310 Chr.8: 467759-467759 on Build GRCh38
rs995450583 Chr.8: 467854-467854 on Build GRCh38
rs1055796588 Chr.8: 467860-467860 on Build GRCh38
rs757284702 Chr.8: 468066-468066 on Build GRCh38
rs201708248 Chr.8: 467710-467710 on Build GRCh38
rs748230367 Chr.8: 467685-467685 on Build GRCh38
rs755521753 Chr.8: 467675-467675 on Build GRCh38
rs772202305 Chr.8: 467698-467698 on Build GRCh38
rs1045824140 Chr.8: 468054-468054 on Build GRCh38
rs566620699 Chr.8: 467929-467929 on Build GRCh38
rs1014168011 Chr.8: 467883-467883 on Build GRCh38
rs747283349 Chr.8: 467703-467703 on Build GRCh38
rs183326199 Chr.8: 467840-467840 on Build GRCh38
rs896645308 Chr.8: 468028-468028 on Build GRCh38
rs116561723 Chr.8: 467835-467835 on Build GRCh38
rs386720483 Chr.8: 467998-468000 on Build GRCh38
rs773369441 Chr.8: 467827-467827 on Build GRCh38
rs758367840 Chr.8: 467772-467772 on Build GRCh38
rs190725845 Chr.8: 467832-467832 on Build GRCh38
rs935846814 Chr.8: 467801-467801 on Build GRCh38
rs1006915542 Chr.8: 468063-468063 on Build GRCh38
rs762304042 Chr.8: 467729-467729 on Build GRCh38
rs776615200 Chr.8: 467707-467707 on Build GRCh38
rs370120502 Chr.8: 467771-467771 on Build GRCh38
rs758008753 Chr.8: 467960-467960 on Build GRCh38
rs1033932106 Chr.8: 467739-467739 on Build GRCh38
rs200378033 Chr.8: 467715-467715 on Build GRCh38
rs771317602 Chr.8: 467780-467780 on Build GRCh38
rs879029484 Chr.8: 468094-468094 on Build GRCh38
rs960916453 Chr.8: 467740-467740 on Build GRCh38
rs767719986 Chr.8: 467865-467865 on Build GRCh38
rs531587898 Chr.8: 467844-467844 on Build GRCh38
rs989347780 Chr.8: 467950-467950 on Build GRCh38
rs950662045 Chr.8: 468092-468092 on Build GRCh38
rs61010875 Chr.8: 467705-467705 on Build GRCh38
rs566148545 Chr.8: 467814-467814 on Build GRCh38
rs138504314 Chr.8: 467903-467903 on Build GRCh38
rs1055272812 Chr.8: 467864-467864 on Build GRCh38
rs199572421 Chr.8: 467728-467728 on Build GRCh38
rs953221401 Chr.8: 468102-468102 on Build GRCh38
rs759854400 Chr.8: 467770-467770 on Build GRCh38
rs954099295 Chr.8: 467921-467921 on Build GRCh38
rs1013879968 Chr.8: 468038-468038 on Build GRCh38
rs368087109 Chr.8: 467671-467671 on Build GRCh38
rs376812883 Chr.8: 467998-467998 on Build GRCh38
rs551393722 Chr.8: 467884-467884 on Build GRCh38
rs957894152 Chr.8: 467936-467936 on Build GRCh38
rs753703281 Chr.8: 468011-468011 on Build GRCh38
rs766738888 Chr.8: 467670-467671 on Build GRCh38
rs765053140 Chr.8: 467925-467925 on Build GRCh38
rs183654675 Chr.8: 467779-467779 on Build GRCh38
rs887482355 Chr.8: 467841-467841 on Build GRCh38
rs921719407 Chr.8: 467798-467798 on Build GRCh38
rs754235713 Chr.8: 467698-467701 on Build GRCh38
rs779050056 Chr.8: 467682-467682 on Build GRCh38
rs186764647 Chr.8: 467830-467830 on Build GRCh38
rs539232655 Chr.8: 467663-467663 on Build GRCh38
rs147333165 Chr.8: 467992-467992 on Build GRCh38
rs555632434 Chr.8: 467947-467947 on Build GRCh38
rs796848627 Chr.8: 467835-467836 on Build GRCh38
rs59258529 Chr.8: 467998-467998 on Build GRCh38
rs575266302 Chr.8: 467769-467769 on Build GRCh38
rs954151799 Chr.8: 468058-468058 on Build GRCh38
rs541880905 Chr.8: 468026-468026 on Build GRCh38
rs978307802 Chr.8: 467776-467776 on Build GRCh38
rs929080239 Chr.8: 467838-467838 on Build GRCh38
rs752362497 Chr.8: 467956-467956 on Build GRCh38
rs367797583 Chr.8: 467981-467981 on Build GRCh38
rs559992461 Chr.8: 468089-468089 on Build GRCh38
rs143976843 Chr.8: 467940-467940 on Build GRCh38
rs372196730 Chr.8: 467785-467785 on Build GRCh38
rs555134641 Chr.8: 467924-467924 on Build GRCh38
rs34788617 Chr.8: 468044-468045 on Build GRCh38
rs921647367 Chr.8: 467955-467955 on Build GRCh38
rs775511222 Chr.8: 467724-467724 on Build GRCh38
rs778540733 Chr.8: 468082-468082 on Build GRCh38
rs765331447 Chr.8: 467716-467716 on Build GRCh38
rs987105206 Chr.8: 467958-467958 on Build GRCh38
rs1039987927 Chr.8: 467653-467653 on Build GRCh38

View Full Product Details