Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 468924-469422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO25
Entrez Gene ID
26260
Gene Name
F-box protein 25
Gene Aliases
FBX25
UniGene
Hs.438454
Gene Chromosome Location
Chr.8: 406808-477967 on Build GRCh38
Transcripts
NM_012173.3 NM_183420.1 NM_183421.1 XM_011534748.2 XM_011534749.1 XM_017013313.1 XM_017013314.1

SNP Detail

SNP ID Chromosome Location
rs927805761 Chr.8: 469091-469091 on Build GRCh38
rs748507121 Chr.8: 469171-469171 on Build GRCh38
rs3779746 Chr.8: 469066-469066 on Build GRCh38
rs1047692172 Chr.8: 469384-469384 on Build GRCh38
rs1031766309 Chr.8: 469396-469396 on Build GRCh38
rs547328409 Chr.8: 469039-469039 on Build GRCh38
rs887278663 Chr.8: 469151-469151 on Build GRCh38
rs114751187 Chr.8: 469392-469392 on Build GRCh38
rs1023604778 Chr.8: 469209-469209 on Build GRCh38
rs1013160423 Chr.8: 469378-469378 on Build GRCh38
rs899610733 Chr.8: 469143-469143 on Build GRCh38
rs184536965 Chr.8: 469377-469377 on Build GRCh38
rs371579221 Chr.8: 469352-469352 on Build GRCh38
rs1020083512 Chr.8: 468950-468950 on Build GRCh38
rs893687823 Chr.8: 469125-469125 on Build GRCh38
rs111570050 Chr.8: 469067-469068 on Build GRCh38
rs747838849 Chr.8: 469056-469057 on Build GRCh38
rs901323999 Chr.8: 469182-469182 on Build GRCh38
rs888531239 Chr.8: 469197-469197 on Build GRCh38
rs1053307501 Chr.8: 469055-469055 on Build GRCh38
rs941435511 Chr.8: 469390-469390 on Build GRCh38
rs1039368856 Chr.8: 469306-469306 on Build GRCh38
rs981540286 Chr.8: 469214-469214 on Build GRCh38
rs927827967 Chr.8: 469215-469215 on Build GRCh38
rs952705870 Chr.8: 469075-469075 on Build GRCh38
rs994553944 Chr.8: 468963-468963 on Build GRCh38
rs117758085 Chr.8: 469362-469362 on Build GRCh38
rs142351881 Chr.8: 469066-469067 on Build GRCh38
rs752858133 Chr.8: 469162-469163 on Build GRCh38
rs547801004 Chr.8: 469260-469260 on Build GRCh38
rs148904975 Chr.8: 469115-469115 on Build GRCh38
rs552157978 Chr.8: 469060-469060 on Build GRCh38
rs572144210 Chr.8: 469358-469358 on Build GRCh38
rs979905343 Chr.8: 469031-469031 on Build GRCh38
rs577440235 Chr.8: 468957-468957 on Build GRCh38
rs3735920 Chr.8: 469391-469391 on Build GRCh38
rs933772885 Chr.8: 469351-469351 on Build GRCh38
rs575243688 Chr.8: 469174-469174 on Build GRCh38
rs1007024544 Chr.8: 469063-469063 on Build GRCh38
rs942386581 Chr.8: 469357-469357 on Build GRCh38
rs994775734 Chr.8: 469186-469186 on Build GRCh38
rs888500569 Chr.8: 469386-469386 on Build GRCh38
rs8049 Chr.8: 468993-468993 on Build GRCh38
rs1047106292 Chr.8: 469195-469195 on Build GRCh38
rs35458213 Chr.8: 469055-469056 on Build GRCh38
rs928473962 Chr.8: 469124-469124 on Build GRCh38
rs950087501 Chr.8: 469074-469074 on Build GRCh38
rs771428068 Chr.8: 469011-469011 on Build GRCh38
rs73173390 Chr.8: 469363-469363 on Build GRCh38
rs1022951902 Chr.8: 469207-469207 on Build GRCh38
rs1025658489 Chr.8: 468975-468975 on Build GRCh38
rs867864009 Chr.8: 469208-469208 on Build GRCh38
rs548993393 Chr.8: 469020-469020 on Build GRCh38
rs531871915 Chr.8: 469132-469132 on Build GRCh38
rs984669394 Chr.8: 468978-468978 on Build GRCh38
rs997927858 Chr.8: 469147-469147 on Build GRCh38
rs757557444 Chr.8: 469057-469058 on Build GRCh38
rs553286404 Chr.8: 468947-468947 on Build GRCh38
rs559800713 Chr.8: 469339-469339 on Build GRCh38
rs537419001 Chr.8: 469176-469176 on Build GRCh38
rs143577289 Chr.8: 469131-469131 on Build GRCh38
rs1041237300 Chr.8: 469064-469064 on Build GRCh38
rs938457880 Chr.8: 469053-469053 on Build GRCh38
rs1023004397 Chr.8: 469387-469387 on Build GRCh38
rs1029808704 Chr.8: 469044-469044 on Build GRCh38
rs566103935 Chr.8: 469142-469142 on Build GRCh38
rs563754157 Chr.8: 468951-468951 on Build GRCh38
rs376747992 Chr.8: 469121-469121 on Build GRCh38
rs554951756 Chr.8: 469156-469156 on Build GRCh38
rs1010652762 Chr.8: 468970-468970 on Build GRCh38
rs145546328 Chr.8: 468977-468977 on Build GRCh38
rs112205005 Chr.8: 469200-469200 on Build GRCh38
rs564896072 Chr.8: 468991-468991 on Build GRCh38
rs79240478 Chr.8: 469068-469068 on Build GRCh38
rs189579155 Chr.8: 469052-469052 on Build GRCh38
rs982376555 Chr.8: 469077-469077 on Build GRCh38
rs909166369 Chr.8: 468992-468992 on Build GRCh38
rs912877871 Chr.8: 469056-469059 on Build GRCh38
rs551591881 Chr.8: 469291-469291 on Build GRCh38
rs959892521 Chr.8: 469243-469243 on Build GRCh38
rs538320583 Chr.8: 469117-469117 on Build GRCh38
rs758716727 Chr.8: 469296-469296 on Build GRCh38
rs1016094112 Chr.8: 469090-469090 on Build GRCh38
rs142773400 Chr.8: 469065-469066 on Build GRCh38
rs546278439 Chr.8: 469203-469203 on Build GRCh38
rs1050909 Chr.8: 469368-469368 on Build GRCh38
rs935280802 Chr.8: 469108-469108 on Build GRCh38
rs1036054430 Chr.8: 469233-469233 on Build GRCh38
rs988347827 Chr.8: 469274-469274 on Build GRCh38
rs535227020 Chr.8: 469148-469148 on Build GRCh38
rs1005515387 Chr.8: 469081-469081 on Build GRCh38
rs956570888 Chr.8: 469270-469270 on Build GRCh38
rs1017389018 Chr.8: 469057-469057 on Build GRCh38
rs1017193453 Chr.8: 469159-469159 on Build GRCh38
rs141533847 Chr.8: 469201-469201 on Build GRCh38
rs530424217 Chr.8: 469056-469056 on Build GRCh38
rs1039316377 Chr.8: 469140-469140 on Build GRCh38

View Full Product Details