Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161171963-161172461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT3
Entrez Gene ID
8703
Gene Name
beta-1,4-galactosyltransferase 3
Gene Aliases
beta4Gal-T3
UniGene
Hs.321231
Gene Chromosome Location
Chr.1: 161171310-161177968 on Build GRCh38
Transcripts
NM_001199873.1 NM_001199874.1 NM_003779.3 XM_005245566.1 XM_011510093.1 XM_017002714.1 XM_005245291.3 XM_005245295.3 XM_006711402.2 XM_006711403.2 XM_006711406.2 XM_011509663.2 XM_011509664.1 XM_011509665.2 XM_011509666.2 XM_011509667.2 XM_011509668.2 XM_011509669.1 XM_011509670.2 XM_011509673.2 XM_011509674.2 XM_011509676.1 XM_011509677.1 XM_011509678.1 XM_011509679.1 XM_011509681.1 XM_011509682.1 XM_017001561.1 XM_017001562.1 XM_017001563.1 XM_017001564.1 XM_017001570.1 XM_017001571.1

Gene Symbol
PPOX
Entrez Gene ID
5498
Gene Name
protoporphyrinogen oxidase
Gene Aliases
PPO, V290M, VP
UniGene
Hs.517373
Gene Chromosome Location
Chr.1: 161165728-161178277 on Build GRCh38
Transcripts
NM_001199873.1 NM_001199874.1 NM_003779.3 XM_005245566.1 XM_011510093.1 XM_017002714.1 XM_005245291.3 XM_005245295.3 XM_006711402.2 XM_006711403.2 XM_006711406.2 XM_011509663.2 XM_011509664.1 XM_011509665.2 XM_011509666.2 XM_011509667.2 XM_011509668.2 XM_011509669.1 XM_011509670.2 XM_011509673.2 XM_011509674.2 XM_011509676.1 XM_011509677.1 XM_011509678.1 XM_011509679.1 XM_011509681.1 XM_011509682.1 XM_017001561.1 XM_017001562.1 XM_017001563.1 XM_017001564.1 XM_017001570.1 XM_017001571.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5727903 c.859A>G p.K287E Chr.1: 161172276-161172276 on Build GRCh38
6567149 c.892G>A p.E298K Chr.1: 161172243-161172243 on Build GRCh38
4922350 c.946C>A p.Q316K Chr.1: 161172052-161172052 on Build GRCh38
4024333 c.969A>G p.T323T Chr.1: 161172029-161172029 on Build GRCh38
6302730 c.940T>G p.W314G Chr.1: 161172058-161172058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780404324 Chr.1: 161172072-161172072 on Build GRCh38
rs370188042 Chr.1: 161172125-161172125 on Build GRCh38
rs747156753 Chr.1: 161172076-161172076 on Build GRCh38
rs201732378 Chr.1: 161172326-161172326 on Build GRCh38
rs755467809 Chr.1: 161172371-161172371 on Build GRCh38
rs1009090346 Chr.1: 161172163-161172163 on Build GRCh38
rs766980374 Chr.1: 161172222-161172222 on Build GRCh38
rs1000032128 Chr.1: 161172242-161172242 on Build GRCh38
rs765636658 Chr.1: 161172109-161172109 on Build GRCh38
rs749135032 Chr.1: 161172281-161172281 on Build GRCh38
rs775022821 Chr.1: 161172030-161172030 on Build GRCh38
rs775005640 Chr.1: 161172186-161172186 on Build GRCh38
rs200759995 Chr.1: 161172345-161172345 on Build GRCh38
rs767239787 Chr.1: 161172033-161172033 on Build GRCh38
rs752000957 Chr.1: 161172229-161172229 on Build GRCh38
rs762438234 Chr.1: 161172025-161172025 on Build GRCh38
rs774403075 Chr.1: 161172294-161172294 on Build GRCh38
rs1032811066 Chr.1: 161172256-161172256 on Build GRCh38
rs750835537 Chr.1: 161172203-161172203 on Build GRCh38
rs772007951 Chr.1: 161172324-161172324 on Build GRCh38
rs567814897 Chr.1: 161172131-161172131 on Build GRCh38
rs765233285 Chr.1: 161172099-161172115 on Build GRCh38
rs908645185 Chr.1: 161172418-161172418 on Build GRCh38
rs748196045 Chr.1: 161172253-161172253 on Build GRCh38
rs190810180 Chr.1: 161172053-161172053 on Build GRCh38
rs150162843 Chr.1: 161171996-161171996 on Build GRCh38
rs781133882 Chr.1: 161172252-161172252 on Build GRCh38
rs373052685 Chr.1: 161172325-161172325 on Build GRCh38
rs377190536 Chr.1: 161172181-161172181 on Build GRCh38
rs146691911 Chr.1: 161172129-161172129 on Build GRCh38
rs890108384 Chr.1: 161172143-161172143 on Build GRCh38
rs781536903 Chr.1: 161172100-161172100 on Build GRCh38
rs770627750 Chr.1: 161172291-161172291 on Build GRCh38
rs530341614 Chr.1: 161172260-161172260 on Build GRCh38
rs751099833 Chr.1: 161172013-161172013 on Build GRCh38
rs751887590 Chr.1: 161172370-161172370 on Build GRCh38
rs368847387 Chr.1: 161172266-161172266 on Build GRCh38
rs1017682635 Chr.1: 161172282-161172282 on Build GRCh38
rs770071099 Chr.1: 161172108-161172108 on Build GRCh38
rs1057311925 Chr.1: 161172010-161172010 on Build GRCh38
rs761907963 Chr.1: 161172091-161172092 on Build GRCh38
rs763689228 Chr.1: 161172191-161172191 on Build GRCh38
rs758732350 Chr.1: 161172215-161172215 on Build GRCh38
rs773170451 Chr.1: 161172111-161172111 on Build GRCh38
rs941553438 Chr.1: 161172038-161172038 on Build GRCh38
rs754441286 Chr.1: 161172265-161172265 on Build GRCh38
rs768017177 Chr.1: 161172379-161172379 on Build GRCh38
rs958645421 Chr.1: 161172261-161172261 on Build GRCh38
rs752286852 Chr.1: 161172034-161172034 on Build GRCh38
rs964727068 Chr.1: 161172288-161172288 on Build GRCh38
rs763776973 Chr.1: 161172338-161172338 on Build GRCh38
rs1024750925 Chr.1: 161172300-161172300 on Build GRCh38
rs372601868 Chr.1: 161172054-161172054 on Build GRCh38
rs776265561 Chr.1: 161172354-161172354 on Build GRCh38
rs766921603 Chr.1: 161172182-161172182 on Build GRCh38
rs375516892 Chr.1: 161172099-161172099 on Build GRCh38
rs755458341 Chr.1: 161172235-161172235 on Build GRCh38
rs745804708 Chr.1: 161172295-161172295 on Build GRCh38
rs374576177 Chr.1: 161172347-161172347 on Build GRCh38
rs376425088 Chr.1: 161172369-161172369 on Build GRCh38
rs199883927 Chr.1: 161172365-161172365 on Build GRCh38
rs748398250 Chr.1: 161172106-161172106 on Build GRCh38

View Full Product Details