Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161367750-161368187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP126
Entrez Gene ID
257177
Gene Name
cilia and flagella associated protein 126
Gene Aliases
C1orf192, Flattop, Fltp
UniGene
Hs.534593
Gene Chromosome Location
Chr.1: 161364731-161367883 on Build GRCh38
Transcripts
NM_001013625.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667358 c.7A>G p.T3A Chr.1: 161367862-161367862 on Build GRCh38
5844277 c.5C>T p.A2V Chr.1: 161367864-161367864 on Build GRCh38
4748062 c.15_16delCA p.Y5fs*1 Chr.1: 161367853-161367854 on Build GRCh38
7257183 c.18T>C p.S6S Chr.1: 161367851-161367851 on Build GRCh38
5809278 c.4G>T p.A2S Chr.1: 161367865-161367865 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368322828 Chr.1: 161367789-161367789 on Build GRCh38
rs34234733 Chr.1: 161367924-161367925 on Build GRCh38
rs762117666 Chr.1: 161367894-161367894 on Build GRCh38
rs765533793 Chr.1: 161367911-161367911 on Build GRCh38
rs41270861 Chr.1: 161367814-161367814 on Build GRCh38
rs770662304 Chr.1: 161367839-161367840 on Build GRCh38
rs754955202 Chr.1: 161367813-161367813 on Build GRCh38
rs571379171 Chr.1: 161368083-161368083 on Build GRCh38
rs1038049683 Chr.1: 161368008-161368008 on Build GRCh38
rs959467093 Chr.1: 161368126-161368126 on Build GRCh38
rs191500294 Chr.1: 161367915-161367915 on Build GRCh38
rs777505329 Chr.1: 161367830-161367831 on Build GRCh38
rs1020889992 Chr.1: 161368085-161368085 on Build GRCh38
rs751333787 Chr.1: 161368130-161368130 on Build GRCh38
rs982430508 Chr.1: 161368035-161368035 on Build GRCh38
rs891033724 Chr.1: 161368071-161368071 on Build GRCh38
rs761347061 Chr.1: 161368158-161368158 on Build GRCh38
rs775473901 Chr.1: 161367886-161367886 on Build GRCh38
rs751732066 Chr.1: 161367811-161367811 on Build GRCh38
rs146363043 Chr.1: 161367841-161367841 on Build GRCh38
rs903650745 Chr.1: 161368086-161368086 on Build GRCh38
rs763083457 Chr.1: 161367914-161367914 on Build GRCh38
rs183593674 Chr.1: 161368106-161368106 on Build GRCh38
rs759102455 Chr.1: 161367880-161367880 on Build GRCh38
rs762769993 Chr.1: 161367947-161367947 on Build GRCh38
rs750593361 Chr.1: 161367913-161367913 on Build GRCh38
rs760182392 Chr.1: 161368068-161368068 on Build GRCh38
rs915744501 Chr.1: 161368083-161368083 on Build GRCh38
rs1033756764 Chr.1: 161368125-161368125 on Build GRCh38
rs771467014 Chr.1: 161367938-161367938 on Build GRCh38
rs983733008 Chr.1: 161367966-161367966 on Build GRCh38
rs561201332 Chr.1: 161367925-161367925 on Build GRCh38
rs749111529 Chr.1: 161367838-161367838 on Build GRCh38
rs964598239 Chr.1: 161367935-161367935 on Build GRCh38
rs199688258 Chr.1: 161367798-161367798 on Build GRCh38
rs952341582 Chr.1: 161368032-161368032 on Build GRCh38
rs1000708596 Chr.1: 161368096-161368096 on Build GRCh38
rs563307424 Chr.1: 161367991-161367991 on Build GRCh38
rs950764879 Chr.1: 161368141-161368141 on Build GRCh38
rs1021237722 Chr.1: 161367817-161367817 on Build GRCh38
rs773727814 Chr.1: 161367856-161367856 on Build GRCh38
rs566373841 Chr.1: 161368041-161368041 on Build GRCh38
rs770405399 Chr.1: 161367852-161367852 on Build GRCh38
rs942204294 Chr.1: 161367992-161367992 on Build GRCh38
rs920177210 Chr.1: 161367997-161367997 on Build GRCh38
rs376533639 Chr.1: 161368136-161368136 on Build GRCh38
rs909475180 Chr.1: 161368164-161368164 on Build GRCh38
rs375119685 Chr.1: 161367889-161367889 on Build GRCh38
rs932319215 Chr.1: 161368052-161368052 on Build GRCh38
rs777431624 Chr.1: 161367838-161367838 on Build GRCh38
rs909360731 Chr.1: 161367968-161367968 on Build GRCh38
rs552210540 Chr.1: 161368112-161368112 on Build GRCh38
rs546365575 Chr.1: 161368033-161368033 on Build GRCh38
rs747925489 Chr.1: 161367833-161367833 on Build GRCh38
rs551453290 Chr.1: 161368038-161368038 on Build GRCh38
rs534604465 Chr.1: 161367867-161367867 on Build GRCh38
rs369766369 Chr.1: 161368056-161368056 on Build GRCh38
rs774855232 Chr.1: 161367945-161367945 on Build GRCh38
rs769738913 Chr.1: 161367836-161367836 on Build GRCh38
rs752209788 Chr.1: 161367821-161367821 on Build GRCh38
rs372324478 Chr.1: 161367932-161367932 on Build GRCh38

View Full Product Details