Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161595266-161595665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR2C
Entrez Gene ID
9103
Gene Name
Fc fragment of IgG receptor IIc (gene/pseudogene)
Gene Aliases
CD32, CD32C, CDW32, FCG2, FCRIIC, IGFR2
UniGene
Hs.656750
Gene Chromosome Location
Chr.1: 161581339-161601220 on Build GRCh38
Transcripts
NM_201563.5 NR_047648.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5844283 c.5C>G p.S2C Chr.1: 161595559-161595559 on Build GRCh38
1138742 c.28C>A p.Q10K Chr.1: 161595582-161595582 on Build GRCh38
3664958 c.80T>G p.F27C Chr.1: 161595634-161595634 on Build GRCh38
4142922 c.37A>G p.M13V Chr.1: 161595591-161595591 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs545302485 Chr.1: 161595602-161595602 on Build GRCh38
rs368294871 Chr.1: 161595625-161595625 on Build GRCh38
rs758142531 Chr.1: 161595620-161595620 on Build GRCh38
rs546911244 Chr.1: 161595424-161595424 on Build GRCh38
rs779783073 Chr.1: 161595613-161595613 on Build GRCh38
rs79316700 Chr.1: 161595524-161595524 on Build GRCh38
rs76277413 Chr.1: 161595591-161595591 on Build GRCh38
rs761511323 Chr.1: 161595607-161595607 on Build GRCh38
rs569432965 Chr.1: 161595493-161595493 on Build GRCh38
rs537528011 Chr.1: 161595540-161595540 on Build GRCh38
rs573882616 Chr.1: 161595481-161595481 on Build GRCh38
rs1026757504 Chr.1: 161595432-161595432 on Build GRCh38
rs778419926 Chr.1: 161595530-161595530 on Build GRCh38
rs536777647 Chr.1: 161595496-161595496 on Build GRCh38
rs757948234 Chr.1: 161595535-161595535 on Build GRCh38
rs565080407 Chr.1: 161595329-161595329 on Build GRCh38
rs114010589 Chr.1: 161595618-161595618 on Build GRCh38
rs183211628 Chr.1: 161595392-161595392 on Build GRCh38
rs748612961 Chr.1: 161595546-161595546 on Build GRCh38
rs200272379 Chr.1: 161595576-161595576 on Build GRCh38
rs952425402 Chr.1: 161595460-161595460 on Build GRCh38
rs61801822 Chr.1: 161595534-161595534 on Build GRCh38
rs148370482 Chr.1: 161595353-161595353 on Build GRCh38
rs753874517 Chr.1: 161595606-161595606 on Build GRCh38
rs778127956 Chr.1: 161595639-161595639 on Build GRCh38
rs777672721 Chr.1: 161595415-161595415 on Build GRCh38
rs760543419 Chr.1: 161595596-161595596 on Build GRCh38
rs1028317796 Chr.1: 161595386-161595386 on Build GRCh38
rs748818876 Chr.1: 161595363-161595364 on Build GRCh38
rs759805230 Chr.1: 161595568-161595568 on Build GRCh38
rs746889049 Chr.1: 161595447-161595447 on Build GRCh38
rs566778555 Chr.1: 161595439-161595439 on Build GRCh38
rs779667437 Chr.1: 161595624-161595624 on Build GRCh38
rs770455106 Chr.1: 161595547-161595547 on Build GRCh38
rs964082914 Chr.1: 161595408-161595408 on Build GRCh38
rs763914181 Chr.1: 161595521-161595521 on Build GRCh38
rs536228097 Chr.1: 161595550-161595550 on Build GRCh38
rs573076151 Chr.1: 161595304-161595304 on Build GRCh38
rs542487799 Chr.1: 161595494-161595494 on Build GRCh38
rs367710761 Chr.1: 161595562-161595562 on Build GRCh38
rs765118061 Chr.1: 161595608-161595608 on Build GRCh38
rs758396257 Chr.1: 161595336-161595336 on Build GRCh38
rs996521295 Chr.1: 161595413-161595413 on Build GRCh38
rs373596625 Chr.1: 161595514-161595514 on Build GRCh38
rs552538594 Chr.1: 161595492-161595492 on Build GRCh38
rs530003830 Chr.1: 161595414-161595414 on Build GRCh38
rs892733522 Chr.1: 161595307-161595307 on Build GRCh38
rs756586067 Chr.1: 161595631-161595631 on Build GRCh38
rs553320687 Chr.1: 161595564-161595564 on Build GRCh38
rs771179835 Chr.1: 161595640-161595640 on Build GRCh38
rs532579273 Chr.1: 161595467-161595467 on Build GRCh38
rs756068057 Chr.1: 161595503-161595503 on Build GRCh38
rs545181797 Chr.1: 161595315-161595315 on Build GRCh38
rs775578095 Chr.1: 161595593-161595593 on Build GRCh38
rs982625707 Chr.1: 161595478-161595478 on Build GRCh38
rs1019282501 Chr.1: 161595359-161595359 on Build GRCh38
rs561159402 Chr.1: 161595393-161595393 on Build GRCh38

View Full Product Details