Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161623706-161624208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR3B
Entrez Gene ID
2215
Gene Name
Fc fragment of IgG receptor IIIb
Gene Aliases
CD16, CD16b, FCG3, FCGR3, FCR-10, FCRIII, FCRIIIb
UniGene
Hs.372679
Gene Chromosome Location
Chr.1: 161623196-161631963 on Build GRCh38
Transcripts
NM_000570.4 NM_001244753.1 NM_001271035.1 NM_001271036.1 NM_001271037.1

SNP Detail

SNP ID Chromosome Location
rs998568553 Chr.1: 161623761-161623762 on Build GRCh38
rs965010055 Chr.1: 161623860-161623860 on Build GRCh38
rs192152686 Chr.1: 161624103-161624103 on Build GRCh38
rs575181423 Chr.1: 161624181-161624181 on Build GRCh38
rs536720275 Chr.1: 161624115-161624115 on Build GRCh38
rs552283660 Chr.1: 161624005-161624005 on Build GRCh38
rs779295860 Chr.1: 161623996-161623996 on Build GRCh38
rs544010053 Chr.1: 161623782-161623782 on Build GRCh38
rs376105479 Chr.1: 161623804-161623804 on Build GRCh38
rs994755339 Chr.1: 161623897-161623897 on Build GRCh38
rs960426353 Chr.1: 161624121-161624121 on Build GRCh38
rs67373090 Chr.1: 161624090-161624091 on Build GRCh38
rs916070566 Chr.1: 161624162-161624162 on Build GRCh38
rs1052814892 Chr.1: 161624094-161624094 on Build GRCh38
rs993435250 Chr.1: 161624015-161624015 on Build GRCh38
rs958131305 Chr.1: 161624064-161624064 on Build GRCh38
rs67167367 Chr.1: 161624168-161624168 on Build GRCh38
rs435985 Chr.1: 161624027-161624027 on Build GRCh38
rs553067383 Chr.1: 161624108-161624108 on Build GRCh38
rs934301836 Chr.1: 161624093-161624093 on Build GRCh38
rs998810338 Chr.1: 161623957-161623957 on Build GRCh38
rs72702127 Chr.1: 161624041-161624041 on Build GRCh38
rs370393406 Chr.1: 161623776-161623776 on Build GRCh38
rs61803005 Chr.1: 161623781-161623781 on Build GRCh38
rs1027022498 Chr.1: 161623902-161623902 on Build GRCh38
rs187307864 Chr.1: 161624029-161624029 on Build GRCh38
rs900326509 Chr.1: 161624109-161624109 on Build GRCh38
rs79636887 Chr.1: 161624090-161624090 on Build GRCh38
rs952709389 Chr.1: 161624117-161624117 on Build GRCh38
rs1008628103 Chr.1: 161624104-161624104 on Build GRCh38
rs952657453 Chr.1: 161623948-161623948 on Build GRCh38
rs982867554 Chr.1: 161623969-161623969 on Build GRCh38
rs908606308 Chr.1: 161623987-161623987 on Build GRCh38
rs1015585353 Chr.1: 161624118-161624118 on Build GRCh38
rs925622058 Chr.1: 161624088-161624089 on Build GRCh38
rs566833549 Chr.1: 161623878-161623878 on Build GRCh38
rs768179714 Chr.1: 161623805-161623805 on Build GRCh38
rs61803004 Chr.1: 161623761-161623761 on Build GRCh38
rs550980464 Chr.1: 161624032-161624032 on Build GRCh38
rs67411091 Chr.1: 161624172-161624172 on Build GRCh38
rs779368371 Chr.1: 161623787-161623787 on Build GRCh38
rs61803006 Chr.1: 161623928-161623928 on Build GRCh38
rs150375393 Chr.1: 161624184-161624184 on Build GRCh38
rs1031341331 Chr.1: 161623779-161623779 on Build GRCh38
rs530155808 Chr.1: 161623845-161623845 on Build GRCh38
rs530970633 Chr.1: 161624006-161624006 on Build GRCh38
rs144575913 Chr.1: 161624116-161624116 on Build GRCh38
rs555286707 Chr.1: 161624120-161624120 on Build GRCh38
rs904187776 Chr.1: 161623747-161623747 on Build GRCh38
rs1043253536 Chr.1: 161624068-161624068 on Build GRCh38
rs181609413 Chr.1: 161623846-161623846 on Build GRCh38
rs190021467 Chr.1: 161623802-161623802 on Build GRCh38
rs992925399 Chr.1: 161624145-161624145 on Build GRCh38

View Full Product Details