Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167117248-167117751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP27
Entrez Gene ID
92235
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
-
UniGene
Hs.632462
Gene Chromosome Location
Chr.1: 167094048-167129165 on Build GRCh38
Transcripts
NM_001080426.1 XM_005245603.3 XM_011510146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3376021 c.322A>G p.S108G Chr.1: 167117444-167117444 on Build GRCh38
7365670 c.355C>A p.Q119K Chr.1: 167117477-167117477 on Build GRCh38
676463 c.348G>C p.L116L Chr.1: 167117470-167117470 on Build GRCh38
676464 c.294C>T p.N98N Chr.1: 167117416-167117416 on Build GRCh38
6541183 c.404A>T p.D135V Chr.1: 167117526-167117526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928998973 Chr.1: 167117316-167117316 on Build GRCh38
rs919399224 Chr.1: 167117425-167117425 on Build GRCh38
rs767587590 Chr.1: 167117293-167117293 on Build GRCh38
rs753740372 Chr.1: 167117314-167117314 on Build GRCh38
rs766654484 Chr.1: 167117279-167117279 on Build GRCh38
rs771502951 Chr.1: 167117340-167117340 on Build GRCh38
rs1054660389 Chr.1: 167117730-167117730 on Build GRCh38
rs148991366 Chr.1: 167117484-167117484 on Build GRCh38
rs757110190 Chr.1: 167117324-167117324 on Build GRCh38
rs952201435 Chr.1: 167117622-167117622 on Build GRCh38
rs779575347 Chr.1: 167117341-167117341 on Build GRCh38
rs766205649 Chr.1: 167117578-167117578 on Build GRCh38
rs773962560 Chr.1: 167117580-167117580 on Build GRCh38
rs1003131358 Chr.1: 167117714-167117714 on Build GRCh38
rs558563058 Chr.1: 167117613-167117613 on Build GRCh38
rs530324682 Chr.1: 167117287-167117287 on Build GRCh38
rs965697852 Chr.1: 167117586-167117586 on Build GRCh38
rs146929565 Chr.1: 167117649-167117649 on Build GRCh38
rs780240980 Chr.1: 167117530-167117530 on Build GRCh38
rs571219118 Chr.1: 167117384-167117384 on Build GRCh38
rs770391450 Chr.1: 167117345-167117345 on Build GRCh38
rs890337509 Chr.1: 167117632-167117632 on Build GRCh38
rs145466273 Chr.1: 167117488-167117488 on Build GRCh38
rs767502047 Chr.1: 167117364-167117364 on Build GRCh38
rs758812932 Chr.1: 167117528-167117528 on Build GRCh38
rs149732757 Chr.1: 167117481-167117481 on Build GRCh38
rs146625016 Chr.1: 167117473-167117473 on Build GRCh38
rs147427450 Chr.1: 167117362-167117362 on Build GRCh38
rs963166513 Chr.1: 167117272-167117272 on Build GRCh38
rs985397750 Chr.1: 167117647-167117647 on Build GRCh38
rs776784375 Chr.1: 167117495-167117495 on Build GRCh38
rs751943869 Chr.1: 167117483-167117483 on Build GRCh38
rs376336322 Chr.1: 167117381-167117381 on Build GRCh38
rs150498762 Chr.1: 167117357-167117357 on Build GRCh38
rs890389836 Chr.1: 167117454-167117454 on Build GRCh38
rs1043565710 Chr.1: 167117379-167117379 on Build GRCh38
rs752610061 Chr.1: 167117295-167117295 on Build GRCh38
rs748280075 Chr.1: 167117558-167117558 on Build GRCh38
rs753849128 Chr.1: 167117447-167117447 on Build GRCh38
rs760845336 Chr.1: 167117439-167117439 on Build GRCh38
rs1020094307 Chr.1: 167117278-167117278 on Build GRCh38
rs139783903 Chr.1: 167117385-167117385 on Build GRCh38
rs758124971 Chr.1: 167117458-167117458 on Build GRCh38
rs995102513 Chr.1: 167117664-167117664 on Build GRCh38
rs763223361 Chr.1: 167117396-167117397 on Build GRCh38
rs767220703 Chr.1: 167117602-167117602 on Build GRCh38
rs934618960 Chr.1: 167117610-167117610 on Build GRCh38
rs773224803 Chr.1: 167117568-167117568 on Build GRCh38
rs547250427 Chr.1: 167117420-167117420 on Build GRCh38
rs778748208 Chr.1: 167117328-167117328 on Build GRCh38
rs201259394 Chr.1: 167117598-167117598 on Build GRCh38
rs760655250 Chr.1: 167117373-167117373 on Build GRCh38
rs935848594 Chr.1: 167117722-167117722 on Build GRCh38
rs768021668 Chr.1: 167117417-167117417 on Build GRCh38
rs777439948 Chr.1: 167117303-167117303 on Build GRCh38
rs778305194 Chr.1: 167117418-167117418 on Build GRCh38
rs140490170 Chr.1: 167117464-167117464 on Build GRCh38
rs746596317 Chr.1: 167117412-167117412 on Build GRCh38
rs76502020 Chr.1: 167117657-167117657 on Build GRCh38
rs186068292 Chr.1: 167117671-167117671 on Build GRCh38
rs1051773888 Chr.1: 167117627-167117627 on Build GRCh38
rs769973578 Chr.1: 167117561-167117561 on Build GRCh38
rs138760972 Chr.1: 167117522-167117522 on Build GRCh38
rs370416497 Chr.1: 167117542-167117542 on Build GRCh38
rs370198107 Chr.1: 167117509-167117509 on Build GRCh38
rs141168445 Chr.1: 167117621-167117621 on Build GRCh38
rs973440420 Chr.1: 167117298-167117298 on Build GRCh38
rs139638553 Chr.1: 167117356-167117356 on Build GRCh38
rs752761956 Chr.1: 167117370-167117370 on Build GRCh38
rs769867286 Chr.1: 167117441-167117441 on Build GRCh38
rs755103142 Chr.1: 167117291-167117291 on Build GRCh38
rs761799923 Chr.1: 167117448-167117448 on Build GRCh38
rs762763544 Chr.1: 167117576-167117576 on Build GRCh38
rs924707101 Chr.1: 167117584-167117584 on Build GRCh38
rs768886871 Chr.1: 167117490-167117490 on Build GRCh38
rs775566671 Chr.1: 167117438-167117438 on Build GRCh38
rs749750951 Chr.1: 167117419-167117419 on Build GRCh38
rs148733748 Chr.1: 167117433-167117433 on Build GRCh38
rs925391410 Chr.1: 167117718-167117718 on Build GRCh38
rs373710646 Chr.1: 167117396-167117396 on Build GRCh38
rs779561513 Chr.1: 167117411-167117411 on Build GRCh38
rs755613898 Chr.1: 167117516-167117516 on Build GRCh38
rs760681958 Chr.1: 167117508-167117508 on Build GRCh38
rs142938681 Chr.1: 167117353-167117353 on Build GRCh38
rs151287120 Chr.1: 167117455-167117455 on Build GRCh38
rs765335271 Chr.1: 167117499-167117499 on Build GRCh38
rs746335429 Chr.1: 167117527-167117527 on Build GRCh38
rs748618592 Chr.1: 167117344-167117344 on Build GRCh38
rs761973363 Chr.1: 167117498-167117498 on Build GRCh38
rs745492802 Chr.1: 167117330-167117330 on Build GRCh38
rs749802234 Chr.1: 167117478-167117478 on Build GRCh38
rs758012853 Chr.1: 167117410-167117410 on Build GRCh38
rs926814805 Chr.1: 167117648-167117648 on Build GRCh38
rs879246949 Chr.1: 167117553-167117553 on Build GRCh38
rs762216703 Chr.1: 167117518-167117518 on Build GRCh38
rs757706679 Chr.1: 167117480-167117480 on Build GRCh38
rs960290773 Chr.1: 167117485-167117485 on Build GRCh38
rs918927674 Chr.1: 167117305-167117305 on Build GRCh38
rs1039003898 Chr.1: 167117653-167117653 on Build GRCh38
rs750129646 Chr.1: 167117404-167117404 on Build GRCh38
rs1026045474 Chr.1: 167117611-167117611 on Build GRCh38
rs776989552 Chr.1: 167117555-167117555 on Build GRCh38
rs945382184 Chr.1: 167117696-167117696 on Build GRCh38
rs138492285 Chr.1: 167117349-167117349 on Build GRCh38
rs757165697 Chr.1: 167117386-167117386 on Build GRCh38
rs752259487 Chr.1: 167117515-167117515 on Build GRCh38
rs950535180 Chr.1: 167117710-167117710 on Build GRCh38
rs1026042604 Chr.1: 167117676-167117676 on Build GRCh38
rs368162787 Chr.1: 167117302-167117302 on Build GRCh38
rs371926436 Chr.1: 167117313-167117313 on Build GRCh38
rs772394225 Chr.1: 167117437-167117437 on Build GRCh38
rs371621251 Chr.1: 167117457-167117457 on Build GRCh38
rs767030589 Chr.1: 167117510-167117510 on Build GRCh38
rs766096668 Chr.1: 167117460-167117460 on Build GRCh38
rs747256963 Chr.1: 167117541-167117541 on Build GRCh38
rs992435682 Chr.1: 167117654-167117654 on Build GRCh38

View Full Product Details