Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167126841-167127340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP27
Entrez Gene ID
92235
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
-
UniGene
Hs.632462
Gene Chromosome Location
Chr.1: 167094048-167129165 on Build GRCh38
Transcripts
NM_001080426.1 XM_005245603.3 XM_011510146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4763403 c.1745C>G p.S582C Chr.1: 167126876-167126876 on Build GRCh38
7413368 c.2108C>A p.P703H Chr.1: 167127239-167127239 on Build GRCh38
3376823 c.1944G>A p.A648A Chr.1: 167127075-167127075 on Build GRCh38
3477743 c.2169C>T p.I723I Chr.1: 167127300-167127300 on Build GRCh38
3477741 c.1920C>T p.S640S Chr.1: 167127051-167127051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759664430 Chr.1: 167127169-167127171 on Build GRCh38
rs747125220 Chr.1: 167127074-167127074 on Build GRCh38
rs757597404 Chr.1: 167127055-167127055 on Build GRCh38
rs769487453 Chr.1: 167127087-167127087 on Build GRCh38
rs750689944 Chr.1: 167127065-167127065 on Build GRCh38
rs747943857 Chr.1: 167127029-167127029 on Build GRCh38
rs529604128 Chr.1: 167126936-167126936 on Build GRCh38
rs145537197 Chr.1: 167127064-167127064 on Build GRCh38
rs368739494 Chr.1: 167127314-167127314 on Build GRCh38
rs76205550 Chr.1: 167127158-167127158 on Build GRCh38
rs749647831 Chr.1: 167126865-167126865 on Build GRCh38
rs767627024 Chr.1: 167127218-167127218 on Build GRCh38
rs372973596 Chr.1: 167127156-167127156 on Build GRCh38
rs1035702827 Chr.1: 167127241-167127241 on Build GRCh38
rs750042465 Chr.1: 167126918-167126918 on Build GRCh38
rs771828185 Chr.1: 167127246-167127246 on Build GRCh38
rs756161976 Chr.1: 167127234-167127234 on Build GRCh38
rs370561353 Chr.1: 167127184-167127184 on Build GRCh38
rs906677033 Chr.1: 167126906-167126906 on Build GRCh38
rs753215878 Chr.1: 167126942-167126942 on Build GRCh38
rs781275928 Chr.1: 167127145-167127145 on Build GRCh38
rs767017134 Chr.1: 167127109-167127109 on Build GRCh38
rs762325833 Chr.1: 167127114-167127114 on Build GRCh38
rs766782601 Chr.1: 167127192-167127192 on Build GRCh38
rs781021697 Chr.1: 167127076-167127076 on Build GRCh38
rs768233594 Chr.1: 167127254-167127254 on Build GRCh38
rs1037207381 Chr.1: 167126874-167126874 on Build GRCh38
rs758806552 Chr.1: 167127138-167127138 on Build GRCh38
rs763434444 Chr.1: 167127191-167127191 on Build GRCh38
rs745641105 Chr.1: 167127242-167127242 on Build GRCh38
rs773965871 Chr.1: 167127105-167127105 on Build GRCh38
rs772856160 Chr.1: 167127031-167127031 on Build GRCh38
rs1003246008 Chr.1: 167127220-167127220 on Build GRCh38
rs759858923 Chr.1: 167127199-167127199 on Build GRCh38
rs761664135 Chr.1: 167126907-167126907 on Build GRCh38
rs755017675 Chr.1: 167127144-167127144 on Build GRCh38
rs760626149 Chr.1: 167126899-167126899 on Build GRCh38
rs201239266 Chr.1: 167127219-167127219 on Build GRCh38
rs775602117 Chr.1: 167126894-167126894 on Build GRCh38
rs144793075 Chr.1: 167126889-167126889 on Build GRCh38
rs753790234 Chr.1: 167127289-167127289 on Build GRCh38
rs779159793 Chr.1: 167126878-167126878 on Build GRCh38
rs778716331 Chr.1: 167127313-167127313 on Build GRCh38
rs752808434 Chr.1: 167127282-167127282 on Build GRCh38
rs778626028 Chr.1: 167127239-167127239 on Build GRCh38
rs756668951 Chr.1: 167126956-167126956 on Build GRCh38
rs764553686 Chr.1: 167127039-167127039 on Build GRCh38
rs760772326 Chr.1: 167127283-167127283 on Build GRCh38
rs956126023 Chr.1: 167126985-167126985 on Build GRCh38
rs764091553 Chr.1: 167127287-167127287 on Build GRCh38
rs753731247 Chr.1: 167127236-167127236 on Build GRCh38
rs141118809 Chr.1: 167127113-167127113 on Build GRCh38
rs771582809 Chr.1: 167127086-167127086 on Build GRCh38
rs369939829 Chr.1: 167126881-167126881 on Build GRCh38
rs773833411 Chr.1: 167127186-167127186 on Build GRCh38
rs745461356 Chr.1: 167127163-167127163 on Build GRCh38
rs532059690 Chr.1: 167127048-167127048 on Build GRCh38
rs896852713 Chr.1: 167126861-167126861 on Build GRCh38
rs764839180 Chr.1: 167126913-167126913 on Build GRCh38
rs751742989 Chr.1: 167127197-167127197 on Build GRCh38
rs143802055 Chr.1: 167127247-167127247 on Build GRCh38
rs545061292 Chr.1: 167126981-167126981 on Build GRCh38
rs773047471 Chr.1: 167127094-167127094 on Build GRCh38
rs1002949988 Chr.1: 167126959-167126959 on Build GRCh38
rs767826523 Chr.1: 167127274-167127274 on Build GRCh38
rs768511723 Chr.1: 167126900-167126900 on Build GRCh38
rs750886063 Chr.1: 167127118-167127118 on Build GRCh38
rs202017285 Chr.1: 167127038-167127038 on Build GRCh38
rs758106454 Chr.1: 167127316-167127316 on Build GRCh38
rs139295413 Chr.1: 167127317-167127317 on Build GRCh38
rs771500645 Chr.1: 167127166-167127166 on Build GRCh38
rs374488804 Chr.1: 167126984-167126984 on Build GRCh38
rs778336860 Chr.1: 167126970-167126970 on Build GRCh38
rs758753192 Chr.1: 167127071-167127071 on Build GRCh38
rs770607161 Chr.1: 167127104-167127104 on Build GRCh38
rs757387713 Chr.1: 167126872-167126872 on Build GRCh38
rs562950788 Chr.1: 167126920-167126920 on Build GRCh38
rs867643169 Chr.1: 167126974-167126974 on Build GRCh38
rs149689505 Chr.1: 167126931-167126931 on Build GRCh38
rs771122589 Chr.1: 167126877-167126877 on Build GRCh38
rs776349275 Chr.1: 167126901-167126901 on Build GRCh38
rs920927738 Chr.1: 167127058-167127058 on Build GRCh38
rs757616235 Chr.1: 167126982-167126982 on Build GRCh38
rs549179166 Chr.1: 167127032-167127032 on Build GRCh38
rs976666073 Chr.1: 167127014-167127014 on Build GRCh38
rs1011449034 Chr.1: 167126971-167126971 on Build GRCh38
rs747073864 Chr.1: 167127005-167127005 on Build GRCh38
rs772284726 Chr.1: 167126993-167126993 on Build GRCh38
rs374598924 Chr.1: 167127259-167127259 on Build GRCh38
rs376593703 Chr.1: 167127159-167127159 on Build GRCh38
rs375160987 Chr.1: 167127150-167127150 on Build GRCh38
rs772233606 Chr.1: 167126891-167126891 on Build GRCh38
rs369410112 Chr.1: 167127117-167127117 on Build GRCh38
rs1042877254 Chr.1: 167127231-167127231 on Build GRCh38
rs771548929 Chr.1: 167127263-167127263 on Build GRCh38
rs566588987 Chr.1: 167127165-167127165 on Build GRCh38
rs756472833 Chr.1: 167126863-167126863 on Build GRCh38
rs746051072 Chr.1: 167126992-167126992 on Build GRCh38
rs539069832 Chr.1: 167127147-167127147 on Build GRCh38
rs200893444 Chr.1: 167127141-167127141 on Build GRCh38
rs777654054 Chr.1: 167127155-167127155 on Build GRCh38
rs757245031 Chr.1: 167127295-167127295 on Build GRCh38
rs144006881 Chr.1: 167127100-167127100 on Build GRCh38
rs774846028 Chr.1: 167127268-167127268 on Build GRCh38
rs188895908 Chr.1: 167127004-167127004 on Build GRCh38
rs768544982 Chr.1: 167127020-167127020 on Build GRCh38
rs768387689 Chr.1: 167127075-167127075 on Build GRCh38
rs140347809 Chr.1: 167127142-167127142 on Build GRCh38
rs776600431 Chr.1: 167127022-167127022 on Build GRCh38
rs757053635 Chr.1: 167127238-167127238 on Build GRCh38
rs987900395 Chr.1: 167127028-167127028 on Build GRCh38
rs777705433 Chr.1: 167127235-167127235 on Build GRCh38
rs773888541 Chr.1: 167127257-167127257 on Build GRCh38
rs780422021 Chr.1: 167127073-167127073 on Build GRCh38
rs759787935 Chr.1: 167127272-167127272 on Build GRCh38
rs61103736 Chr.1: 167127056-167127056 on Build GRCh38
rs370847886 Chr.1: 167127185-167127185 on Build GRCh38
rs769590965 Chr.1: 167127030-167127030 on Build GRCh38
rs770986916 Chr.1: 167127265-167127265 on Build GRCh38
rs912261228 Chr.1: 167127129-167127129 on Build GRCh38

View Full Product Details