Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 74361823-74362327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP1
Entrez Gene ID
54332
Gene Name
ganglioside induced differentiation associated protein 1
Gene Aliases
CMT4, CMT4A, CMTRIA
UniGene
Hs.168950
Gene Chromosome Location
Chr.8: 74349251-74488867 on Build GRCh38
Transcripts
NM_001040875.2 NM_018972.2 NR_046346.1 XM_011517551.2 XM_011517552.2 XM_017013585.1 XM_017013586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
301183 c.572G>A p.R191Q Chr.8: 74361971-74361971 on Build GRCh38
7112038 c.502G>A p.E168K Chr.8: 74361901-74361901 on Build GRCh38
3786041 c.485-10T>A p.? Chr.8: 74361874-74361874 on Build GRCh38
1458217 c.490A>T p.I164F Chr.8: 74361889-74361889 on Build GRCh38
6260319 c.550G>T p.A184S Chr.8: 74361949-74361949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75294039 Chr.8: 74362018-74362018 on Build GRCh38
rs898015725 Chr.8: 74362294-74362294 on Build GRCh38
rs928148303 Chr.8: 74362296-74362296 on Build GRCh38
rs938416998 Chr.8: 74362052-74362052 on Build GRCh38
rs758326016 Chr.8: 74361894-74361894 on Build GRCh38
rs915439087 Chr.8: 74362007-74362007 on Build GRCh38
rs772634708 Chr.8: 74361849-74361849 on Build GRCh38
rs3832592 Chr.8: 74362097-74362098 on Build GRCh38
rs864622501 Chr.8: 74361979-74361979 on Build GRCh38
rs527810029 Chr.8: 74362168-74362168 on Build GRCh38
rs948357639 Chr.8: 74362089-74362089 on Build GRCh38
rs773723198 Chr.8: 74362048-74362048 on Build GRCh38
rs551975044 Chr.8: 74362083-74362083 on Build GRCh38
rs1050824692 Chr.8: 74362189-74362189 on Build GRCh38
rs924393289 Chr.8: 74362038-74362038 on Build GRCh38
rs924476816 Chr.8: 74362040-74362040 on Build GRCh38
rs964744192 Chr.8: 74362157-74362157 on Build GRCh38
rs761421699 Chr.8: 74361999-74361999 on Build GRCh38
rs778079435 Chr.8: 74361912-74361912 on Build GRCh38
rs1056785757 Chr.8: 74362082-74362082 on Build GRCh38
rs575531593 Chr.8: 74362269-74362269 on Build GRCh38
rs993357236 Chr.8: 74362099-74362099 on Build GRCh38
rs1008997357 Chr.8: 74362134-74362134 on Build GRCh38
rs104894077 Chr.8: 74361886-74361886 on Build GRCh38
rs146014328 Chr.8: 74362002-74362002 on Build GRCh38
rs990490728 Chr.8: 74362130-74362130 on Build GRCh38
rs989884398 Chr.8: 74362191-74362191 on Build GRCh38
rs776391567 Chr.8: 74361992-74361992 on Build GRCh38
rs746812771 Chr.8: 74362153-74362153 on Build GRCh38
rs774281600 Chr.8: 74362223-74362223 on Build GRCh38
rs758593282 Chr.8: 74362082-74362083 on Build GRCh38
rs549336887 Chr.8: 74362235-74362235 on Build GRCh38
rs376427753 Chr.8: 74362017-74362017 on Build GRCh38
rs890268724 Chr.8: 74362107-74362107 on Build GRCh38
rs761332159 Chr.8: 74361881-74361881 on Build GRCh38
rs749501057 Chr.8: 74361916-74361916 on Build GRCh38
rs527426824 Chr.8: 74362036-74362036 on Build GRCh38
rs771176873 Chr.8: 74361928-74361928 on Build GRCh38
rs1026131693 Chr.8: 74362102-74362102 on Build GRCh38
rs957266807 Chr.8: 74362098-74362098 on Build GRCh38
rs373277845 Chr.8: 74361890-74361890 on Build GRCh38
rs570458493 Chr.8: 74362166-74362166 on Build GRCh38
rs764793211 Chr.8: 74362000-74362000 on Build GRCh38
rs779894269 Chr.8: 74361943-74361943 on Build GRCh38
rs753388747 Chr.8: 74361879-74361879 on Build GRCh38
rs763802909 Chr.8: 74361878-74361878 on Build GRCh38
rs771534068 Chr.8: 74362161-74362161 on Build GRCh38
rs779132911 Chr.8: 74361942-74361942 on Build GRCh38
rs766295477 Chr.8: 74362096-74362097 on Build GRCh38
rs11554166 Chr.8: 74361906-74361906 on Build GRCh38
rs1036517376 Chr.8: 74362273-74362273 on Build GRCh38
rs890592656 Chr.8: 74361867-74361867 on Build GRCh38
rs1012971469 Chr.8: 74362010-74362010 on Build GRCh38
rs772722746 Chr.8: 74361971-74361971 on Build GRCh38
rs978258090 Chr.8: 74362293-74362293 on Build GRCh38
rs768849850 Chr.8: 74361987-74361987 on Build GRCh38
rs537418707 Chr.8: 74362231-74362231 on Build GRCh38
rs770658701 Chr.8: 74361956-74361956 on Build GRCh38
rs886041386 Chr.8: 74361900-74361900 on Build GRCh38
rs760895192 Chr.8: 74361869-74361869 on Build GRCh38
rs1037629250 Chr.8: 74362093-74362094 on Build GRCh38
rs73345392 Chr.8: 74361864-74361864 on Build GRCh38
rs144199299 Chr.8: 74361973-74361973 on Build GRCh38
rs148508128 Chr.8: 74361955-74361955 on Build GRCh38
rs965041280 Chr.8: 74361968-74361968 on Build GRCh38
rs779770529 Chr.8: 74361905-74361905 on Build GRCh38
rs995060932 Chr.8: 74362307-74362307 on Build GRCh38
rs995113212 Chr.8: 74361950-74361950 on Build GRCh38
rs746438323 Chr.8: 74361951-74361951 on Build GRCh38
rs761128477 Chr.8: 74361984-74361984 on Build GRCh38

View Full Product Details