Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 74364300-74364799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP1
Entrez Gene ID
54332
Gene Name
ganglioside induced differentiation associated protein 1
Gene Aliases
CMT4, CMT4A, CMTRIA
UniGene
Hs.168950
Gene Chromosome Location
Chr.8: 74349251-74488867 on Build GRCh38
Transcripts
NM_001040875.2 NM_018972.2 NR_046346.1 XM_011517551.2 XM_011517552.2 XM_017013585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5037082 c.1037G>T p.S346I Chr.8: 74364327-74364327 on Build GRCh38
5979543 c.1062A>T p.R354S Chr.8: 74364352-74364352 on Build GRCh38
4458155 c.1074C>G p.F358L Chr.8: 74364364-74364364 on Build GRCh38
1458219 c.1032T>C p.L344L Chr.8: 74364322-74364322 on Build GRCh38
5455689 c.1060A>G p.R354G Chr.8: 74364350-74364350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969634329 Chr.8: 74364623-74364623 on Build GRCh38
rs764207679 Chr.8: 74364521-74364521 on Build GRCh38
rs939300895 Chr.8: 74364694-74364694 on Build GRCh38
rs1135715 Chr.8: 74364724-74364724 on Build GRCh38
rs775647278 Chr.8: 74364507-74364507 on Build GRCh38
rs892103161 Chr.8: 74364663-74364663 on Build GRCh38
rs559133670 Chr.8: 74364664-74364664 on Build GRCh38
rs765609133 Chr.8: 74364356-74364356 on Build GRCh38
rs200836590 Chr.8: 74364386-74364386 on Build GRCh38
rs762813366 Chr.8: 74364474-74364474 on Build GRCh38
rs541078199 Chr.8: 74364647-74364647 on Build GRCh38
rs754305814 Chr.8: 74364778-74364789 on Build GRCh38
rs368943246 Chr.8: 74364335-74364335 on Build GRCh38
rs1007465964 Chr.8: 74364759-74364759 on Build GRCh38
rs754319162 Chr.8: 74364349-74364349 on Build GRCh38
rs1007110652 Chr.8: 74364434-74364434 on Build GRCh38
rs756089950 Chr.8: 74364761-74364761 on Build GRCh38
rs539004055 Chr.8: 74364519-74364519 on Build GRCh38
rs775559344 Chr.8: 74364332-74364332 on Build GRCh38
rs946460857 Chr.8: 74364728-74364728 on Build GRCh38
rs190777043 Chr.8: 74364414-74364414 on Build GRCh38
rs903612555 Chr.8: 74364648-74364648 on Build GRCh38
rs766677581 Chr.8: 74364509-74364510 on Build GRCh38
rs1002438138 Chr.8: 74364714-74364714 on Build GRCh38
rs982807152 Chr.8: 74364732-74364732 on Build GRCh38
rs965721851 Chr.8: 74364749-74364749 on Build GRCh38
rs909686879 Chr.8: 74364561-74364561 on Build GRCh38
rs1025272910 Chr.8: 74364677-74364677 on Build GRCh38
rs147945220 Chr.8: 74364385-74364385 on Build GRCh38
rs762380919 Chr.8: 74364525-74364525 on Build GRCh38
rs1052033443 Chr.8: 74364698-74364698 on Build GRCh38
rs145548839 Chr.8: 74364624-74364624 on Build GRCh38
rs182598397 Chr.8: 74364489-74364489 on Build GRCh38
rs1052038409 Chr.8: 74364661-74364661 on Build GRCh38
rs1043876709 Chr.8: 74364731-74364731 on Build GRCh38
rs765918987 Chr.8: 74364651-74364651 on Build GRCh38
rs773046770 Chr.8: 74364486-74364486 on Build GRCh38
rs757607617 Chr.8: 74364352-74364352 on Build GRCh38
rs112786851 Chr.8: 74364329-74364329 on Build GRCh38
rs764613529 Chr.8: 74364340-74364340 on Build GRCh38
rs375005505 Chr.8: 74364395-74364395 on Build GRCh38
rs777048240 Chr.8: 74364522-74364522 on Build GRCh38
rs772284929 Chr.8: 74364497-74364497 on Build GRCh38
rs575461751 Chr.8: 74364533-74364533 on Build GRCh38
rs766161487 Chr.8: 74364576-74364576 on Build GRCh38
rs554709374 Chr.8: 74364621-74364621 on Build GRCh38
rs73345399 Chr.8: 74364428-74364428 on Build GRCh38
rs59242435 Chr.8: 74364529-74364529 on Build GRCh38
rs542835607 Chr.8: 74364589-74364589 on Build GRCh38
rs748096043 Chr.8: 74364429-74364429 on Build GRCh38
rs1027109078 Chr.8: 74364720-74364720 on Build GRCh38
rs748093295 Chr.8: 74364742-74364742 on Build GRCh38
rs965656378 Chr.8: 74364510-74364510 on Build GRCh38
rs532934290 Chr.8: 74364684-74364684 on Build GRCh38
rs984629672 Chr.8: 74364557-74364557 on Build GRCh38
rs980670692 Chr.8: 74364637-74364637 on Build GRCh38
rs750114170 Chr.8: 74364371-74364371 on Build GRCh38
rs758771523 Chr.8: 74364563-74364563 on Build GRCh38
rs1039927839 Chr.8: 74364778-74364778 on Build GRCh38

View Full Product Details